Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 84622484-84622977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN2
Entrez Gene ID
54993
Gene Name
zinc finger and SCAN domain containing 2
Gene Aliases
ZFP29, ZNF854
UniGene
Hs.594023
Gene Chromosome Location
Chr.15: 84600986-84623716 on Build GRCh38
Transcripts
NM_181877.3

SNP Detail

SNP ID Chromosome Location
rs142208872 Chr.15: 84622667-84622667 on Build GRCh38
rs920252048 Chr.15: 84622951-84622968 on Build GRCh38
rs747968928 Chr.15: 84622689-84622696 on Build GRCh38
rs765357706 Chr.15: 84622720-84622720 on Build GRCh38
rs1018133162 Chr.15: 84622893-84622893 on Build GRCh38
rs759656340 Chr.15: 84622718-84622718 on Build GRCh38
rs899412812 Chr.15: 84622951-84622951 on Build GRCh38
rs761510434 Chr.15: 84622641-84622641 on Build GRCh38
rs759789153 Chr.15: 84622832-84622832 on Build GRCh38
rs763363710 Chr.15: 84622918-84622918 on Build GRCh38
rs896923318 Chr.15: 84622822-84622822 on Build GRCh38
rs766825948 Chr.15: 84622697-84622697 on Build GRCh38
rs1034553811 Chr.15: 84622603-84622604 on Build GRCh38
rs765432787 Chr.15: 84622834-84622834 on Build GRCh38
rs138979916 Chr.15: 84622608-84622608 on Build GRCh38
rs773556404 Chr.15: 84622668-84622668 on Build GRCh38
rs1017597399 Chr.15: 84622523-84622523 on Build GRCh38
rs34175071 Chr.15: 84622833-84622834 on Build GRCh38
rs568775004 Chr.15: 84622799-84622799 on Build GRCh38
rs890950076 Chr.15: 84622854-84622854 on Build GRCh38
rs950283247 Chr.15: 84622746-84622746 on Build GRCh38
rs770978937 Chr.15: 84622704-84622704 on Build GRCh38
rs761199202 Chr.15: 84622671-84622671 on Build GRCh38
rs1015094330 Chr.15: 84622507-84622507 on Build GRCh38
rs940993900 Chr.15: 84622781-84622781 on Build GRCh38
rs117370461 Chr.15: 84622752-84622752 on Build GRCh38
rs774337892 Chr.15: 84622510-84622510 on Build GRCh38
rs552339503 Chr.15: 84622705-84622705 on Build GRCh38
rs565611643 Chr.15: 84622513-84622513 on Build GRCh38
rs977121312 Chr.15: 84622712-84622712 on Build GRCh38
rs1033079266 Chr.15: 84622617-84622617 on Build GRCh38
rs537046115 Chr.15: 84622831-84622831 on Build GRCh38
rs747135554 Chr.15: 84622539-84622539 on Build GRCh38
rs775732146 Chr.15: 84622886-84622886 on Build GRCh38
rs1006720994 Chr.15: 84622516-84622516 on Build GRCh38
rs967001541 Chr.15: 84622708-84622708 on Build GRCh38
rs978843813 Chr.15: 84622560-84622560 on Build GRCh38
rs35373066 Chr.15: 84622562-84622563 on Build GRCh38
rs752920867 Chr.15: 84622723-84622723 on Build GRCh38
rs866190841 Chr.15: 84622653-84622653 on Build GRCh38
rs773940427 Chr.15: 84622616-84622616 on Build GRCh38
rs925857355 Chr.15: 84622730-84622730 on Build GRCh38
rs1051814280 Chr.15: 84622872-84622872 on Build GRCh38
rs992569645 Chr.15: 84622671-84622671 on Build GRCh38
rs929678707 Chr.15: 84622826-84622826 on Build GRCh38
rs191013510 Chr.15: 84622765-84622765 on Build GRCh38
rs768530916 Chr.15: 84622506-84622506 on Build GRCh38
rs548556091 Chr.15: 84622787-84622787 on Build GRCh38
rs1024478272 Chr.15: 84622682-84622682 on Build GRCh38
rs917371888 Chr.15: 84622721-84622721 on Build GRCh38

View Full Product Details