Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11254325-11254818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS1
Entrez Gene ID
8651
Gene Name
suppressor of cytokine signaling 1
Gene Aliases
CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3
UniGene
Hs.50640
Gene Chromosome Location
Chr.16: 11254417-11256182 on Build GRCh38
Transcripts
NM_003745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
36162 c.? p.? Chr.16: 11254783-11254784 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938748385 Chr.16: 11254714-11254714 on Build GRCh38
rs958461470 Chr.16: 11254686-11254686 on Build GRCh38
rs1005106801 Chr.16: 11254659-11254659 on Build GRCh38
rs35739545 Chr.16: 11254417-11254417 on Build GRCh38
rs964236927 Chr.16: 11254736-11254736 on Build GRCh38
rs986962150 Chr.16: 11254402-11254402 on Build GRCh38
rs187731105 Chr.16: 11254649-11254649 on Build GRCh38
rs934892112 Chr.16: 11254416-11254417 on Build GRCh38
rs943857763 Chr.16: 11254566-11254566 on Build GRCh38
rs996948668 Chr.16: 11254764-11254765 on Build GRCh38
rs575199723 Chr.16: 11254789-11254789 on Build GRCh38
rs567196031 Chr.16: 11254628-11254628 on Build GRCh38
rs990187265 Chr.16: 11254419-11254419 on Build GRCh38
rs908037420 Chr.16: 11254401-11254401 on Build GRCh38
rs1031822587 Chr.16: 11254657-11254657 on Build GRCh38
rs1034517453 Chr.16: 11254678-11254678 on Build GRCh38
rs984133546 Chr.16: 11254398-11254398 on Build GRCh38
rs113504623 Chr.16: 11254408-11254408 on Build GRCh38
rs199732720 Chr.16: 11254417-11254417 on Build GRCh38
rs1016613925 Chr.16: 11254542-11254542 on Build GRCh38
rs530331412 Chr.16: 11254584-11254584 on Build GRCh38
rs370928470 Chr.16: 11254795-11254795 on Build GRCh38
rs753605441 Chr.16: 11254791-11254791 on Build GRCh38
rs555047093 Chr.16: 11254705-11254705 on Build GRCh38
rs1056214690 Chr.16: 11254498-11254498 on Build GRCh38
rs34665126 Chr.16: 11254387-11254387 on Build GRCh38
rs1011299449 Chr.16: 11254796-11254796 on Build GRCh38
rs752261706 Chr.16: 11254669-11254669 on Build GRCh38
rs568420700 Chr.16: 11254724-11254724 on Build GRCh38
rs115979730 Chr.16: 11254418-11254418 on Build GRCh38
rs182354564 Chr.16: 11254573-11254573 on Build GRCh38
rs755754896 Chr.16: 11254685-11254685 on Build GRCh38
rs8060459 Chr.16: 11254416-11254416 on Build GRCh38
rs977299722 Chr.16: 11254703-11254703 on Build GRCh38
rs759589457 Chr.16: 11254408-11254409 on Build GRCh38
rs1040450017 Chr.16: 11254572-11254572 on Build GRCh38
rs1056245042 Chr.16: 11254716-11254716 on Build GRCh38
rs975539353 Chr.16: 11254375-11254375 on Build GRCh38
rs945970861 Chr.16: 11254451-11254451 on Build GRCh38
rs754064101 Chr.16: 11254527-11254528 on Build GRCh38
rs76979931 Chr.16: 11254415-11254418 on Build GRCh38
rs1004847464 Chr.16: 11254768-11254768 on Build GRCh38
rs34927167 Chr.16: 11254604-11254604 on Build GRCh38
rs990333772 Chr.16: 11254692-11254692 on Build GRCh38
rs898407449 Chr.16: 11254755-11254755 on Build GRCh38
rs572828363 Chr.16: 11254765-11254765 on Build GRCh38
rs537999563 Chr.16: 11254682-11254682 on Build GRCh38
rs577171732 Chr.16: 11254792-11254796 on Build GRCh38
rs981116754 Chr.16: 11254409-11254409 on Build GRCh38
rs564874619 Chr.16: 11254415-11254415 on Build GRCh38
rs569224392 Chr.16: 11254675-11254675 on Build GRCh38
rs1049625966 Chr.16: 11254751-11254751 on Build GRCh38
rs538539048 Chr.16: 11254770-11254770 on Build GRCh38
rs887049001 Chr.16: 11254524-11254524 on Build GRCh38
rs113432632 Chr.16: 11254664-11254664 on Build GRCh38
rs914244924 Chr.16: 11254449-11254449 on Build GRCh38
rs1028106475 Chr.16: 11254394-11254394 on Build GRCh38
rs558316619 Chr.16: 11254773-11254773 on Build GRCh38
rs971552083 Chr.16: 11254407-11254407 on Build GRCh38
rs1015148445 Chr.16: 11254660-11254660 on Build GRCh38
rs1028113719 Chr.16: 11254626-11254626 on Build GRCh38
rs945719913 Chr.16: 11254700-11254700 on Build GRCh38
rs939222408 Chr.16: 11254480-11254480 on Build GRCh38
rs887761913 Chr.16: 11254758-11254758 on Build GRCh38
rs879102216 Chr.16: 11254760-11254760 on Build GRCh38
rs796537318 Chr.16: 11254426-11254426 on Build GRCh38
rs980065389 Chr.16: 11254701-11254701 on Build GRCh38
rs1041666735 Chr.16: 11254522-11254522 on Build GRCh38
rs981567045 Chr.16: 11254672-11254672 on Build GRCh38
rs1039556584 Chr.16: 11254740-11254740 on Build GRCh38
rs144458712 Chr.16: 11254408-11254408 on Build GRCh38
rs868458526 Chr.16: 11254372-11254372 on Build GRCh38
rs532835330 Chr.16: 11254642-11254642 on Build GRCh38
rs552720624 Chr.16: 11254748-11254748 on Build GRCh38
rs750485560 Chr.16: 11254404-11254406 on Build GRCh38
rs964670987 Chr.16: 11254363-11254363 on Build GRCh38
rs536056934 Chr.16: 11254741-11254741 on Build GRCh38
rs111727771 Chr.16: 11254395-11254395 on Build GRCh38
rs914531385 Chr.16: 11254427-11254427 on Build GRCh38
rs534143848 Chr.16: 11254407-11254408 on Build GRCh38
rs370488138 Chr.16: 11254416-11254416 on Build GRCh38
rs1016992055 Chr.16: 11254347-11254347 on Build GRCh38
rs199828233 Chr.16: 11254407-11254407 on Build GRCh38
rs33932899 Chr.16: 11254354-11254354 on Build GRCh38
rs971503261 Chr.16: 11254670-11254670 on Build GRCh38
rs777439416 Chr.16: 11254787-11254787 on Build GRCh38
rs890937787 Chr.16: 11254541-11254541 on Build GRCh38
rs907461367 Chr.16: 11254781-11254781 on Build GRCh38
rs760814084 Chr.16: 11254794-11254796 on Build GRCh38
rs1027637675 Chr.16: 11254640-11254640 on Build GRCh38
rs948841688 Chr.16: 11254448-11254448 on Build GRCh38
rs754377333 Chr.16: 11254472-11254472 on Build GRCh38
rs1044836151 Chr.16: 11254461-11254461 on Build GRCh38
rs1007953682 Chr.16: 11254776-11254776 on Build GRCh38
rs544292351 Chr.16: 11254444-11254444 on Build GRCh38
rs912493125 Chr.16: 11254734-11254734 on Build GRCh38
rs914211995 Chr.16: 11254696-11254696 on Build GRCh38

View Full Product Details