Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20559097-20559502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM2B
Entrez Gene ID
348158
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Gene Aliases
ACSM2, HXMA, HYST1046
UniGene
Hs.298252
Gene Chromosome Location
Chr.16: 20536261-20576379 on Build GRCh38
Transcripts
NM_001105069.1 NM_182617.3 XM_005255304.3 XM_011545824.2 XM_017023205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4256928 c.232G>A p.E78K Chr.16: 20559393-20559393 on Build GRCh38
2130039 c.178-1G>T p.? Chr.16: 20559448-20559448 on Build GRCh38
4903953 c.205C>T p.L69L Chr.16: 20559420-20559420 on Build GRCh38
4409505 c.294G>A p.S98S Chr.16: 20559331-20559331 on Build GRCh38
4892336 c.188G>A p.R63Q Chr.16: 20559437-20559437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765684143 Chr.16: 20559479-20559479 on Build GRCh38
rs767397573 Chr.16: 20559256-20559256 on Build GRCh38
rs188171953 Chr.16: 20559465-20559465 on Build GRCh38
rs778557354 Chr.16: 20559369-20559369 on Build GRCh38
rs28712600 Chr.16: 20559144-20559144 on Build GRCh38
rs371900959 Chr.16: 20559426-20559426 on Build GRCh38
rs780168424 Chr.16: 20559277-20559277 on Build GRCh38
rs888468250 Chr.16: 20559292-20559292 on Build GRCh38
rs775213714 Chr.16: 20559383-20559383 on Build GRCh38
rs765303609 Chr.16: 20559414-20559415 on Build GRCh38
rs769424453 Chr.16: 20559240-20559240 on Build GRCh38
rs766364341 Chr.16: 20559403-20559403 on Build GRCh38
rs745409071 Chr.16: 20559435-20559435 on Build GRCh38
rs924836372 Chr.16: 20559428-20559428 on Build GRCh38
rs9927854 Chr.16: 20559453-20559453 on Build GRCh38
rs1041234995 Chr.16: 20559275-20559275 on Build GRCh38
rs563800838 Chr.16: 20559457-20559457 on Build GRCh38
rs748713260 Chr.16: 20559231-20559231 on Build GRCh38
rs764482959 Chr.16: 20559475-20559475 on Build GRCh38
rs767810251 Chr.16: 20559409-20559409 on Build GRCh38
rs554694059 Chr.16: 20559284-20559284 on Build GRCh38
rs774158462 Chr.16: 20559248-20559248 on Build GRCh38
rs553128923 Chr.16: 20559333-20559333 on Build GRCh38
rs371467475 Chr.16: 20559274-20559274 on Build GRCh38
rs781663184 Chr.16: 20559282-20559282 on Build GRCh38
rs201089298 Chr.16: 20559281-20559281 on Build GRCh38
rs374648082 Chr.16: 20559291-20559291 on Build GRCh38
rs759628455 Chr.16: 20559464-20559464 on Build GRCh38
rs776526040 Chr.16: 20559398-20559398 on Build GRCh38
rs141121285 Chr.16: 20559312-20559312 on Build GRCh38
rs766528681 Chr.16: 20559340-20559340 on Build GRCh38
rs530395411 Chr.16: 20559438-20559438 on Build GRCh38
rs745441280 Chr.16: 20559372-20559372 on Build GRCh38
rs559044619 Chr.16: 20559305-20559305 on Build GRCh38
rs746715033 Chr.16: 20559447-20559447 on Build GRCh38
rs759627168 Chr.16: 20559407-20559407 on Build GRCh38
rs538802177 Chr.16: 20559242-20559242 on Build GRCh38
rs751672845 Chr.16: 20559405-20559405 on Build GRCh38
rs781353979 Chr.16: 20559358-20559358 on Build GRCh38
rs757950871 Chr.16: 20559437-20559437 on Build GRCh38
rs139440992 Chr.16: 20559259-20559259 on Build GRCh38
rs775838979 Chr.16: 20559469-20559469 on Build GRCh38
rs747976990 Chr.16: 20559456-20559456 on Build GRCh38
rs753136707 Chr.16: 20559359-20559359 on Build GRCh38
rs958703513 Chr.16: 20559134-20559134 on Build GRCh38
rs75808894 Chr.16: 20559143-20559143 on Build GRCh38
rs759295384 Chr.16: 20559309-20559309 on Build GRCh38
rs542088831 Chr.16: 20559353-20559353 on Build GRCh38
rs375844763 Chr.16: 20559276-20559276 on Build GRCh38
rs755233431 Chr.16: 20559356-20559356 on Build GRCh38
rs8056693 Chr.16: 20559339-20559339 on Build GRCh38
rs758956993 Chr.16: 20559480-20559480 on Build GRCh38
rs756106862 Chr.16: 20559268-20559268 on Build GRCh38
rs752568940 Chr.16: 20559264-20559264 on Build GRCh38
rs370716308 Chr.16: 20559200-20559200 on Build GRCh38
rs756522282 Chr.16: 20559422-20559422 on Build GRCh38
rs1048991666 Chr.16: 20559472-20559472 on Build GRCh38
rs977090394 Chr.16: 20559413-20559413 on Build GRCh38
rs148136861 Chr.16: 20559303-20559303 on Build GRCh38
rs764038642 Chr.16: 20559332-20559332 on Build GRCh38
rs760492137 Chr.16: 20559392-20559392 on Build GRCh38
rs765231557 Chr.16: 20559271-20559271 on Build GRCh38
rs968405178 Chr.16: 20559381-20559381 on Build GRCh38
rs577119435 Chr.16: 20559311-20559311 on Build GRCh38
rs766154898 Chr.16: 20559247-20559247 on Build GRCh38
rs761041455 Chr.16: 20559473-20559473 on Build GRCh38
rs753006019 Chr.16: 20559410-20559410 on Build GRCh38
rs377623779 Chr.16: 20559430-20559430 on Build GRCh38
rs566287967 Chr.16: 20559267-20559267 on Build GRCh38
rs749862085 Chr.16: 20559365-20559365 on Build GRCh38
rs771766731 Chr.16: 20559378-20559378 on Build GRCh38
rs761868480 Chr.16: 20559400-20559400 on Build GRCh38
rs267604440 Chr.16: 20559243-20559243 on Build GRCh38
rs776972411 Chr.16: 20559209-20559209 on Build GRCh38
rs756651036 Chr.16: 20559364-20559364 on Build GRCh38
rs770337678 Chr.16: 20559367-20559367 on Build GRCh38
rs369411104 Chr.16: 20559331-20559331 on Build GRCh38
rs774254811 Chr.16: 20559462-20559462 on Build GRCh38
rs563151418 Chr.16: 20559395-20559395 on Build GRCh38
rs74479331 Chr.16: 20559434-20559434 on Build GRCh38
rs757791905 Chr.16: 20559432-20559432 on Build GRCh38
rs148987328 Chr.16: 20559255-20559255 on Build GRCh38
rs750919229 Chr.16: 20559452-20559452 on Build GRCh38
rs551819110 Chr.16: 20559171-20559171 on Build GRCh38
rs200636112 Chr.16: 20559452-20559452 on Build GRCh38
rs143779927 Chr.16: 20559302-20559302 on Build GRCh38

View Full Product Details