Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20733542-20734083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THUMPD1
Entrez Gene ID
55623
Gene Name
THUMP domain containing 1
Gene Aliases
Tan1
UniGene
Hs.700004
Gene Chromosome Location
Chr.16: 20730255-20741964 on Build GRCh38
Transcripts
NM_001304550.1 NM_017736.4 XM_011545891.1 XM_017023433.1

SNP Detail

SNP ID Chromosome Location
rs957076330 Chr.16: 20733580-20733580 on Build GRCh38
rs1009668842 Chr.16: 20733863-20733863 on Build GRCh38
rs929343973 Chr.16: 20733670-20733670 on Build GRCh38
rs887051356 Chr.16: 20733686-20733686 on Build GRCh38
rs1031419043 Chr.16: 20733572-20733572 on Build GRCh38
rs745887626 Chr.16: 20733605-20733605 on Build GRCh38
rs556942323 Chr.16: 20733594-20733595 on Build GRCh38
rs987748504 Chr.16: 20733875-20733875 on Build GRCh38
rs957759653 Chr.16: 20733826-20733826 on Build GRCh38
rs1002034333 Chr.16: 20733739-20733739 on Build GRCh38
rs1037754431 Chr.16: 20733655-20733655 on Build GRCh38
rs573662332 Chr.16: 20733728-20733728 on Build GRCh38
rs1058904 Chr.16: 20733955-20733955 on Build GRCh38
rs751180385 Chr.16: 20733586-20733590 on Build GRCh38
rs370556639 Chr.16: 20733771-20733771 on Build GRCh38
rs954497459 Chr.16: 20733874-20733875 on Build GRCh38
rs943504973 Chr.16: 20733654-20733654 on Build GRCh38
rs8058815 Chr.16: 20734039-20734039 on Build GRCh38
rs573256645 Chr.16: 20733684-20733688 on Build GRCh38
rs990309214 Chr.16: 20733593-20733593 on Build GRCh38
rs538038885 Chr.16: 20734056-20734056 on Build GRCh38
rs921418722 Chr.16: 20733600-20733600 on Build GRCh38
rs954369154 Chr.16: 20733618-20733618 on Build GRCh38
rs905036473 Chr.16: 20733737-20733737 on Build GRCh38
rs1058905 Chr.16: 20733933-20733933 on Build GRCh38
rs917271792 Chr.16: 20733957-20733957 on Build GRCh38
rs770337957 Chr.16: 20733634-20733634 on Build GRCh38
rs910763060 Chr.16: 20733881-20733881 on Build GRCh38
rs111573937 Chr.16: 20733983-20733983 on Build GRCh38
rs549488354 Chr.16: 20734022-20734022 on Build GRCh38
rs775818275 Chr.16: 20733727-20733727 on Build GRCh38
rs973534735 Chr.16: 20733923-20733924 on Build GRCh38
rs929376917 Chr.16: 20733935-20733935 on Build GRCh38
rs987057240 Chr.16: 20733623-20733623 on Build GRCh38
rs910733022 Chr.16: 20733652-20733652 on Build GRCh38
rs867872519 Chr.16: 20733829-20733829 on Build GRCh38
rs184650960 Chr.16: 20734048-20734048 on Build GRCh38
rs75930210 Chr.16: 20733864-20733864 on Build GRCh38
rs551806303 Chr.16: 20733896-20733896 on Build GRCh38
rs950060783 Chr.16: 20734033-20734033 on Build GRCh38
rs940021731 Chr.16: 20733649-20733649 on Build GRCh38
rs753414122 Chr.16: 20734055-20734055 on Build GRCh38
rs371378377 Chr.16: 20733643-20733646 on Build GRCh38

View Full Product Details