Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67280790-67281206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG4
Entrez Gene ID
25894
Gene Name
pleckstrin homology and RhoGEF domain containing G4
Gene Aliases
ARHGEF44, PRTPHN1, SCA4
UniGene
Hs.188781
Gene Chromosome Location
Chr.16: 67277820-67289500 on Build GRCh38
Transcripts
NM_001129727.2 NM_001129728.1 NM_001129729.2 NM_001129731.2 XM_011522985.2 XM_011522986.2 XM_011522987.2 XM_011522988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5814565 c.665A>T p.E222V Chr.16: 67280951-67280951 on Build GRCh38
5604539 c.782C>A p.S261Y Chr.16: 67281153-67281153 on Build GRCh38
1609514 c.776C>A p.P259Q Chr.16: 67281147-67281147 on Build GRCh38
7292976 c.687C>T p.I229I Chr.16: 67280973-67280973 on Build GRCh38
263000 c.764G>A p.R255Q Chr.16: 67281135-67281135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757990422 Chr.16: 67280988-67280988 on Build GRCh38
rs1034879954 Chr.16: 67280931-67280931 on Build GRCh38
rs781004890 Chr.16: 67281131-67281131 on Build GRCh38
rs368176497 Chr.16: 67281003-67281003 on Build GRCh38
rs918702277 Chr.16: 67280826-67280826 on Build GRCh38
rs777342739 Chr.16: 67280838-67280838 on Build GRCh38
rs777483435 Chr.16: 67280877-67280877 on Build GRCh38
rs778302356 Chr.16: 67281034-67281035 on Build GRCh38
rs201712586 Chr.16: 67280926-67280926 on Build GRCh38
rs902297806 Chr.16: 67280890-67280890 on Build GRCh38
rs543547835 Chr.16: 67280979-67280979 on Build GRCh38
rs752768730 Chr.16: 67280984-67280984 on Build GRCh38
rs771380636 Chr.16: 67280882-67280882 on Build GRCh38
rs535977084 Chr.16: 67281094-67281094 on Build GRCh38
rs199594589 Chr.16: 67280843-67280843 on Build GRCh38
rs1039952135 Chr.16: 67280872-67280872 on Build GRCh38
rs376153070 Chr.16: 67280816-67280816 on Build GRCh38
rs759343530 Chr.16: 67281070-67281070 on Build GRCh38
rs371376758 Chr.16: 67280839-67280839 on Build GRCh38
rs767760292 Chr.16: 67281064-67281064 on Build GRCh38
rs1029160923 Chr.16: 67281120-67281120 on Build GRCh38
rs777109981 Chr.16: 67280886-67280886 on Build GRCh38
rs752793302 Chr.16: 67280840-67280840 on Build GRCh38
rs190827633 Chr.16: 67280960-67280960 on Build GRCh38
rs140748514 Chr.16: 67281163-67281163 on Build GRCh38
rs770455111 Chr.16: 67280898-67280898 on Build GRCh38
rs142455848 Chr.16: 67281041-67281041 on Build GRCh38
rs561875011 Chr.16: 67280992-67280992 on Build GRCh38
rs199714439 Chr.16: 67280987-67280987 on Build GRCh38
rs753603363 Chr.16: 67280975-67280975 on Build GRCh38
rs1021764688 Chr.16: 67281093-67281093 on Build GRCh38
rs386791752 Chr.16: 67280859-67280882 on Build GRCh38
rs747851059 Chr.16: 67280830-67280830 on Build GRCh38
rs763366247 Chr.16: 67281040-67281040 on Build GRCh38
rs149840175 Chr.16: 67281134-67281134 on Build GRCh38
rs373837124 Chr.16: 67281034-67281034 on Build GRCh38
rs771437333 Chr.16: 67280861-67280861 on Build GRCh38
rs773378992 Chr.16: 67281068-67281068 on Build GRCh38
rs968828956 Chr.16: 67281097-67281097 on Build GRCh38
rs372449397 Chr.16: 67281056-67281056 on Build GRCh38
rs760870289 Chr.16: 67281182-67281182 on Build GRCh38
rs199563453 Chr.16: 67281172-67281172 on Build GRCh38
rs149229558 Chr.16: 67280887-67280887 on Build GRCh38
rs559572262 Chr.16: 67281107-67281107 on Build GRCh38
rs770224196 Chr.16: 67281037-67281037 on Build GRCh38
rs140708063 Chr.16: 67281095-67281095 on Build GRCh38
rs771729893 Chr.16: 67280831-67280831 on Build GRCh38
rs953883189 Chr.16: 67281124-67281124 on Build GRCh38
rs746720610 Chr.16: 67280993-67280993 on Build GRCh38
rs761126537 Chr.16: 67280834-67280834 on Build GRCh38
rs951495494 Chr.16: 67281118-67281118 on Build GRCh38
rs114723164 Chr.16: 67280974-67280974 on Build GRCh38
rs374144842 Chr.16: 67280879-67280879 on Build GRCh38
rs751061606 Chr.16: 67281108-67281108 on Build GRCh38
rs778438224 Chr.16: 67280855-67280855 on Build GRCh38
rs746256512 Chr.16: 67281031-67281031 on Build GRCh38
rs377700816 Chr.16: 67280852-67280852 on Build GRCh38
rs747689846 Chr.16: 67280859-67280859 on Build GRCh38
rs745446321 Chr.16: 67281133-67281133 on Build GRCh38
rs763106385 Chr.16: 67280903-67280903 on Build GRCh38
rs761928081 Chr.16: 67280965-67280965 on Build GRCh38
rs780951600 Chr.16: 67281019-67281019 on Build GRCh38
rs755943666 Chr.16: 67280853-67280853 on Build GRCh38
rs773430423 Chr.16: 67281181-67281181 on Build GRCh38
rs776088693 Chr.16: 67280900-67280900 on Build GRCh38
rs182286285 Chr.16: 67281044-67281044 on Build GRCh38
rs764397738 Chr.16: 67280846-67280846 on Build GRCh38
rs764015407 Chr.16: 67280923-67280923 on Build GRCh38
rs149654792 Chr.16: 67280818-67280818 on Build GRCh38
rs754891531 Chr.16: 67280854-67280854 on Build GRCh38
rs776143749 Chr.16: 67281039-67281039 on Build GRCh38
rs758243484 Chr.16: 67281103-67281103 on Build GRCh38
rs866008873 Chr.16: 67281158-67281158 on Build GRCh38
rs778846844 Chr.16: 67280983-67280983 on Build GRCh38
rs766583217 Chr.16: 67280837-67280837 on Build GRCh38
rs145413083 Chr.16: 67280829-67280829 on Build GRCh38
rs34936270 Chr.16: 67281082-67281082 on Build GRCh38
rs60087996 Chr.16: 67280824-67280824 on Build GRCh38
rs145862477 Chr.16: 67281135-67281135 on Build GRCh38
rs763633832 Chr.16: 67281178-67281178 on Build GRCh38
rs756712177 Chr.16: 67280987-67280988 on Build GRCh38
rs756740484 Chr.16: 67281126-67281126 on Build GRCh38
rs752576320 Chr.16: 67280984-67280984 on Build GRCh38
rs893350612 Chr.16: 67280876-67280876 on Build GRCh38
rs556014414 Chr.16: 67280894-67280894 on Build GRCh38

View Full Product Details