Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69330999-69331510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG8
Entrez Gene ID
84342
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
CDG2H, DOR1
UniGene
Hs.130849
Gene Chromosome Location
Chr.16: 69328621-69339623 on Build GRCh38
Transcripts
NM_032382.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3387557 c.1641C>A p.A547A Chr.16: 69331037-69331037 on Build GRCh38
3272178 c.1583G>A p.G528D Chr.16: 69331095-69331095 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs121434387 Chr.16: 69331067-69331067 on Build GRCh38
rs780751637 Chr.16: 69331129-69331130 on Build GRCh38
rs1022908241 Chr.16: 69331221-69331221 on Build GRCh38
rs186439067 Chr.16: 69331385-69331385 on Build GRCh38
rs535913998 Chr.16: 69331074-69331074 on Build GRCh38
rs144045802 Chr.16: 69331141-69331141 on Build GRCh38
rs1015182755 Chr.16: 69331287-69331287 on Build GRCh38
rs947827372 Chr.16: 69331216-69331216 on Build GRCh38
rs746737547 Chr.16: 69331084-69331084 on Build GRCh38
rs753870306 Chr.16: 69331140-69331140 on Build GRCh38
rs181040346 Chr.16: 69331219-69331219 on Build GRCh38
rs1029033156 Chr.16: 69331160-69331160 on Build GRCh38
rs763454942 Chr.16: 69331021-69331021 on Build GRCh38
rs1008106051 Chr.16: 69331301-69331301 on Build GRCh38
rs189199610 Chr.16: 69331058-69331058 on Build GRCh38
rs527885323 Chr.16: 69331305-69331305 on Build GRCh38
rs956951928 Chr.16: 69331185-69331185 on Build GRCh38
rs565175745 Chr.16: 69331176-69331176 on Build GRCh38
rs997033711 Chr.16: 69331229-69331229 on Build GRCh38
rs957183498 Chr.16: 69331379-69331379 on Build GRCh38
rs918056100 Chr.16: 69331210-69331210 on Build GRCh38
rs538509057 Chr.16: 69331372-69331372 on Build GRCh38
rs966529559 Chr.16: 69331153-69331153 on Build GRCh38
rs774861686 Chr.16: 69331106-69331106 on Build GRCh38
rs139824417 Chr.16: 69331250-69331250 on Build GRCh38
rs887044170 Chr.16: 69331270-69331270 on Build GRCh38
rs751924599 Chr.16: 69331040-69331040 on Build GRCh38
rs571146279 Chr.16: 69331312-69331312 on Build GRCh38
rs777572193 Chr.16: 69331076-69331076 on Build GRCh38
rs1029065698 Chr.16: 69331319-69331319 on Build GRCh38
rs939393768 Chr.16: 69331448-69331448 on Build GRCh38
rs973888130 Chr.16: 69331151-69331151 on Build GRCh38
rs764532175 Chr.16: 69331022-69331022 on Build GRCh38
rs769321092 Chr.16: 69331094-69331094 on Build GRCh38
rs918117529 Chr.16: 69331392-69331392 on Build GRCh38
rs750627777 Chr.16: 69331124-69331124 on Build GRCh38
rs111336106 Chr.16: 69331035-69331035 on Build GRCh38
rs114861924 Chr.16: 69331089-69331089 on Build GRCh38
rs764658768 Chr.16: 69331103-69331103 on Build GRCh38
rs968039469 Chr.16: 69331169-69331169 on Build GRCh38
rs754723553 Chr.16: 69331228-69331228 on Build GRCh38
rs1019419838 Chr.16: 69331148-69331148 on Build GRCh38
rs149657356 Chr.16: 69331304-69331304 on Build GRCh38
rs906312296 Chr.16: 69331485-69331485 on Build GRCh38
rs568421717 Chr.16: 69331418-69331418 on Build GRCh38
rs540188221 Chr.16: 69331218-69331219 on Build GRCh38
rs77548036 Chr.16: 69331477-69331477 on Build GRCh38
rs996241699 Chr.16: 69331159-69331159 on Build GRCh38
rs1046684095 Chr.16: 69331266-69331266 on Build GRCh38
rs928073768 Chr.16: 69331390-69331390 on Build GRCh38
rs370982886 Chr.16: 69331101-69331101 on Build GRCh38
rs924178127 Chr.16: 69331180-69331180 on Build GRCh38
rs766499868 Chr.16: 69331127-69331127 on Build GRCh38
rs762253690 Chr.16: 69331107-69331107 on Build GRCh38
rs1045155186 Chr.16: 69331199-69331199 on Build GRCh38
rs757586660 Chr.16: 69331041-69331041 on Build GRCh38
rs576761446 Chr.16: 69331178-69331178 on Build GRCh38
rs866749880 Chr.16: 69331408-69331408 on Build GRCh38
rs968070773 Chr.16: 69331324-69331324 on Build GRCh38
rs1000883985 Chr.16: 69331329-69331329 on Build GRCh38
rs376430929 Chr.16: 69331065-69331065 on Build GRCh38
rs566819490 Chr.16: 69331254-69331254 on Build GRCh38
rs982428881 Chr.16: 69331282-69331282 on Build GRCh38
rs552538846 Chr.16: 69331306-69331306 on Build GRCh38
rs368338806 Chr.16: 69331126-69331126 on Build GRCh38
rs755315633 Chr.16: 69331128-69331129 on Build GRCh38
rs915771016 Chr.16: 69331355-69331355 on Build GRCh38
rs545646295 Chr.16: 69331219-69331219 on Build GRCh38
rs1029421323 Chr.16: 69331233-69331233 on Build GRCh38
rs747868745 Chr.16: 69331144-69331144 on Build GRCh38
rs200972444 Chr.16: 69331057-69331057 on Build GRCh38
rs745439091 Chr.16: 69331091-69331091 on Build GRCh38
rs1044683243 Chr.16: 69331479-69331479 on Build GRCh38
rs9928658 Chr.16: 69331220-69331220 on Build GRCh38
rs992756520 Chr.16: 69331205-69331205 on Build GRCh38
rs781410055 Chr.16: 69331048-69331048 on Build GRCh38
rs908466902 Chr.16: 69331144-69331145 on Build GRCh38
rs751980937 Chr.16: 69331057-69331058 on Build GRCh38
rs76253513 Chr.16: 69331087-69331087 on Build GRCh38
rs1052448431 Chr.16: 69331473-69331473 on Build GRCh38
rs906563626 Chr.16: 69331213-69331213 on Build GRCh38
rs1056248553 Chr.16: 69331049-69331049 on Build GRCh38
rs996705229 Chr.16: 69331316-69331316 on Build GRCh38
rs111432987 Chr.16: 69331219-69331220 on Build GRCh38
rs374439014 Chr.16: 69331115-69331115 on Build GRCh38

View Full Product Details