Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70659890-70660383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
UniGene
Hs.461214
Gene Chromosome Location
Chr.16: 70579895-70660682 on Build GRCh38
Transcripts
NM_001172771.1 NM_001172772.1 NM_152456.2 XM_011522902.2 XM_011522903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
417291 c.686C>T p.S229L Chr.16: 70660144-70660144 on Build GRCh38
6685336 c.552C>T p.S184S Chr.16: 70660010-70660010 on Build GRCh38
417292 c.623C>T p.A208V Chr.16: 70660081-70660081 on Build GRCh38
5971897 c.659C>A p.P220H Chr.16: 70660117-70660117 on Build GRCh38
4062388 c.618G>C p.Q206H Chr.16: 70660076-70660076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048635773 Chr.16: 70660162-70660162 on Build GRCh38
rs921788134 Chr.16: 70659915-70659916 on Build GRCh38
rs753771953 Chr.16: 70660158-70660158 on Build GRCh38
rs4985556 Chr.16: 70660097-70660097 on Build GRCh38
rs774449875 Chr.16: 70660002-70660002 on Build GRCh38
rs370957526 Chr.16: 70660194-70660194 on Build GRCh38
rs770634170 Chr.16: 70659951-70659951 on Build GRCh38
rs112385962 Chr.16: 70659925-70659925 on Build GRCh38
rs746778123 Chr.16: 70659948-70659948 on Build GRCh38
rs754912577 Chr.16: 70660164-70660164 on Build GRCh38
rs927551998 Chr.16: 70659962-70659962 on Build GRCh38
rs980832612 Chr.16: 70660299-70660299 on Build GRCh38
rs201952472 Chr.16: 70659981-70659981 on Build GRCh38
rs7206509 Chr.16: 70660042-70660042 on Build GRCh38
rs755937255 Chr.16: 70660112-70660112 on Build GRCh38
rs766224487 Chr.16: 70660154-70660154 on Build GRCh38
rs750167732 Chr.16: 70660111-70660111 on Build GRCh38
rs780602039 Chr.16: 70660175-70660175 on Build GRCh38
rs530529839 Chr.16: 70660298-70660298 on Build GRCh38
rs3214847 Chr.16: 70659970-70659971 on Build GRCh38
rs1052897020 Chr.16: 70659940-70659940 on Build GRCh38
rs747223979 Chr.16: 70660126-70660126 on Build GRCh38
rs749574027 Chr.16: 70660237-70660237 on Build GRCh38
rs772052353 Chr.16: 70660087-70660087 on Build GRCh38
rs141513638 Chr.16: 70660153-70660153 on Build GRCh38
rs926860502 Chr.16: 70660311-70660311 on Build GRCh38
rs766132144 Chr.16: 70660208-70660208 on Build GRCh38
rs777183793 Chr.16: 70660045-70660045 on Build GRCh38
rs771592770 Chr.16: 70659985-70659986 on Build GRCh38
rs144464320 Chr.16: 70660173-70660173 on Build GRCh38
rs200144613 Chr.16: 70660109-70660109 on Build GRCh38
rs769403976 Chr.16: 70660094-70660094 on Build GRCh38
rs773778670 Chr.16: 70660144-70660144 on Build GRCh38
rs577167294 Chr.16: 70659967-70659967 on Build GRCh38
rs1036683289 Chr.16: 70660283-70660283 on Build GRCh38
rs917604278 Chr.16: 70660275-70660275 on Build GRCh38
rs757397918 Chr.16: 70660227-70660227 on Build GRCh38
rs758011880 Chr.16: 70660062-70660062 on Build GRCh38
rs575041676 Chr.16: 70660315-70660315 on Build GRCh38
rs748188942 Chr.16: 70660083-70660083 on Build GRCh38
rs768176846 Chr.16: 70660142-70660142 on Build GRCh38
rs780153224 Chr.16: 70659996-70659996 on Build GRCh38
rs762466829 Chr.16: 70660103-70660103 on Build GRCh38
rs576160948 Chr.16: 70659970-70659970 on Build GRCh38
rs570123541 Chr.16: 70660184-70660184 on Build GRCh38
rs777016133 Chr.16: 70660129-70660129 on Build GRCh38
rs189265997 Chr.16: 70659980-70659980 on Build GRCh38
rs747715186 Chr.16: 70659988-70659988 on Build GRCh38
rs369568962 Chr.16: 70660034-70660034 on Build GRCh38
rs1019565461 Chr.16: 70659913-70659913 on Build GRCh38
rs953329368 Chr.16: 70660265-70660265 on Build GRCh38
rs375275661 Chr.16: 70660216-70660216 on Build GRCh38
rs745592299 Chr.16: 70659983-70659984 on Build GRCh38
rs780442530 Chr.16: 70660236-70660236 on Build GRCh38
rs993058245 Chr.16: 70660274-70660274 on Build GRCh38
rs749776814 Chr.16: 70660182-70660182 on Build GRCh38
rs761632445 Chr.16: 70659922-70659922 on Build GRCh38
rs772292258 Chr.16: 70660010-70660010 on Build GRCh38
rs112639369 Chr.16: 70660170-70660170 on Build GRCh38
rs978910378 Chr.16: 70659949-70659949 on Build GRCh38
rs199910768 Chr.16: 70660082-70660082 on Build GRCh38
rs779361515 Chr.16: 70659981-70659981 on Build GRCh38
rs763733561 Chr.16: 70660063-70660063 on Build GRCh38
rs781489072 Chr.16: 70660231-70660231 on Build GRCh38
rs908679598 Chr.16: 70660352-70660352 on Build GRCh38
rs889733429 Chr.16: 70660105-70660105 on Build GRCh38
rs756450696 Chr.16: 70660235-70660235 on Build GRCh38
rs145782768 Chr.16: 70660076-70660076 on Build GRCh38
rs796937232 Chr.16: 70659970-70659971 on Build GRCh38
rs763570329 Chr.16: 70660106-70660106 on Build GRCh38
rs772842086 Chr.16: 70660200-70660200 on Build GRCh38
rs533578139 Chr.16: 70659939-70659939 on Build GRCh38
rs866381841 Chr.16: 70660210-70660210 on Build GRCh38
rs746523737 Chr.16: 70660163-70660164 on Build GRCh38
rs956580661 Chr.16: 70660279-70660280 on Build GRCh38
rs551796755 Chr.16: 70660183-70660183 on Build GRCh38
rs752692287 Chr.16: 70660225-70660225 on Build GRCh38
rs745769402 Chr.16: 70660078-70660078 on Build GRCh38
rs768715713 Chr.16: 70659993-70659993 on Build GRCh38
rs747139320 Chr.16: 70659969-70659970 on Build GRCh38
rs75183493 Chr.16: 70659933-70659933 on Build GRCh38
rs746167704 Chr.16: 70660131-70660131 on Build GRCh38
rs931056144 Chr.16: 70660350-70660350 on Build GRCh38
rs746149647 Chr.16: 70660171-70660171 on Build GRCh38
rs779902217 Chr.16: 70660113-70660113 on Build GRCh38
rs949101643 Chr.16: 70660281-70660281 on Build GRCh38
rs761996710 Chr.16: 70660006-70660006 on Build GRCh38
rs397711055 Chr.16: 70659980-70659981 on Build GRCh38
rs759399789 Chr.16: 70660214-70660214 on Build GRCh38
rs760390116 Chr.16: 70660207-70660207 on Build GRCh38
rs918011858 Chr.16: 70660297-70660297 on Build GRCh38
rs752305128 Chr.16: 70660057-70660057 on Build GRCh38
rs150579141 Chr.16: 70660169-70660169 on Build GRCh38
rs200444183 Chr.16: 70660139-70660139 on Build GRCh38
rs755146539 Chr.16: 70659964-70659964 on Build GRCh38
rs781232520 Chr.16: 70660147-70660147 on Build GRCh38
rs765046412 Chr.16: 70660217-70660217 on Build GRCh38
rs770090002 Chr.16: 70660285-70660285 on Build GRCh38
rs372998917 Chr.16: 70660138-70660138 on Build GRCh38
rs931826870 Chr.16: 70659915-70659915 on Build GRCh38
rs1048578402 Chr.16: 70660343-70660343 on Build GRCh38
rs931578543 Chr.16: 70660330-70660330 on Build GRCh38
rs757060030 Chr.16: 70660072-70660072 on Build GRCh38
rs1048222155 Chr.16: 70660361-70660361 on Build GRCh38
rs138439611 Chr.16: 70660088-70660088 on Build GRCh38
rs202122982 Chr.16: 70660098-70660098 on Build GRCh38
rs201784459 Chr.16: 70660102-70660102 on Build GRCh38
rs201277640 Chr.16: 70660081-70660081 on Build GRCh38
rs148286339 Chr.16: 70660108-70660108 on Build GRCh38
rs79922158 Chr.16: 70660067-70660067 on Build GRCh38
rs777887915 Chr.16: 70660167-70660167 on Build GRCh38
rs779754742 Chr.16: 70659958-70659958 on Build GRCh38
rs1028832359 Chr.16: 70660246-70660246 on Build GRCh38
rs192356278 Chr.16: 70660145-70660145 on Build GRCh38
rs764810390 Chr.16: 70660055-70660055 on Build GRCh38
rs751216629 Chr.16: 70660107-70660107 on Build GRCh38
rs537802994 Chr.16: 70660232-70660232 on Build GRCh38
rs749242332 Chr.16: 70660114-70660114 on Build GRCh38
rs989262229 Chr.16: 70659926-70659926 on Build GRCh38
rs775115913 Chr.16: 70660041-70660042 on Build GRCh38
rs532486484 Chr.16: 70660117-70660117 on Build GRCh38
rs761163867 Chr.16: 70660191-70660191 on Build GRCh38
rs558111511 Chr.16: 70659959-70659959 on Build GRCh38
rs753626247 Chr.16: 70660211-70660211 on Build GRCh38
rs773341434 Chr.16: 70660011-70660011 on Build GRCh38
rs550608624 Chr.16: 70660127-70660127 on Build GRCh38

View Full Product Details