Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8753182-8753706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDYE4
Entrez Gene ID
388333
Gene Name
speedy/RINGO cell cycle regulator family member E4
Gene Aliases
-
UniGene
Hs.368429
Gene Chromosome Location
Chr.17: 8746868-8759428 on Build GRCh38
Transcripts
NM_001128076.1 XM_005256632.4 XM_005256633.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3890740 c.592C>T p.R198* Chr.17: 8753383-8753383 on Build GRCh38
5539614 c.635C>T p.S212F Chr.17: 8753340-8753340 on Build GRCh38
986358 c.540C>A p.F180L Chr.17: 8753435-8753435 on Build GRCh38
986357 c.559A>G p.N187D Chr.17: 8753416-8753416 on Build GRCh38
6427402 c.533A>G p.D178G Chr.17: 8753442-8753442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762397158 Chr.17: 8753441-8753441 on Build GRCh38
rs776561313 Chr.17: 8753433-8753433 on Build GRCh38
rs61319764 Chr.17: 8753641-8753641 on Build GRCh38
rs779116942 Chr.17: 8753300-8753300 on Build GRCh38
rs751248906 Chr.17: 8753462-8753462 on Build GRCh38
rs764250329 Chr.17: 8753354-8753354 on Build GRCh38
rs758072371 Chr.17: 8753377-8753377 on Build GRCh38
rs56985159 Chr.17: 8753533-8753533 on Build GRCh38
rs774678760 Chr.17: 8753281-8753281 on Build GRCh38
rs772500424 Chr.17: 8753473-8753473 on Build GRCh38
rs539761927 Chr.17: 8753333-8753333 on Build GRCh38
rs1012491354 Chr.17: 8753576-8753576 on Build GRCh38
rs555868095 Chr.17: 8753461-8753461 on Build GRCh38
rs147104347 Chr.17: 8753662-8753662 on Build GRCh38
rs141770543 Chr.17: 8753657-8753657 on Build GRCh38
rs778582254 Chr.17: 8753209-8753209 on Build GRCh38
rs568054759 Chr.17: 8753246-8753246 on Build GRCh38
rs1012028682 Chr.17: 8753397-8753397 on Build GRCh38
rs769602268 Chr.17: 8753404-8753404 on Build GRCh38
rs750829507 Chr.17: 8753450-8753450 on Build GRCh38
rs780646534 Chr.17: 8753403-8753403 on Build GRCh38
rs776778791 Chr.17: 8753536-8753536 on Build GRCh38
rs748998840 Chr.17: 8753406-8753406 on Build GRCh38
rs1030644747 Chr.17: 8753624-8753624 on Build GRCh38
rs986991168 Chr.17: 8753471-8753471 on Build GRCh38
rs561849273 Chr.17: 8753606-8753606 on Build GRCh38
rs897053734 Chr.17: 8753504-8753504 on Build GRCh38
rs577934195 Chr.17: 8753544-8753544 on Build GRCh38
rs976256393 Chr.17: 8753598-8753598 on Build GRCh38
rs957615959 Chr.17: 8753560-8753560 on Build GRCh38
rs757488898 Chr.17: 8753281-8753282 on Build GRCh38
rs768611408 Chr.17: 8753316-8753316 on Build GRCh38
rs141972356 Chr.17: 8753277-8753292 on Build GRCh38
rs747008775 Chr.17: 8753244-8753244 on Build GRCh38
rs896344573 Chr.17: 8753574-8753574 on Build GRCh38
rs765925123 Chr.17: 8753276-8753277 on Build GRCh38
rs756110114 Chr.17: 8753503-8753503 on Build GRCh38
rs757161745 Chr.17: 8753521-8753521 on Build GRCh38
rs1056312865 Chr.17: 8753380-8753380 on Build GRCh38
rs529003094 Chr.17: 8753617-8753617 on Build GRCh38
rs777525943 Chr.17: 8753505-8753505 on Build GRCh38
rs184746576 Chr.17: 8753344-8753344 on Build GRCh38
rs1056383537 Chr.17: 8753569-8753569 on Build GRCh38
rs886348041 Chr.17: 8753358-8753358 on Build GRCh38
rs752088667 Chr.17: 8753484-8753484 on Build GRCh38
rs565016558 Chr.17: 8753578-8753578 on Build GRCh38
rs759857191 Chr.17: 8753337-8753337 on Build GRCh38
rs1040502436 Chr.17: 8753391-8753391 on Build GRCh38
rs756695989 Chr.17: 8753289-8753289 on Build GRCh38
rs754617529 Chr.17: 8753477-8753477 on Build GRCh38
rs940605730 Chr.17: 8753564-8753564 on Build GRCh38
rs768923830 Chr.17: 8753427-8753427 on Build GRCh38
rs896292502 Chr.17: 8753382-8753382 on Build GRCh38
rs757690015 Chr.17: 8753297-8753297 on Build GRCh38
rs913510391 Chr.17: 8753568-8753568 on Build GRCh38
rs961212264 Chr.17: 8753257-8753257 on Build GRCh38
rs151292250 Chr.17: 8753604-8753604 on Build GRCh38
rs752769179 Chr.17: 8753286-8753286 on Build GRCh38
rs566674246 Chr.17: 8753349-8753349 on Build GRCh38
rs919965904 Chr.17: 8753335-8753335 on Build GRCh38
rs781057475 Chr.17: 8753323-8753323 on Build GRCh38
rs550287502 Chr.17: 8753285-8753285 on Build GRCh38
rs754531721 Chr.17: 8753392-8753392 on Build GRCh38
rs556305105 Chr.17: 8753538-8753538 on Build GRCh38
rs756721954 Chr.17: 8753241-8753241 on Build GRCh38
rs757975494 Chr.17: 8753308-8753308 on Build GRCh38
rs750697337 Chr.17: 8753367-8753367 on Build GRCh38
rs780809689 Chr.17: 8753483-8753483 on Build GRCh38
rs368429426 Chr.17: 8753282-8753297 on Build GRCh38
rs542798931 Chr.17: 8753605-8753605 on Build GRCh38
rs770354561 Chr.17: 8753539-8753539 on Build GRCh38
rs201401384 Chr.17: 8753618-8753618 on Build GRCh38
rs532912241 Chr.17: 8753640-8753640 on Build GRCh38
rs754281323 Chr.17: 8753292-8753292 on Build GRCh38
rs746109628 Chr.17: 8753537-8753537 on Build GRCh38
rs775906704 Chr.17: 8753416-8753416 on Build GRCh38
rs868253554 Chr.17: 8753226-8753226 on Build GRCh38
rs749429473 Chr.17: 8753277-8753277 on Build GRCh38
rs775746897 Chr.17: 8753348-8753348 on Build GRCh38
rs765595061 Chr.17: 8753442-8753442 on Build GRCh38
rs370452621 Chr.17: 8753317-8753317 on Build GRCh38
rs568809932 Chr.17: 8753325-8753325 on Build GRCh38
rs776413786 Chr.17: 8753247-8753247 on Build GRCh38
rs754409502 Chr.17: 8753359-8753359 on Build GRCh38
rs761629081 Chr.17: 8753275-8753275 on Build GRCh38
rs773402571 Chr.17: 8753527-8753527 on Build GRCh38
rs568773370 Chr.17: 8753628-8753628 on Build GRCh38
rs779551915 Chr.17: 8753309-8753309 on Build GRCh38
rs763254969 Chr.17: 8753451-8753451 on Build GRCh38
rs202236200 Chr.17: 8753379-8753379 on Build GRCh38
rs7224469 Chr.17: 8753672-8753672 on Build GRCh38
rs142364566 Chr.17: 8753423-8753423 on Build GRCh38
rs759643509 Chr.17: 8753282-8753282 on Build GRCh38
rs772449283 Chr.17: 8753411-8753411 on Build GRCh38
rs751091456 Chr.17: 8753383-8753383 on Build GRCh38
rs897107459 Chr.17: 8753671-8753671 on Build GRCh38
rs771652566 Chr.17: 8753210-8753210 on Build GRCh38
rs956347099 Chr.17: 8753639-8753639 on Build GRCh38
rs371127650 Chr.17: 8753547-8753547 on Build GRCh38
rs373073240 Chr.17: 8753278-8753278 on Build GRCh38
rs765693202 Chr.17: 8753361-8753361 on Build GRCh38
rs746447428 Chr.17: 8753312-8753312 on Build GRCh38
rs35709422 Chr.17: 8753237-8753238 on Build GRCh38
rs774978572 Chr.17: 8753219-8753219 on Build GRCh38
rs888118045 Chr.17: 8753449-8753449 on Build GRCh38
rs1000933615 Chr.17: 8753443-8753443 on Build GRCh38
rs745602744 Chr.17: 8753307-8753307 on Build GRCh38
rs779074461 Chr.17: 8753431-8753433 on Build GRCh38
rs764660808 Chr.17: 8753291-8753291 on Build GRCh38
rs574032029 Chr.17: 8753507-8753507 on Build GRCh38

View Full Product Details