Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9240590-9241084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTN1
Entrez Gene ID
9423
Gene Name
netrin 1
Gene Aliases
NTN1L
UniGene
Hs.660885
Gene Chromosome Location
Chr.17: 9021542-9244000 on Build GRCh38
Transcripts
NM_004822.2

SNP Detail

SNP ID Chromosome Location
rs980849577 Chr.17: 9240881-9240882 on Build GRCh38
rs945482053 Chr.17: 9240696-9240697 on Build GRCh38
rs1037487042 Chr.17: 9240992-9240992 on Build GRCh38
rs988277487 Chr.17: 9241045-9241045 on Build GRCh38
rs538469402 Chr.17: 9240973-9240973 on Build GRCh38
rs969614164 Chr.17: 9240976-9240976 on Build GRCh38
rs993238229 Chr.17: 9240864-9240864 on Build GRCh38
rs398058568 Chr.17: 9240749-9240754 on Build GRCh38
rs924992258 Chr.17: 9240733-9240733 on Build GRCh38
rs932434138 Chr.17: 9240913-9240913 on Build GRCh38
rs372110023 Chr.17: 9240931-9240931 on Build GRCh38
rs917617226 Chr.17: 9240642-9240642 on Build GRCh38
rs969518659 Chr.17: 9240703-9240703 on Build GRCh38
rs1015355058 Chr.17: 9240686-9240686 on Build GRCh38
rs3826394 Chr.17: 9240614-9240614 on Build GRCh38
rs913576535 Chr.17: 9240827-9240827 on Build GRCh38
rs192604291 Chr.17: 9240878-9240878 on Build GRCh38
rs1036474091 Chr.17: 9240944-9240944 on Build GRCh38
rs1002292191 Chr.17: 9240991-9240991 on Build GRCh38
rs1014237505 Chr.17: 9240692-9240692 on Build GRCh38
rs913679843 Chr.17: 9241046-9241046 on Build GRCh38
rs932628090 Chr.17: 9240943-9240943 on Build GRCh38
rs537922778 Chr.17: 9240737-9240742 on Build GRCh38
rs993756633 Chr.17: 9241002-9241002 on Build GRCh38
rs569182354 Chr.17: 9240777-9240777 on Build GRCh38
rs1041104539 Chr.17: 9240931-9240932 on Build GRCh38
rs1050337166 Chr.17: 9240718-9240718 on Build GRCh38
rs567845429 Chr.17: 9240965-9240965 on Build GRCh38
rs987737866 Chr.17: 9240823-9240823 on Build GRCh38
rs754523914 Chr.17: 9240811-9240811 on Build GRCh38
rs971704099 Chr.17: 9240631-9240631 on Build GRCh38
rs3031879 Chr.17: 9240731-9240736 on Build GRCh38
rs957759069 Chr.17: 9241008-9241008 on Build GRCh38
rs572332142 Chr.17: 9240676-9240676 on Build GRCh38
rs751194819 Chr.17: 9240932-9240933 on Build GRCh38
rs958308865 Chr.17: 9240895-9240895 on Build GRCh38
rs1024687479 Chr.17: 9241013-9241013 on Build GRCh38
rs1006329180 Chr.17: 9240936-9240936 on Build GRCh38
rs188454225 Chr.17: 9240817-9240817 on Build GRCh38
rs962299084 Chr.17: 9240690-9240690 on Build GRCh38
rs897809130 Chr.17: 9240955-9240955 on Build GRCh38
rs887864944 Chr.17: 9240621-9240621 on Build GRCh38
rs556257410 Chr.17: 9240742-9240743 on Build GRCh38
rs965414425 Chr.17: 9241065-9241065 on Build GRCh38
rs6145985 Chr.17: 9240731-9240742 on Build GRCh38
rs1015926154 Chr.17: 9240619-9240619 on Build GRCh38
rs1025158875 Chr.17: 9240875-9240875 on Build GRCh38
rs1032484924 Chr.17: 9240995-9240995 on Build GRCh38
rs12453129 Chr.17: 9240617-9240617 on Build GRCh38
rs753800949 Chr.17: 9240698-9240698 on Build GRCh38
rs372928070 Chr.17: 9240740-9240740 on Build GRCh38
rs971376346 Chr.17: 9240646-9240646 on Build GRCh38
rs961805281 Chr.17: 9240624-9240624 on Build GRCh38
rs777476558 Chr.17: 9241001-9241001 on Build GRCh38
rs573751114 Chr.17: 9240809-9240809 on Build GRCh38
rs550435979 Chr.17: 9240649-9240649 on Build GRCh38
rs958256806 Chr.17: 9241066-9241066 on Build GRCh38
rs980943327 Chr.17: 9240717-9240717 on Build GRCh38
rs1006004490 Chr.17: 9240807-9240807 on Build GRCh38
rs117799337 Chr.17: 9240885-9240885 on Build GRCh38
rs756187774 Chr.17: 9240615-9240615 on Build GRCh38
rs184534380 Chr.17: 9240886-9240886 on Build GRCh38
rs1048266943 Chr.17: 9240915-9240915 on Build GRCh38
rs921230543 Chr.17: 9240912-9240912 on Build GRCh38
rs926887216 Chr.17: 9240657-9240657 on Build GRCh38
rs374885476 Chr.17: 9240772-9240772 on Build GRCh38
rs533845629 Chr.17: 9240924-9240924 on Build GRCh38
rs970892789 Chr.17: 9241044-9241044 on Build GRCh38

View Full Product Details