Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9612227-9612735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP52
Entrez Gene ID
146845
Gene Name
cilia and flagella associated protein 52
Gene Aliases
WDR16, WDRPUH
UniGene
Hs.232270
Gene Chromosome Location
Chr.17: 9576627-9643459 on Build GRCh38
Transcripts
NM_001080556.1 NM_145054.4 XM_017024227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
265546 c.936G>A p.S312S Chr.17: 9612390-9612390 on Build GRCh38
3890747 c.1014G>A p.A338A Chr.17: 9612438-9612438 on Build GRCh38
5546548 c.960G>A p.T320T Chr.17: 9612414-9612414 on Build GRCh38
223849 c.986C>T p.T329I Chr.17: 9612440-9612440 on Build GRCh38
5519455 c.905G>T p.G302V Chr.17: 9612329-9612329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377565869 Chr.17: 9612373-9612373 on Build GRCh38
rs112097194 Chr.17: 9612620-9612620 on Build GRCh38
rs777291573 Chr.17: 9612450-9612450 on Build GRCh38
rs376661862 Chr.17: 9612314-9612314 on Build GRCh38
rs956980942 Chr.17: 9612598-9612598 on Build GRCh38
rs139171654 Chr.17: 9612372-9612372 on Build GRCh38
rs747834863 Chr.17: 9612393-9612393 on Build GRCh38
rs747787922 Chr.17: 9612310-9612310 on Build GRCh38
rs201352201 Chr.17: 9612389-9612389 on Build GRCh38
rs1031668856 Chr.17: 9612590-9612590 on Build GRCh38
rs1019836835 Chr.17: 9612492-9612492 on Build GRCh38
rs986860039 Chr.17: 9612255-9612255 on Build GRCh38
rs752058115 Chr.17: 9612438-9612438 on Build GRCh38
rs776337185 Chr.17: 9612306-9612306 on Build GRCh38
rs188788396 Chr.17: 9612551-9612551 on Build GRCh38
rs748964845 Chr.17: 9612459-9612459 on Build GRCh38
rs759502082 Chr.17: 9612420-9612420 on Build GRCh38
rs374020468 Chr.17: 9612362-9612362 on Build GRCh38
rs369634065 Chr.17: 9612349-9612349 on Build GRCh38
rs760499389 Chr.17: 9612429-9612429 on Build GRCh38
rs767415440 Chr.17: 9612350-9612350 on Build GRCh38
rs956908574 Chr.17: 9612254-9612254 on Build GRCh38
rs1047412036 Chr.17: 9612374-9612374 on Build GRCh38
rs544679690 Chr.17: 9612503-9612503 on Build GRCh38
rs768138371 Chr.17: 9612350-9612352 on Build GRCh38
rs769465711 Chr.17: 9612396-9612396 on Build GRCh38
rs149304215 Chr.17: 9612601-9612601 on Build GRCh38
rs73975746 Chr.17: 9612539-9612539 on Build GRCh38
rs146057434 Chr.17: 9612390-9612390 on Build GRCh38
rs952643946 Chr.17: 9612668-9612668 on Build GRCh38
rs747349187 Chr.17: 9612295-9612295 on Build GRCh38
rs754169117 Chr.17: 9612360-9612360 on Build GRCh38
rs771996883 Chr.17: 9612498-9612499 on Build GRCh38
rs772674669 Chr.17: 9612316-9612316 on Build GRCh38
rs774282830 Chr.17: 9612340-9612340 on Build GRCh38
rs774221667 Chr.17: 9612418-9612418 on Build GRCh38
rs1056801078 Chr.17: 9612401-9612401 on Build GRCh38
rs112612589 Chr.17: 9612285-9612285 on Build GRCh38
rs538483197 Chr.17: 9612289-9612289 on Build GRCh38
rs765517839 Chr.17: 9612369-9612369 on Build GRCh38
rs755450527 Chr.17: 9612440-9612440 on Build GRCh38
rs370297079 Chr.17: 9612414-9612414 on Build GRCh38
rs760552565 Chr.17: 9612512-9612512 on Build GRCh38
rs757258446 Chr.17: 9612353-9612353 on Build GRCh38
rs908450836 Chr.17: 9612346-9612346 on Build GRCh38
rs773367785 Chr.17: 9612526-9612526 on Build GRCh38
rs770407483 Chr.17: 9612416-9612416 on Build GRCh38
rs200953170 Chr.17: 9612445-9612445 on Build GRCh38
rs781632763 Chr.17: 9612444-9612444 on Build GRCh38
rs6503235 Chr.17: 9612460-9612460 on Build GRCh38
rs368707227 Chr.17: 9612446-9612446 on Build GRCh38
rs150116794 Chr.17: 9612457-9612457 on Build GRCh38
rs778553088 Chr.17: 9612415-9612416 on Build GRCh38
rs766245608 Chr.17: 9612333-9612333 on Build GRCh38
rs539845945 Chr.17: 9612665-9612665 on Build GRCh38
rs964100405 Chr.17: 9612627-9612627 on Build GRCh38
rs745840181 Chr.17: 9612480-9612480 on Build GRCh38
rs1032552660 Chr.17: 9612437-9612437 on Build GRCh38
rs774470421 Chr.17: 9612421-9612422 on Build GRCh38
rs564399873 Chr.17: 9612604-9612604 on Build GRCh38
rs1019641366 Chr.17: 9612610-9612610 on Build GRCh38
rs181222295 Chr.17: 9612648-9612648 on Build GRCh38
rs370755204 Chr.17: 9612505-9612505 on Build GRCh38
rs767468275 Chr.17: 9612423-9612423 on Build GRCh38
rs778463578 Chr.17: 9612479-9612479 on Build GRCh38
rs149106566 Chr.17: 9612381-9612381 on Build GRCh38
rs370813607 Chr.17: 9612408-9612408 on Build GRCh38
rs765570854 Chr.17: 9612430-9612430 on Build GRCh38
rs745722527 Chr.17: 9612491-9612492 on Build GRCh38
rs750491859 Chr.17: 9612370-9612370 on Build GRCh38
rs766472420 Chr.17: 9612434-9612434 on Build GRCh38
rs1012631419 Chr.17: 9612456-9612456 on Build GRCh38
rs763013883 Chr.17: 9612433-9612433 on Build GRCh38
rs376260961 Chr.17: 9612413-9612413 on Build GRCh38
rs569400249 Chr.17: 9612301-9612301 on Build GRCh38
rs762482958 Chr.17: 9612327-9612327 on Build GRCh38
rs775458752 Chr.17: 9612428-9612428 on Build GRCh38
rs374415605 Chr.17: 9612521-9612521 on Build GRCh38
rs750593072 Chr.17: 9612432-9612432 on Build GRCh38
rs908398784 Chr.17: 9612651-9612651 on Build GRCh38

View Full Product Details