Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9645711-9646214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP43
Entrez Gene ID
124739
Gene Name
ubiquitin specific peptidase 43
Gene Aliases
-
UniGene
Hs.709621
Gene Chromosome Location
Chr.17: 9643493-9729687 on Build GRCh38
Transcripts
NM_001267576.1 NM_153210.4 XM_011523639.2 XM_011523640.1 XM_011523641.1 XM_011523642.2 XM_017024159.1 XM_017024162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5712769 c.298G>T p.A100S Chr.17: 9645930-9645930 on Build GRCh38
6556120 c.443C>T p.A148V Chr.17: 9646075-9646075 on Build GRCh38
6052089 c.387G>A p.L129L Chr.17: 9646019-9646019 on Build GRCh38
5740866 c.353G>A p.C118Y Chr.17: 9645985-9645985 on Build GRCh38
6052090 c.468T>G p.T156T Chr.17: 9646100-9646100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776092372 Chr.17: 9645884-9645884 on Build GRCh38
rs541261453 Chr.17: 9645844-9645844 on Build GRCh38
rs751791413 Chr.17: 9645876-9645876 on Build GRCh38
rs773934217 Chr.17: 9646050-9646050 on Build GRCh38
rs969054628 Chr.17: 9646016-9646016 on Build GRCh38
rs977044741 Chr.17: 9646028-9646028 on Build GRCh38
rs959931811 Chr.17: 9645878-9645878 on Build GRCh38
rs995146207 Chr.17: 9645811-9645811 on Build GRCh38
rs1039977841 Chr.17: 9645810-9645810 on Build GRCh38
rs755107578 Chr.17: 9645902-9645902 on Build GRCh38
rs944285870 Chr.17: 9645733-9645733 on Build GRCh38
rs114382285 Chr.17: 9646043-9646043 on Build GRCh38
rs865778620 Chr.17: 9646033-9646033 on Build GRCh38
rs184688281 Chr.17: 9645874-9645874 on Build GRCh38
rs920154058 Chr.17: 9646147-9646147 on Build GRCh38
rs1022843265 Chr.17: 9646010-9646010 on Build GRCh38
rs115314145 Chr.17: 9646064-9646064 on Build GRCh38
rs756708050 Chr.17: 9645978-9645978 on Build GRCh38
rs891333808 Chr.17: 9645744-9645744 on Build GRCh38
rs775457368 Chr.17: 9645814-9645814 on Build GRCh38
rs958594387 Chr.17: 9645880-9645880 on Build GRCh38
rs778370139 Chr.17: 9646029-9646029 on Build GRCh38
rs988283391 Chr.17: 9646170-9646170 on Build GRCh38
rs1051260667 Chr.17: 9645743-9645743 on Build GRCh38
rs912702235 Chr.17: 9646113-9646113 on Build GRCh38
rs931561385 Chr.17: 9646160-9646160 on Build GRCh38
rs548183484 Chr.17: 9645833-9645833 on Build GRCh38
rs538668559 Chr.17: 9645885-9645885 on Build GRCh38
rs745514629 Chr.17: 9646053-9646053 on Build GRCh38
rs1021519853 Chr.17: 9645923-9645923 on Build GRCh38
rs552978792 Chr.17: 9645857-9645857 on Build GRCh38
rs1002902644 Chr.17: 9646181-9646181 on Build GRCh38
rs3744756 Chr.17: 9646124-9646124 on Build GRCh38
rs530371655 Chr.17: 9645868-9645868 on Build GRCh38
rs757010193 Chr.17: 9645867-9645867 on Build GRCh38
rs942933453 Chr.17: 9645806-9645806 on Build GRCh38
rs775013276 Chr.17: 9646058-9646058 on Build GRCh38
rs1025606679 Chr.17: 9645853-9645853 on Build GRCh38
rs995489477 Chr.17: 9645848-9645848 on Build GRCh38
rs530834310 Chr.17: 9646056-9646056 on Build GRCh38
rs767001982 Chr.17: 9645756-9645756 on Build GRCh38
rs748194066 Chr.17: 9645973-9645973 on Build GRCh38
rs1039990258 Chr.17: 9645801-9645801 on Build GRCh38
rs781387050 Chr.17: 9645929-9645929 on Build GRCh38
rs900087160 Chr.17: 9645809-9645809 on Build GRCh38
rs867911990 Chr.17: 9646116-9646116 on Build GRCh38
rs1047049731 Chr.17: 9646162-9646162 on Build GRCh38
rs556802757 Chr.17: 9646039-9646039 on Build GRCh38
rs755058755 Chr.17: 9645916-9645916 on Build GRCh38
rs1040287967 Chr.17: 9646133-9646133 on Build GRCh38
rs912715396 Chr.17: 9646188-9646188 on Build GRCh38
rs76808429 Chr.17: 9646172-9646172 on Build GRCh38
rs760575803 Chr.17: 9645830-9645830 on Build GRCh38
rs898759754 Chr.17: 9645822-9645822 on Build GRCh38
rs935400497 Chr.17: 9646140-9646140 on Build GRCh38
rs987247507 Chr.17: 9646103-9646103 on Build GRCh38
rs887146309 Chr.17: 9646179-9646179 on Build GRCh38

View Full Product Details