Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10444625-10445134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH4
Entrez Gene ID
4622
Gene Name
myosin heavy chain 4
Gene Aliases
MYH2B, MyHC-2B, MyHC-IIb
UniGene
Hs.711141
Gene Chromosome Location
Chr.17: 10443058-10469559 on Build GRCh38
Transcripts
NM_017533.2 XM_017024676.1 NR_125367.1

Gene Symbol
MYHAS
Entrez Gene ID
100128560
Gene Name
myosin heavy chain gene cluster antisense RNA
Gene Aliases
-
UniGene
Hs.661174
Gene Chromosome Location
Chr.17: 10383132-10625540 on Build GRCh38
Transcripts
NM_017533.2 XM_017024676.1 NR_125367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3712200 c.5319G>A p.L1773L Chr.17: 10445123-10445123 on Build GRCh38
5966780 c.5533C>T p.R1845C Chr.17: 10444833-10444833 on Build GRCh38
6719413 c.5466G>A p.R1822R Chr.17: 10444976-10444976 on Build GRCh38
6523269 c.5483G>T p.S1828I Chr.17: 10444883-10444883 on Build GRCh38
3513706 c.5527G>A p.G1843S Chr.17: 10444839-10444839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015196587 Chr.17: 10445106-10445106 on Build GRCh38
rs375534836 Chr.17: 10444787-10444787 on Build GRCh38
rs150826087 Chr.17: 10444917-10444917 on Build GRCh38
rs766547496 Chr.17: 10444654-10444656 on Build GRCh38
rs763965549 Chr.17: 10444708-10444708 on Build GRCh38
rs755154771 Chr.17: 10445076-10445078 on Build GRCh38
rs375718466 Chr.17: 10445085-10445085 on Build GRCh38
rs773065772 Chr.17: 10444976-10444976 on Build GRCh38
rs79179246 Chr.17: 10444918-10444918 on Build GRCh38
rs755044579 Chr.17: 10445036-10445036 on Build GRCh38
rs773605554 Chr.17: 10445090-10445090 on Build GRCh38
rs185824725 Chr.17: 10444832-10444832 on Build GRCh38
rs138375236 Chr.17: 10444653-10444653 on Build GRCh38
rs572596249 Chr.17: 10444779-10444779 on Build GRCh38
rs750983058 Chr.17: 10444925-10444925 on Build GRCh38
rs2277649 Chr.17: 10445037-10445037 on Build GRCh38
rs372264460 Chr.17: 10444713-10444713 on Build GRCh38
rs554344219 Chr.17: 10444731-10444731 on Build GRCh38
rs754220373 Chr.17: 10444923-10444923 on Build GRCh38
rs867027969 Chr.17: 10444907-10444907 on Build GRCh38
rs766870001 Chr.17: 10445110-10445110 on Build GRCh38
rs759041439 Chr.17: 10445062-10445062 on Build GRCh38
rs927957828 Chr.17: 10445102-10445102 on Build GRCh38
rs878978320 Chr.17: 10444972-10444972 on Build GRCh38
rs773887491 Chr.17: 10444981-10444981 on Build GRCh38
rs370263974 Chr.17: 10444978-10444978 on Build GRCh38
rs200732393 Chr.17: 10445019-10445019 on Build GRCh38
rs973277465 Chr.17: 10444696-10444696 on Build GRCh38
rs776367481 Chr.17: 10444711-10444711 on Build GRCh38
rs893951001 Chr.17: 10444838-10444838 on Build GRCh38
rs148072812 Chr.17: 10444997-10444997 on Build GRCh38
rs779599645 Chr.17: 10444794-10444794 on Build GRCh38
rs760340692 Chr.17: 10444695-10444695 on Build GRCh38
rs1043444570 Chr.17: 10444663-10444669 on Build GRCh38
rs776399558 Chr.17: 10444856-10444856 on Build GRCh38
rs775219097 Chr.17: 10445071-10445071 on Build GRCh38
rs763267052 Chr.17: 10445101-10445101 on Build GRCh38
rs775815764 Chr.17: 10444691-10444691 on Build GRCh38
rs34260986 Chr.17: 10444687-10444687 on Build GRCh38
rs780476111 Chr.17: 10445031-10445031 on Build GRCh38
rs199974986 Chr.17: 10445063-10445063 on Build GRCh38
rs141762664 Chr.17: 10445018-10445018 on Build GRCh38
rs761645152 Chr.17: 10444869-10444869 on Build GRCh38
rs758347372 Chr.17: 10444929-10444929 on Build GRCh38
rs751094422 Chr.17: 10444911-10444911 on Build GRCh38
rs1018529810 Chr.17: 10444888-10444888 on Build GRCh38
rs75173310 Chr.17: 10445058-10445058 on Build GRCh38
rs762214845 Chr.17: 10445016-10445016 on Build GRCh38
rs757811767 Chr.17: 10444792-10444792 on Build GRCh38
rs575593634 Chr.17: 10444962-10444962 on Build GRCh38
rs768902070 Chr.17: 10444950-10444950 on Build GRCh38
rs759003357 Chr.17: 10444993-10444993 on Build GRCh38
rs757933878 Chr.17: 10444774-10444774 on Build GRCh38
rs61521510 Chr.17: 10444833-10444833 on Build GRCh38
rs780179940 Chr.17: 10444947-10444947 on Build GRCh38
rs758931342 Chr.17: 10445027-10445027 on Build GRCh38
rs770067160 Chr.17: 10445044-10445044 on Build GRCh38
rs749814839 Chr.17: 10444669-10444669 on Build GRCh38
rs765916631 Chr.17: 10444908-10444908 on Build GRCh38
rs369333277 Chr.17: 10444956-10444956 on Build GRCh38
rs752423627 Chr.17: 10445012-10445012 on Build GRCh38
rs777527554 Chr.17: 10445045-10445045 on Build GRCh38
rs139900511 Chr.17: 10445043-10445043 on Build GRCh38
rs778191883 Chr.17: 10444788-10444788 on Build GRCh38
rs532245549 Chr.17: 10444948-10444948 on Build GRCh38
rs779517826 Chr.17: 10444924-10444924 on Build GRCh38
rs762312209 Chr.17: 10444896-10444896 on Build GRCh38
rs267604703 Chr.17: 10444875-10444875 on Build GRCh38
rs376179609 Chr.17: 10444971-10444971 on Build GRCh38
rs538270849 Chr.17: 10444686-10444686 on Build GRCh38
rs749913825 Chr.17: 10444738-10444738 on Build GRCh38
rs935396215 Chr.17: 10444830-10444830 on Build GRCh38
rs867629328 Chr.17: 10445003-10445003 on Build GRCh38
rs760403113 Chr.17: 10445073-10445073 on Build GRCh38
rs373271489 Chr.17: 10444958-10444958 on Build GRCh38
rs968999719 Chr.17: 10444991-10444991 on Build GRCh38
rs772220551 Chr.17: 10444805-10444805 on Build GRCh38
rs751751519 Chr.17: 10444964-10444966 on Build GRCh38
rs567649730 Chr.17: 10444675-10444675 on Build GRCh38
rs775519657 Chr.17: 10444810-10444810 on Build GRCh38
rs879202551 Chr.17: 10444972-10444973 on Build GRCh38
rs749725185 Chr.17: 10444790-10444790 on Build GRCh38
rs778082635 Chr.17: 10444664-10444664 on Build GRCh38
rs749083591 Chr.17: 10445047-10445047 on Build GRCh38
rs146221622 Chr.17: 10445088-10445088 on Build GRCh38
rs751627870 Chr.17: 10445034-10445034 on Build GRCh38
rs765129427 Chr.17: 10444873-10444873 on Build GRCh38
rs773154000 Chr.17: 10444887-10444887 on Build GRCh38
rs867319089 Chr.17: 10444766-10444766 on Build GRCh38

View Full Product Details