Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32454415-32454924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD11
Entrez Gene ID
5717
Gene Name
proteasome 26S subunit, non-ATPase 11
Gene Aliases
Rpn6, S9, p44.5
UniGene
Hs.443379
Gene Chromosome Location
Chr.17: 32444463-32483319 on Build GRCh38
Transcripts
NM_001270482.1 NM_002815.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6570028 c.259C>T p.R87C Chr.17: 32454560-32454560 on Build GRCh38
5962995 c.258T>C p.A86A Chr.17: 32454559-32454559 on Build GRCh38
1189440 c.215A>T p.Y72F Chr.17: 32454516-32454516 on Build GRCh38
1479435 c.274C>G p.L92V Chr.17: 32454575-32454575 on Build GRCh38
4484675 c.283C>G p.L95V Chr.17: 32454584-32454584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977621684 Chr.17: 32454862-32454862 on Build GRCh38
rs527395256 Chr.17: 32454543-32454543 on Build GRCh38
rs141232083 Chr.17: 32454878-32454878 on Build GRCh38
rs558157773 Chr.17: 32454586-32454586 on Build GRCh38
rs769410629 Chr.17: 32454880-32454880 on Build GRCh38
rs577597533 Chr.17: 32454610-32454610 on Build GRCh38
rs772006275 Chr.17: 32454529-32454529 on Build GRCh38
rs148245607 Chr.17: 32454819-32454819 on Build GRCh38
rs888227572 Chr.17: 32454706-32454706 on Build GRCh38
rs764608128 Chr.17: 32454624-32454624 on Build GRCh38
rs746985262 Chr.17: 32454744-32454744 on Build GRCh38
rs755266974 Chr.17: 32454467-32454467 on Build GRCh38
rs563817195 Chr.17: 32454866-32454866 on Build GRCh38
rs1005296702 Chr.17: 32454724-32454724 on Build GRCh38
rs569737552 Chr.17: 32454445-32454445 on Build GRCh38
rs758949015 Chr.17: 32454473-32454473 on Build GRCh38
rs915898231 Chr.17: 32454573-32454573 on Build GRCh38
rs753187420 Chr.17: 32454470-32454470 on Build GRCh38
rs993920465 Chr.17: 32454684-32454684 on Build GRCh38
rs369694838 Chr.17: 32454477-32454477 on Build GRCh38
rs779313601 Chr.17: 32454469-32454469 on Build GRCh38
rs771776678 Chr.17: 32454733-32454733 on Build GRCh38
rs753500719 Chr.17: 32454460-32454460 on Build GRCh38
rs926107336 Chr.17: 32454899-32454899 on Build GRCh38
rs773044556 Chr.17: 32454533-32454533 on Build GRCh38
rs770458809 Chr.17: 32454745-32454745 on Build GRCh38
rs373669061 Chr.17: 32454613-32454613 on Build GRCh38
rs749991107 Chr.17: 32454448-32454448 on Build GRCh38
rs756446665 Chr.17: 32454710-32454710 on Build GRCh38
rs957711019 Chr.17: 32454901-32454901 on Build GRCh38
rs776518579 Chr.17: 32454876-32454876 on Build GRCh38
rs768279549 Chr.17: 32454840-32454840 on Build GRCh38
rs534607477 Chr.17: 32454518-32454518 on Build GRCh38
rs774428385 Chr.17: 32454444-32454444 on Build GRCh38
rs920086981 Chr.17: 32454885-32454885 on Build GRCh38
rs773132940 Chr.17: 32454737-32454737 on Build GRCh38
rs747066502 Chr.17: 32454480-32454480 on Build GRCh38
rs776164726 Chr.17: 32454758-32454758 on Build GRCh38
rs752017045 Chr.17: 32454629-32454629 on Build GRCh38
rs753095750 Chr.17: 32454616-32454616 on Build GRCh38
rs747902980 Chr.17: 32454732-32454732 on Build GRCh38
rs201020248 Chr.17: 32454446-32454446 on Build GRCh38
rs555229692 Chr.17: 32454459-32454459 on Build GRCh38
rs758717454 Chr.17: 32454617-32454617 on Build GRCh38
rs760173860 Chr.17: 32454559-32454559 on Build GRCh38
rs891105341 Chr.17: 32454565-32454565 on Build GRCh38
rs376860597 Chr.17: 32454669-32454669 on Build GRCh38
rs765810001 Chr.17: 32454561-32454561 on Build GRCh38
rs908253730 Chr.17: 32454822-32454822 on Build GRCh38
rs746067171 Chr.17: 32454708-32454708 on Build GRCh38
rs780455126 Chr.17: 32454714-32454714 on Build GRCh38
rs199714157 Chr.17: 32454489-32454489 on Build GRCh38
rs757356221 Chr.17: 32454657-32454657 on Build GRCh38
rs768559815 Chr.17: 32454499-32454499 on Build GRCh38
rs746163534 Chr.17: 32454491-32454491 on Build GRCh38
rs149054306 Chr.17: 32454673-32454673 on Build GRCh38
rs566005068 Chr.17: 32454483-32454483 on Build GRCh38
rs774338442 Chr.17: 32454502-32454502 on Build GRCh38
rs143100533 Chr.17: 32454697-32454697 on Build GRCh38

View Full Product Details