Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35118085-35118595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51D
Entrez Gene ID
5892
Gene Name
RAD51 paralog D
Gene Aliases
BROVCA4, R51H3, RAD51L3, TRAD
UniGene
Hs.631757
Gene Chromosome Location
Chr.17: 35099792-35119869 on Build GRCh38
Transcripts
NM_001142571.1 NM_002878.3 NM_133629.2 NR_037711.1 NR_037712.1 NR_037714.1

Gene Symbol
RAD51L3-RFFL
Entrez Gene ID
100529207
Gene Name
RAD51L3-RFFL readthrough
Gene Aliases
-
UniGene
Hs.13680
Gene Chromosome Location
Chr.17: 35009112-35121522 on Build GRCh38
Transcripts
NM_001142571.1 NM_002878.3 NM_133629.2 NR_037711.1 NR_037712.1 NR_037714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889302 c.203G>A p.G68D Chr.17: 35118561-35118561 on Build GRCh38
977817 c.176C>A p.A59D Chr.17: 35118588-35118588 on Build GRCh38
4970541 c.204C>T p.G68G Chr.17: 35118560-35118560 on Build GRCh38
1382401 c.258T>C p.I86I Chr.17: 35118506-35118506 on Build GRCh38
6538713 c.203G>C p.G68A Chr.17: 35118561-35118561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746929682 Chr.17: 35118541-35118541 on Build GRCh38
rs775045445 Chr.17: 35118562-35118562 on Build GRCh38
rs200018296 Chr.17: 35118514-35118514 on Build GRCh38
rs75802835 Chr.17: 35118133-35118133 on Build GRCh38
rs546461804 Chr.17: 35118564-35118564 on Build GRCh38
rs1043618944 Chr.17: 35118221-35118221 on Build GRCh38
rs143533175 Chr.17: 35118249-35118249 on Build GRCh38
rs976833685 Chr.17: 35118442-35118442 on Build GRCh38
rs745546403 Chr.17: 35118551-35118551 on Build GRCh38
rs765555990 Chr.17: 35118475-35118475 on Build GRCh38
rs774973970 Chr.17: 35118329-35118329 on Build GRCh38
rs867217445 Chr.17: 35118389-35118389 on Build GRCh38
rs181672547 Chr.17: 35118416-35118416 on Build GRCh38
rs573193143 Chr.17: 35118405-35118405 on Build GRCh38
rs200995160 Chr.17: 35118451-35118451 on Build GRCh38
rs895800477 Chr.17: 35118253-35118253 on Build GRCh38
rs993973591 Chr.17: 35118347-35118347 on Build GRCh38
rs559850711 Chr.17: 35118552-35118552 on Build GRCh38
rs376616485 Chr.17: 35118569-35118569 on Build GRCh38
rs975512952 Chr.17: 35118217-35118217 on Build GRCh38
rs371182137 Chr.17: 35118574-35118574 on Build GRCh38
rs1044821376 Chr.17: 35118392-35118392 on Build GRCh38
rs923892456 Chr.17: 35118282-35118282 on Build GRCh38
rs964084917 Chr.17: 35118197-35118197 on Build GRCh38
rs74475325 Chr.17: 35118407-35118407 on Build GRCh38
rs777826765 Chr.17: 35118534-35118534 on Build GRCh38
rs200564819 Chr.17: 35118499-35118499 on Build GRCh38
rs767681165 Chr.17: 35118505-35118505 on Build GRCh38
rs567997074 Chr.17: 35118262-35118262 on Build GRCh38
rs552945580 Chr.17: 35118345-35118345 on Build GRCh38
rs370123666 Chr.17: 35118455-35118455 on Build GRCh38
rs876660173 Chr.17: 35118555-35118555 on Build GRCh38
rs1053825968 Chr.17: 35118306-35118306 on Build GRCh38
rs913009139 Chr.17: 35118187-35118187 on Build GRCh38
rs147268486 Chr.17: 35118397-35118398 on Build GRCh38
rs921732527 Chr.17: 35118234-35118234 on Build GRCh38
rs759161611 Chr.17: 35118461-35118461 on Build GRCh38
rs1006745506 Chr.17: 35118413-35118413 on Build GRCh38
rs1026793130 Chr.17: 35118363-35118363 on Build GRCh38
rs786202179 Chr.17: 35118564-35118579 on Build GRCh38
rs376670250 Chr.17: 35118533-35118533 on Build GRCh38
rs1003706569 Chr.17: 35118441-35118441 on Build GRCh38
rs1019993235 Chr.17: 35118445-35118445 on Build GRCh38
rs568922072 Chr.17: 35118131-35118131 on Build GRCh38
rs9901455 Chr.17: 35118530-35118530 on Build GRCh38
rs200810304 Chr.17: 35118566-35118566 on Build GRCh38
rs765387810 Chr.17: 35118395-35118396 on Build GRCh38
rs887322398 Chr.17: 35118492-35118492 on Build GRCh38
rs947780553 Chr.17: 35118339-35118339 on Build GRCh38
rs56026142 Chr.17: 35118568-35118568 on Build GRCh38
rs976528600 Chr.17: 35118144-35118144 on Build GRCh38
rs369946779 Chr.17: 35118547-35118547 on Build GRCh38
rs750714062 Chr.17: 35118509-35118509 on Build GRCh38
rs375054425 Chr.17: 35118350-35118350 on Build GRCh38
rs56218020 Chr.17: 35118494-35118494 on Build GRCh38
rs533874119 Chr.17: 35118291-35118291 on Build GRCh38
rs79372946 Chr.17: 35118412-35118412 on Build GRCh38
rs764351040 Chr.17: 35118560-35118560 on Build GRCh38
rs763380219 Chr.17: 35118481-35118481 on Build GRCh38
rs148690585 Chr.17: 35118548-35118548 on Build GRCh38
rs766692136 Chr.17: 35118571-35118571 on Build GRCh38
rs753052289 Chr.17: 35118479-35118479 on Build GRCh38
rs587782063 Chr.17: 35118508-35118508 on Build GRCh38
rs746889357 Chr.17: 35118475-35118477 on Build GRCh38
rs763439048 Chr.17: 35118559-35118559 on Build GRCh38
rs757467813 Chr.17: 35118501-35118501 on Build GRCh38
rs545137563 Chr.17: 35118408-35118408 on Build GRCh38
rs876658332 Chr.17: 35118525-35118525 on Build GRCh38
rs142189122 Chr.17: 35118556-35118556 on Build GRCh38
rs778585939 Chr.17: 35118518-35118518 on Build GRCh38
rs760246764 Chr.17: 35118454-35118454 on Build GRCh38
rs933167419 Chr.17: 35118236-35118236 on Build GRCh38
rs776316890 Chr.17: 35118459-35118459 on Build GRCh38
rs777402267 Chr.17: 35118577-35118577 on Build GRCh38

View Full Product Details