Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38898324-38898823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LASP1
Entrez Gene ID
3927
Gene Name
LIM and SH3 protein 1
Gene Aliases
Lasp-1, MLN50
UniGene
Hs.741156
Gene Chromosome Location
Chr.17: 38869859-38921770 on Build GRCh38
Transcripts
NM_001271608.1 NM_006148.3 NR_073384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6576110 c.309C>A p.D103E Chr.17: 38898471-38898471 on Build GRCh38
436473 c.326G>A p.R109K Chr.17: 38898488-38898488 on Build GRCh38
5441617 c.271G>A p.E91K Chr.17: 38898433-38898433 on Build GRCh38
4808210 c.349A>T p.I117F Chr.17: 38898511-38898511 on Build GRCh38
5783025 c.334A>G p.K112E Chr.17: 38898496-38898496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749677702 Chr.17: 38898721-38898721 on Build GRCh38
rs752517092 Chr.17: 38898627-38898627 on Build GRCh38
rs778846955 Chr.17: 38898646-38898646 on Build GRCh38
rs867124374 Chr.17: 38898640-38898640 on Build GRCh38
rs969795148 Chr.17: 38898738-38898738 on Build GRCh38
rs1038352471 Chr.17: 38898699-38898699 on Build GRCh38
rs201616839 Chr.17: 38898789-38898791 on Build GRCh38
rs914506119 Chr.17: 38898563-38898563 on Build GRCh38
rs973332221 Chr.17: 38898393-38898393 on Build GRCh38
rs546857959 Chr.17: 38898574-38898574 on Build GRCh38
rs767559321 Chr.17: 38898571-38898571 on Build GRCh38
rs143184732 Chr.17: 38898707-38898707 on Build GRCh38
rs1053392231 Chr.17: 38898602-38898602 on Build GRCh38
rs139844955 Chr.17: 38898669-38898669 on Build GRCh38
rs756934383 Chr.17: 38898696-38898696 on Build GRCh38
rs767599931 Chr.17: 38898371-38898371 on Build GRCh38
rs780162081 Chr.17: 38898473-38898473 on Build GRCh38
rs769199613 Chr.17: 38898724-38898724 on Build GRCh38
rs1026249803 Chr.17: 38898709-38898709 on Build GRCh38
rs962987675 Chr.17: 38898392-38898392 on Build GRCh38
rs774377425 Chr.17: 38898544-38898544 on Build GRCh38
rs898769724 Chr.17: 38898706-38898706 on Build GRCh38
rs376885755 Chr.17: 38898616-38898616 on Build GRCh38
rs778720031 Chr.17: 38898363-38898363 on Build GRCh38
rs533944880 Chr.17: 38898755-38898755 on Build GRCh38
rs769032251 Chr.17: 38898542-38898542 on Build GRCh38
rs999729187 Chr.17: 38898505-38898505 on Build GRCh38
rs532607531 Chr.17: 38898398-38898398 on Build GRCh38
rs1008771018 Chr.17: 38898357-38898357 on Build GRCh38
rs990094884 Chr.17: 38898559-38898559 on Build GRCh38
rs868550362 Chr.17: 38898547-38898547 on Build GRCh38
rs556876833 Chr.17: 38898615-38898615 on Build GRCh38
rs780669382 Chr.17: 38898714-38898714 on Build GRCh38
rs1048604502 Chr.17: 38898648-38898648 on Build GRCh38
rs758066378 Chr.17: 38898382-38898382 on Build GRCh38
rs567922284 Chr.17: 38898591-38898591 on Build GRCh38
rs1041383185 Chr.17: 38898592-38898592 on Build GRCh38
rs112191648 Chr.17: 38898369-38898369 on Build GRCh38
rs935915564 Chr.17: 38898555-38898555 on Build GRCh38
rs746536575 Chr.17: 38898404-38898404 on Build GRCh38
rs980199906 Chr.17: 38898769-38898769 on Build GRCh38
rs1004237588 Chr.17: 38898440-38898440 on Build GRCh38
rs750355011 Chr.17: 38898682-38898682 on Build GRCh38
rs770292660 Chr.17: 38898477-38898477 on Build GRCh38
rs544545913 Chr.17: 38898347-38898347 on Build GRCh38
rs113209138 Chr.17: 38898523-38898523 on Build GRCh38
rs2286883 Chr.17: 38898631-38898631 on Build GRCh38
rs78076914 Chr.17: 38898713-38898713 on Build GRCh38
rs1039884749 Chr.17: 38898368-38898368 on Build GRCh38
rs958496966 Chr.17: 38898732-38898735 on Build GRCh38
rs538159756 Chr.17: 38898609-38898609 on Build GRCh38
rs753638417 Chr.17: 38898546-38898546 on Build GRCh38
rs1025795967 Chr.17: 38898406-38898406 on Build GRCh38
rs780611002 Chr.17: 38898702-38898702 on Build GRCh38
rs555272469 Chr.17: 38898787-38898787 on Build GRCh38
rs146454293 Chr.17: 38898534-38898534 on Build GRCh38
rs1130638 Chr.17: 38898519-38898519 on Build GRCh38
rs763824278 Chr.17: 38898632-38898632 on Build GRCh38
rs746433445 Chr.17: 38898525-38898525 on Build GRCh38
rs777248488 Chr.17: 38898400-38898400 on Build GRCh38
rs115706870 Chr.17: 38898532-38898532 on Build GRCh38
rs549181618 Chr.17: 38898561-38898561 on Build GRCh38
rs774772576 Chr.17: 38898774-38898774 on Build GRCh38
rs200496433 Chr.17: 38898541-38898541 on Build GRCh38
rs914431763 Chr.17: 38898747-38898747 on Build GRCh38
rs950459854 Chr.17: 38898415-38898415 on Build GRCh38

View Full Product Details