Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 66211941-66212370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOH
Entrez Gene ID
350
Gene Name
apolipoprotein H
Gene Aliases
B2G1, B2GP1, BG
UniGene
Hs.445358
Gene Chromosome Location
Chr.17: 66212029-66229438 on Build GRCh38
Transcripts
NM_000042.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6532535 c.1035C>G p.C345W Chr.17: 66212136-66212136 on Build GRCh38
5434672 c.994C>G p.L332V Chr.17: 66212177-66212177 on Build GRCh38
259472 c.1005G>A p.W335* Chr.17: 66212166-66212166 on Build GRCh38
3755757 c.1004G>C p.W335S Chr.17: 66212167-66212167 on Build GRCh38
6147558 c.1011T>G p.T337T Chr.17: 66212160-66212160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932841813 Chr.17: 66212042-66212042 on Build GRCh38
rs780480286 Chr.17: 66212105-66212105 on Build GRCh38
rs12943614 Chr.17: 66212281-66212281 on Build GRCh38
rs750997155 Chr.17: 66212091-66212091 on Build GRCh38
rs902965612 Chr.17: 66212305-66212305 on Build GRCh38
rs751991480 Chr.17: 66212189-66212189 on Build GRCh38
rs1012505224 Chr.17: 66212163-66212163 on Build GRCh38
rs973343590 Chr.17: 66212222-66212222 on Build GRCh38
rs1015672775 Chr.17: 66212106-66212106 on Build GRCh38
rs756930948 Chr.17: 66212209-66212213 on Build GRCh38
rs113813266 Chr.17: 66212028-66212028 on Build GRCh38
rs750768530 Chr.17: 66212321-66212321 on Build GRCh38
rs778750307 Chr.17: 66212213-66212214 on Build GRCh38
rs1013941331 Chr.17: 66212263-66212263 on Build GRCh38
rs920547287 Chr.17: 66212061-66212061 on Build GRCh38
rs576409449 Chr.17: 66212280-66212280 on Build GRCh38
rs1801690 Chr.17: 66212167-66212167 on Build GRCh38
rs771800586 Chr.17: 66212136-66212136 on Build GRCh38
rs377164737 Chr.17: 66212225-66212225 on Build GRCh38
rs568079868 Chr.17: 66212192-66212192 on Build GRCh38
rs190137879 Chr.17: 66212111-66212111 on Build GRCh38
rs8178951 Chr.17: 66212071-66212071 on Build GRCh38
rs748403358 Chr.17: 66212204-66212204 on Build GRCh38
rs902827652 Chr.17: 66212132-66212132 on Build GRCh38
rs8178950 Chr.17: 66212221-66212221 on Build GRCh38
rs8178949 Chr.17: 66212275-66212275 on Build GRCh38
rs935741938 Chr.17: 66212312-66212312 on Build GRCh38
rs369915117 Chr.17: 66212161-66212161 on Build GRCh38
rs752992912 Chr.17: 66212036-66212036 on Build GRCh38
rs763848928 Chr.17: 66212151-66212151 on Build GRCh38
rs911700469 Chr.17: 66212274-66212274 on Build GRCh38
rs758767603 Chr.17: 66212095-66212095 on Build GRCh38
rs200265067 Chr.17: 66212191-66212191 on Build GRCh38
rs769883181 Chr.17: 66212128-66212128 on Build GRCh38
rs541084675 Chr.17: 66212087-66212087 on Build GRCh38
rs764842797 Chr.17: 66212157-66212157 on Build GRCh38
rs113041144 Chr.17: 66212031-66212031 on Build GRCh38
rs772114469 Chr.17: 66212214-66212214 on Build GRCh38
rs562833065 Chr.17: 66212133-66212133 on Build GRCh38
rs775417167 Chr.17: 66212139-66212139 on Build GRCh38
rs774235022 Chr.17: 66212130-66212130 on Build GRCh38
rs748272877 Chr.17: 66212114-66212114 on Build GRCh38
rs113474683 Chr.17: 66212190-66212190 on Build GRCh38
rs369364551 Chr.17: 66212016-66212016 on Build GRCh38
rs896790610 Chr.17: 66212256-66212256 on Build GRCh38
rs201802172 Chr.17: 66212206-66212206 on Build GRCh38
rs8178861 Chr.17: 66212228-66212228 on Build GRCh38
rs780277635 Chr.17: 66212173-66212173 on Build GRCh38
rs112890054 Chr.17: 66212032-66212032 on Build GRCh38
rs777848115 Chr.17: 66212211-66212211 on Build GRCh38
rs753267983 Chr.17: 66212154-66212154 on Build GRCh38
rs1558359 Chr.17: 66212278-66212278 on Build GRCh38
rs199836919 Chr.17: 66212198-66212198 on Build GRCh38
rs771648761 Chr.17: 66212174-66212174 on Build GRCh38
rs373238657 Chr.17: 66211982-66211982 on Build GRCh38
rs745795914 Chr.17: 66212213-66212213 on Build GRCh38
rs957254163 Chr.17: 66212313-66212313 on Build GRCh38
rs199553389 Chr.17: 66212120-66212120 on Build GRCh38
rs986024149 Chr.17: 66212249-66212249 on Build GRCh38
rs1011391748 Chr.17: 66212331-66212331 on Build GRCh38
rs535705632 Chr.17: 66212282-66212282 on Build GRCh38
rs371044477 Chr.17: 66212155-66212155 on Build GRCh38
rs940726390 Chr.17: 66212002-66212002 on Build GRCh38
rs1041496447 Chr.17: 66212126-66212126 on Build GRCh38
rs775116700 Chr.17: 66212220-66212220 on Build GRCh38
rs760240363 Chr.17: 66212144-66212144 on Build GRCh38
rs569012373 Chr.17: 66212244-66212244 on Build GRCh38
rs6933 Chr.17: 66212112-66212112 on Build GRCh38
rs561622404 Chr.17: 66212030-66212030 on Build GRCh38
rs572178041 Chr.17: 66212076-66212076 on Build GRCh38

View Full Product Details