Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68537438-68537943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20A
Entrez Gene ID
54757
Gene Name
family with sequence similarity 20 member A
Gene Aliases
AI1G, AIGFS, FP2747
UniGene
Hs.268874
Gene Chromosome Location
Chr.17: 68534561-68600954 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Gene Symbol
PRKAR1A
Entrez Gene ID
5573
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Gene Aliases
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1
UniGene
Hs.280342
Gene Chromosome Location
Chr.17: 68413623-68551316 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4975632 c.1450G>A p.D484N Chr.17: 68537653-68537653 on Build GRCh38
5656729 c.1496A>T p.D499V Chr.17: 68537607-68537607 on Build GRCh38
4963147 c.1572C>T p.D524D Chr.17: 68537531-68537531 on Build GRCh38
4431336 c.1527G>A p.V509V Chr.17: 68537576-68537576 on Build GRCh38
983430 c.1422C>T p.S474S Chr.17: 68537681-68537681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369157703 Chr.17: 68537538-68537538 on Build GRCh38
rs774852696 Chr.17: 68537765-68537765 on Build GRCh38
rs1051754649 Chr.17: 68537624-68537624 on Build GRCh38
rs762483955 Chr.17: 68537534-68537534 on Build GRCh38
rs981770103 Chr.17: 68537836-68537836 on Build GRCh38
rs777416324 Chr.17: 68537604-68537604 on Build GRCh38
rs1017808414 Chr.17: 68537704-68537704 on Build GRCh38
rs995104417 Chr.17: 68537721-68537721 on Build GRCh38
rs762117621 Chr.17: 68537788-68537788 on Build GRCh38
rs777626642 Chr.17: 68537706-68537706 on Build GRCh38
rs769671047 Chr.17: 68537601-68537601 on Build GRCh38
rs984185355 Chr.17: 68537896-68537896 on Build GRCh38
rs142137059 Chr.17: 68537555-68537555 on Build GRCh38
rs747067542 Chr.17: 68537557-68537557 on Build GRCh38
rs190227580 Chr.17: 68537617-68537617 on Build GRCh38
rs776996642 Chr.17: 68537787-68537787 on Build GRCh38
rs758433003 Chr.17: 68537689-68537689 on Build GRCh38
rs749033941 Chr.17: 68537716-68537716 on Build GRCh38
rs146689929 Chr.17: 68537592-68537592 on Build GRCh38
rs2907373 Chr.17: 68537514-68537514 on Build GRCh38
rs750525198 Chr.17: 68537686-68537686 on Build GRCh38
rs770729143 Chr.17: 68537719-68537719 on Build GRCh38
rs145258230 Chr.17: 68537722-68537722 on Build GRCh38
rs772882502 Chr.17: 68537533-68537533 on Build GRCh38
rs914185104 Chr.17: 68537462-68537462 on Build GRCh38
rs768982348 Chr.17: 68537784-68537784 on Build GRCh38
rs754141585 Chr.17: 68537569-68537569 on Build GRCh38
rs766042943 Chr.17: 68537536-68537536 on Build GRCh38
rs779968960 Chr.17: 68537691-68537691 on Build GRCh38
rs374740151 Chr.17: 68537472-68537472 on Build GRCh38
rs747009042 Chr.17: 68537510-68537510 on Build GRCh38
rs182816928 Chr.17: 68537631-68537631 on Build GRCh38
rs764331035 Chr.17: 68537565-68537565 on Build GRCh38
rs772483017 Chr.17: 68537670-68537670 on Build GRCh38
rs756997361 Chr.17: 68537587-68537587 on Build GRCh38
rs139620139 Chr.17: 68537671-68537671 on Build GRCh38
rs370179954 Chr.17: 68537501-68537501 on Build GRCh38
rs777997757 Chr.17: 68537499-68537499 on Build GRCh38
rs987829743 Chr.17: 68537814-68537814 on Build GRCh38
rs755526466 Chr.17: 68537477-68537477 on Build GRCh38
rs766899260 Chr.17: 68537461-68537462 on Build GRCh38
rs959151816 Chr.17: 68537845-68537845 on Build GRCh38
rs781046485 Chr.17: 68537783-68537783 on Build GRCh38
rs565340713 Chr.17: 68537596-68537596 on Build GRCh38
rs149451535 Chr.17: 68537488-68537488 on Build GRCh38
rs745483908 Chr.17: 68537725-68537725 on Build GRCh38
rs750439786 Chr.17: 68537591-68537591 on Build GRCh38
rs140248708 Chr.17: 68537815-68537815 on Build GRCh38
rs771658536 Chr.17: 68537729-68537729 on Build GRCh38
rs773992729 Chr.17: 68537546-68537546 on Build GRCh38
rs143893081 Chr.17: 68537493-68537493 on Build GRCh38
rs762205955 Chr.17: 68537656-68537656 on Build GRCh38
rs191443147 Chr.17: 68537705-68537705 on Build GRCh38
rs761314241 Chr.17: 68537777-68537777 on Build GRCh38
rs765838425 Chr.17: 68537689-68537699 on Build GRCh38
rs765389194 Chr.17: 68537683-68537683 on Build GRCh38
rs768580756 Chr.17: 68537513-68537513 on Build GRCh38
rs373457690 Chr.17: 68537589-68537589 on Build GRCh38
rs754152049 Chr.17: 68537672-68537672 on Build GRCh38
rs534446744 Chr.17: 68537901-68537901 on Build GRCh38
rs750897470 Chr.17: 68537744-68537745 on Build GRCh38
rs757459094 Chr.17: 68537576-68537576 on Build GRCh38
rs752731065 Chr.17: 68537701-68537701 on Build GRCh38
rs1036227169 Chr.17: 68537558-68537558 on Build GRCh38
rs551965708 Chr.17: 68537562-68537562 on Build GRCh38
rs781268933 Chr.17: 68537760-68537760 on Build GRCh38
rs569571447 Chr.17: 68537795-68537795 on Build GRCh38
rs756300365 Chr.17: 68537496-68537496 on Build GRCh38
rs965691431 Chr.17: 68537843-68537843 on Build GRCh38
rs944183650 Chr.17: 68537467-68537467 on Build GRCh38
rs770641013 Chr.17: 68537610-68537610 on Build GRCh38
rs1041254326 Chr.17: 68537497-68537497 on Build GRCh38
rs747920001 Chr.17: 68537530-68537530 on Build GRCh38
rs950399055 Chr.17: 68537762-68537762 on Build GRCh38
rs760125148 Chr.17: 68537551-68537551 on Build GRCh38
rs932702227 Chr.17: 68537854-68537854 on Build GRCh38
rs752291813 Chr.17: 68537502-68537508 on Build GRCh38
rs747927473 Chr.17: 68537593-68537593 on Build GRCh38
rs780208759 Chr.17: 68537531-68537531 on Build GRCh38
rs757364783 Chr.17: 68537509-68537509 on Build GRCh38
rs2286556 Chr.17: 68537838-68537838 on Build GRCh38
rs12952571 Chr.17: 68537476-68537476 on Build GRCh38
rs567050051 Chr.17: 68537852-68537852 on Build GRCh38
rs761058695 Chr.17: 68537559-68537559 on Build GRCh38

View Full Product Details