Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70108116-70108649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ16
Entrez Gene ID
3773
Gene Name
potassium voltage-gated channel subfamily J member 16
Gene Aliases
BIR9, KIR5.1
UniGene
Hs.463985
Gene Chromosome Location
Chr.17: 70075225-70135608 on Build GRCh38
Transcripts
NM_001270422.1 NM_001291622.1 NM_001291623.1 NM_001291624.1 NM_001291625.1 NM_018658.2 NM_170741.2 NM_170742.2 XM_005257337.4 XM_006721885.3 XM_006721886.3 XM_006721887.3 XM_011524781.2 XM_017024609.1 XM_017024610.1 XM_017024611.1 XM_017024612.1 XM_017024613.1

SNP Detail

SNP ID Chromosome Location
rs983399709 Chr.17: 70108623-70108623 on Build GRCh38
rs1027703749 Chr.17: 70108582-70108582 on Build GRCh38
rs138347279 Chr.17: 70108514-70108514 on Build GRCh38
rs987242416 Chr.17: 70108363-70108363 on Build GRCh38
rs999806799 Chr.17: 70108532-70108532 on Build GRCh38
rs539938313 Chr.17: 70108416-70108416 on Build GRCh38
rs967335694 Chr.17: 70108518-70108518 on Build GRCh38
rs1031322585 Chr.17: 70108541-70108541 on Build GRCh38
rs973623666 Chr.17: 70108609-70108609 on Build GRCh38
rs773972159 Chr.17: 70108187-70108187 on Build GRCh38
rs966987009 Chr.17: 70108330-70108330 on Build GRCh38
rs973030935 Chr.17: 70108221-70108221 on Build GRCh38
rs751675472 Chr.17: 70108595-70108595 on Build GRCh38
rs950421430 Chr.17: 70108586-70108586 on Build GRCh38
rs954401447 Chr.17: 70108543-70108543 on Build GRCh38
rs10647082 Chr.17: 70108469-70108470 on Build GRCh38
rs189139983 Chr.17: 70108313-70108313 on Build GRCh38
rs890409900 Chr.17: 70108451-70108451 on Build GRCh38
rs999145848 Chr.17: 70108408-70108408 on Build GRCh38
rs1026278689 Chr.17: 70108289-70108289 on Build GRCh38
rs376498608 Chr.17: 70108489-70108489 on Build GRCh38
rs933043194 Chr.17: 70108362-70108362 on Build GRCh38
rs895238692 Chr.17: 70108492-70108492 on Build GRCh38
rs924286028 Chr.17: 70108352-70108352 on Build GRCh38
rs964465352 Chr.17: 70108153-70108153 on Build GRCh38
rs1003565984 Chr.17: 70108444-70108444 on Build GRCh38
rs41282057 Chr.17: 70108387-70108387 on Build GRCh38
rs758430783 Chr.17: 70108480-70108480 on Build GRCh38
rs929260931 Chr.17: 70108404-70108404 on Build GRCh38
rs534018930 Chr.17: 70108267-70108267 on Build GRCh38
rs906605520 Chr.17: 70108296-70108296 on Build GRCh38
rs575741997 Chr.17: 70108329-70108329 on Build GRCh38
rs566924741 Chr.17: 70108257-70108257 on Build GRCh38
rs747856013 Chr.17: 70108227-70108227 on Build GRCh38
rs953051861 Chr.17: 70108266-70108266 on Build GRCh38
rs539590462 Chr.17: 70108367-70108367 on Build GRCh38
rs35293762 Chr.17: 70108468-70108469 on Build GRCh38
rs7217507 Chr.17: 70108499-70108499 on Build GRCh38
rs532900977 Chr.17: 70108306-70108306 on Build GRCh38
rs150291981 Chr.17: 70108244-70108244 on Build GRCh38
rs3760392 Chr.17: 70108435-70108435 on Build GRCh38
rs772670993 Chr.17: 70108309-70108309 on Build GRCh38
rs184891103 Chr.17: 70108589-70108589 on Build GRCh38
rs920468465 Chr.17: 70108397-70108397 on Build GRCh38
rs1047694405 Chr.17: 70108427-70108427 on Build GRCh38
rs557153694 Chr.17: 70108314-70108314 on Build GRCh38
rs181181271 Chr.17: 70108373-70108373 on Build GRCh38
rs535786018 Chr.17: 70108324-70108324 on Build GRCh38
rs750912056 Chr.17: 70108176-70108176 on Build GRCh38
rs987211662 Chr.17: 70108557-70108557 on Build GRCh38
rs910260199 Chr.17: 70108583-70108583 on Build GRCh38
rs533525611 Chr.17: 70108206-70108206 on Build GRCh38
rs568960601 Chr.17: 70108337-70108337 on Build GRCh38

View Full Product Details