Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70176311-70176813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ2
Entrez Gene ID
3759
Gene Name
potassium voltage-gated channel subfamily J member 2
Gene Aliases
ATFB9, HHBIRK1, HHIRK1, IRK1, KIR2.1, LQT7, SQT3
UniGene
Hs.1547
Gene Chromosome Location
Chr.17: 70169535-70180044 on Build GRCh38
Transcripts
NM_000891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1290751 c.1275G>A p.S425S Chr.17: 70176314-70176314 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965985845 Chr.17: 70176418-70176418 on Build GRCh38
rs751951590 Chr.17: 70176751-70176751 on Build GRCh38
rs767742256 Chr.17: 70176360-70176360 on Build GRCh38
rs776522922 Chr.17: 70176343-70176343 on Build GRCh38
rs757826584 Chr.17: 70176370-70176371 on Build GRCh38
rs750382314 Chr.17: 70176350-70176350 on Build GRCh38
rs894850986 Chr.17: 70176393-70176393 on Build GRCh38
rs1010596977 Chr.17: 70176564-70176565 on Build GRCh38
rs920525736 Chr.17: 70176464-70176464 on Build GRCh38
rs954951923 Chr.17: 70176433-70176433 on Build GRCh38
rs548331448 Chr.17: 70176556-70176556 on Build GRCh38
rs891646921 Chr.17: 70176389-70176389 on Build GRCh38
rs929262599 Chr.17: 70176498-70176498 on Build GRCh38
rs996108433 Chr.17: 70176730-70176730 on Build GRCh38
rs984911196 Chr.17: 70176457-70176457 on Build GRCh38
rs1008903280 Chr.17: 70176676-70176676 on Build GRCh38
rs755287950 Chr.17: 70176565-70176569 on Build GRCh38
rs886053326 Chr.17: 70176536-70176536 on Build GRCh38
rs779473102 Chr.17: 70176375-70176375 on Build GRCh38
rs55970278 Chr.17: 70176402-70176402 on Build GRCh38
rs901658982 Chr.17: 70176722-70176722 on Build GRCh38
rs557213178 Chr.17: 70176760-70176761 on Build GRCh38
rs773647270 Chr.17: 70176416-70176416 on Build GRCh38
rs891506685 Chr.17: 70176600-70176600 on Build GRCh38
rs1028890335 Chr.17: 70176732-70176732 on Build GRCh38
rs954878829 Chr.17: 70176613-70176613 on Build GRCh38
rs999412390 Chr.17: 70176430-70176430 on Build GRCh38
rs786205132 Chr.17: 70176341-70176341 on Build GRCh38
rs886053327 Chr.17: 70176647-70176647 on Build GRCh38
rs964690324 Chr.17: 70176447-70176447 on Build GRCh38
rs973357736 Chr.17: 70176748-70176748 on Build GRCh38
rs935644030 Chr.17: 70176536-70176536 on Build GRCh38
rs188588568 Chr.17: 70176431-70176431 on Build GRCh38
rs1040270841 Chr.17: 70176661-70176661 on Build GRCh38
rs948297602 Chr.17: 70176340-70176340 on Build GRCh38
rs886053325 Chr.17: 70176447-70176447 on Build GRCh38
rs918080435 Chr.17: 70176522-70176522 on Build GRCh38
rs775188814 Chr.17: 70176456-70176456 on Build GRCh38
rs1029021700 Chr.17: 70176448-70176448 on Build GRCh38
rs886053324 Chr.17: 70176445-70176445 on Build GRCh38
rs560147745 Chr.17: 70176527-70176527 on Build GRCh38
rs372559719 Chr.17: 70176764-70176765 on Build GRCh38
rs73998781 Chr.17: 70176459-70176459 on Build GRCh38
rs570173316 Chr.17: 70176645-70176645 on Build GRCh38
rs904761645 Chr.17: 70176581-70176581 on Build GRCh38
rs142660363 Chr.17: 70176609-70176609 on Build GRCh38
rs774436667 Chr.17: 70176345-70176345 on Build GRCh38
rs148485313 Chr.17: 70176587-70176587 on Build GRCh38
rs72552029 Chr.17: 70176358-70176358 on Build GRCh38
rs544826396 Chr.17: 70176524-70176524 on Build GRCh38
rs1022310324 Chr.17: 70176413-70176419 on Build GRCh38
rs568408913 Chr.17: 70176399-70176399 on Build GRCh38
rs761297050 Chr.17: 70176453-70176453 on Build GRCh38
rs1017656309 Chr.17: 70176682-70176682 on Build GRCh38
rs987555677 Chr.17: 70176438-70176438 on Build GRCh38
rs766945050 Chr.17: 70176454-70176454 on Build GRCh38
rs757621970 Chr.17: 70176369-70176369 on Build GRCh38
rs1054786098 Chr.17: 70176384-70176384 on Build GRCh38
rs999152921 Chr.17: 70176591-70176591 on Build GRCh38
rs12947476 Chr.17: 70176378-70176378 on Build GRCh38
rs1054216620 Chr.17: 70176585-70176585 on Build GRCh38
rs553864556 Chr.17: 70176363-70176363 on Build GRCh38
rs749666786 Chr.17: 70176366-70176366 on Build GRCh38
rs765230767 Chr.17: 70176346-70176346 on Build GRCh38
rs910513865 Chr.17: 70176445-70176445 on Build GRCh38
rs968892189 Chr.17: 70176417-70176417 on Build GRCh38
rs56194813 Chr.17: 70176534-70176534 on Build GRCh38
rs146282466 Chr.17: 70176533-70176533 on Build GRCh38
rs962243222 Chr.17: 70176474-70176474 on Build GRCh38
rs758970776 Chr.17: 70176508-70176508 on Build GRCh38

View Full Product Details