Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73253844-73254345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF4L
Entrez Gene ID
642843
Gene Name
cleavage and polyadenylation specific factor 4 like
Gene Aliases
-
UniGene
Hs.534707
Gene Chromosome Location
Chr.17: 73242343-73263778 on Build GRCh38
Transcripts
NM_001129885.1 XM_011525115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944073 c.186G>C p.W62C Chr.17: 73253956-73253956 on Build GRCh38
1385691 c.350C>T p.P117L Chr.17: 73253984-73253984 on Build GRCh38
237241 c.419C>T p.A140V Chr.17: 73253915-73253915 on Build GRCh38
1385690 c.158C>T p.P53L Chr.17: 73253984-73253984 on Build GRCh38
4488965 c.340C>T p.H114Y Chr.17: 73253994-73253994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018382783 Chr.17: 73254242-73254242 on Build GRCh38
rs1038141718 Chr.17: 73253950-73253950 on Build GRCh38
rs189371912 Chr.17: 73253912-73253912 on Build GRCh38
rs965373708 Chr.17: 73253891-73253891 on Build GRCh38
rs182020766 Chr.17: 73253960-73253960 on Build GRCh38
rs1012192806 Chr.17: 73254041-73254042 on Build GRCh38
rs138120513 Chr.17: 73254165-73254165 on Build GRCh38
rs367548246 Chr.17: 73253949-73253949 on Build GRCh38
rs923470181 Chr.17: 73253903-73253903 on Build GRCh38
rs879010248 Chr.17: 73253887-73253887 on Build GRCh38
rs145649040 Chr.17: 73254161-73254161 on Build GRCh38
rs1047820940 Chr.17: 73253916-73253916 on Build GRCh38
rs927884184 Chr.17: 73254046-73254046 on Build GRCh38
rs994626826 Chr.17: 73254314-73254314 on Build GRCh38
rs954878932 Chr.17: 73254285-73254285 on Build GRCh38
rs545671735 Chr.17: 73254048-73254048 on Build GRCh38
rs531147925 Chr.17: 73254305-73254305 on Build GRCh38
rs1051180328 Chr.17: 73254213-73254213 on Build GRCh38
rs374908311 Chr.17: 73254093-73254093 on Build GRCh38
rs540248257 Chr.17: 73254132-73254132 on Build GRCh38
rs374373984 Chr.17: 73253932-73253932 on Build GRCh38
rs1040173436 Chr.17: 73254152-73254152 on Build GRCh38
rs11077676 Chr.17: 73254268-73254268 on Build GRCh38
rs536834125 Chr.17: 73254296-73254296 on Build GRCh38
rs1046330935 Chr.17: 73254068-73254068 on Build GRCh38
rs951601384 Chr.17: 73253922-73253922 on Build GRCh38
rs370524848 Chr.17: 73253931-73253931 on Build GRCh38
rs946860218 Chr.17: 73254147-73254147 on Build GRCh38
rs769696298 Chr.17: 73254003-73254003 on Build GRCh38
rs547454365 Chr.17: 73254136-73254136 on Build GRCh38
rs963734210 Chr.17: 73254267-73254267 on Build GRCh38
rs1014576121 Chr.17: 73254289-73254289 on Build GRCh38
rs574410366 Chr.17: 73253865-73253865 on Build GRCh38
rs763036295 Chr.17: 73254037-73254037 on Build GRCh38
rs899995888 Chr.17: 73253899-73253899 on Build GRCh38
rs189559737 Chr.17: 73254273-73254273 on Build GRCh38
rs934174060 Chr.17: 73254210-73254210 on Build GRCh38
rs973560010 Chr.17: 73254002-73254002 on Build GRCh38
rs564211204 Chr.17: 73254075-73254075 on Build GRCh38
rs774594187 Chr.17: 73254071-73254071 on Build GRCh38
rs772041573 Chr.17: 73253947-73253947 on Build GRCh38
rs935197335 Chr.17: 73254103-73254103 on Build GRCh38
rs981914750 Chr.17: 73254027-73254027 on Build GRCh38
rs370545970 Chr.17: 73253870-73253870 on Build GRCh38
rs1014502167 Chr.17: 73253953-73253953 on Build GRCh38
rs898809147 Chr.17: 73254291-73254291 on Build GRCh38
rs1023821010 Chr.17: 73254324-73254324 on Build GRCh38
rs769022504 Chr.17: 73254295-73254295 on Build GRCh38
rs906375978 Chr.17: 73254092-73254092 on Build GRCh38
rs755699974 Chr.17: 73254126-73254126 on Build GRCh38
rs868770841 Chr.17: 73253970-73253970 on Build GRCh38
rs114003900 Chr.17: 73254004-73254004 on Build GRCh38
rs998470835 Chr.17: 73253902-73253902 on Build GRCh38
rs374105252 Chr.17: 73253886-73253886 on Build GRCh38
rs749887952 Chr.17: 73253937-73253937 on Build GRCh38
rs185931168 Chr.17: 73254095-73254095 on Build GRCh38
rs915221239 Chr.17: 73254143-73254143 on Build GRCh38
rs969616327 Chr.17: 73254013-73254013 on Build GRCh38

View Full Product Details