Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74274169-74274678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
UniGene
Hs.147472
Gene Chromosome Location
Chr.17: 74274215-74314884 on Build GRCh38
Transcripts
NM_001172810.1 NM_023036.4 XM_011525125.2

SNP Detail

SNP ID Chromosome Location
rs972776266 Chr.17: 74274202-74274202 on Build GRCh38
rs188822172 Chr.17: 74274587-74274587 on Build GRCh38
rs890049988 Chr.17: 74274458-74274458 on Build GRCh38
rs778315009 Chr.17: 74274504-74274504 on Build GRCh38
rs746625773 Chr.17: 74274565-74274565 on Build GRCh38
rs201298434 Chr.17: 74274453-74274453 on Build GRCh38
rs751029617 Chr.17: 74274287-74274296 on Build GRCh38
rs754497256 Chr.17: 74274340-74274340 on Build GRCh38
rs146925440 Chr.17: 74274475-74274475 on Build GRCh38
rs1042117603 Chr.17: 74274343-74274343 on Build GRCh38
rs918742559 Chr.17: 74274536-74274536 on Build GRCh38
rs959560267 Chr.17: 74274562-74274562 on Build GRCh38
rs370544004 Chr.17: 74274255-74274255 on Build GRCh38
rs112109136 Chr.17: 74274567-74274567 on Build GRCh38
rs999374537 Chr.17: 74274439-74274439 on Build GRCh38
rs763405700 Chr.17: 74274376-74274376 on Build GRCh38
rs894975850 Chr.17: 74274305-74274305 on Build GRCh38
rs34306690 Chr.17: 74274205-74274206 on Build GRCh38
rs952816294 Chr.17: 74274239-74274239 on Build GRCh38
rs34919463 Chr.17: 74274215-74274216 on Build GRCh38
rs965495300 Chr.17: 74274485-74274485 on Build GRCh38
rs1002039018 Chr.17: 74274287-74274288 on Build GRCh38
rs547693786 Chr.17: 74274469-74274469 on Build GRCh38
rs1056190206 Chr.17: 74274606-74274606 on Build GRCh38
rs1877687 Chr.17: 74274303-74274303 on Build GRCh38
rs949141619 Chr.17: 74274342-74274342 on Build GRCh38
rs1048451954 Chr.17: 74274282-74274282 on Build GRCh38
rs952767307 Chr.17: 74274548-74274548 on Build GRCh38
rs549248126 Chr.17: 74274219-74274219 on Build GRCh38
rs917788552 Chr.17: 74274631-74274631 on Build GRCh38
rs900948220 Chr.17: 74274227-74274227 on Build GRCh38
rs763485792 Chr.17: 74274284-74274285 on Build GRCh38
rs1054063957 Chr.17: 74274455-74274455 on Build GRCh38
rs747311219 Chr.17: 74274559-74274559 on Build GRCh38
rs551715321 Chr.17: 74274335-74274335 on Build GRCh38
rs966621636 Chr.17: 74274419-74274419 on Build GRCh38
rs761805361 Chr.17: 74274230-74274230 on Build GRCh38
rs779354774 Chr.17: 74274381-74274381 on Build GRCh38
rs184147791 Chr.17: 74274456-74274456 on Build GRCh38
rs944179640 Chr.17: 74274236-74274236 on Build GRCh38
rs562947150 Chr.17: 74274302-74274302 on Build GRCh38
rs1025723225 Chr.17: 74274535-74274535 on Build GRCh38
rs5822035 Chr.17: 74274285-74274294 on Build GRCh38
rs1877686 Chr.17: 74274443-74274443 on Build GRCh38
rs909893225 Chr.17: 74274565-74274565 on Build GRCh38
rs909105509 Chr.17: 74274265-74274265 on Build GRCh38
rs984547404 Chr.17: 74274558-74274558 on Build GRCh38
rs375965264 Chr.17: 74274385-74274385 on Build GRCh38
rs753355539 Chr.17: 74274336-74274336 on Build GRCh38
rs965535064 Chr.17: 74274490-74274490 on Build GRCh38

View Full Product Details