Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74588427-74588935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf77
Entrez Gene ID
146723
Gene Name
chromosome 17 open reading frame 77
Gene Aliases
-
UniGene
Hs.350775
Gene Chromosome Location
Chr.17: 74584679-74594209 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2 NM_001115152.1

Gene Symbol
CD300LD
Entrez Gene ID
100131439
Gene Name
CD300 molecule like family member d
Gene Aliases
CD300D, CLM-4, CMRF35-A4, CMRF35A4
UniGene
Hs.531099
Gene Chromosome Location
Chr.17: 74579972-74592231 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2 NM_001115152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4904931 c.147G>A p.L49L Chr.17: 74588743-74588743 on Build GRCh38
5444738 c.232C>A p.R78R Chr.17: 74588658-74588658 on Build GRCh38
5508892 c.379+1G>T p.? Chr.17: 74588510-74588510 on Build GRCh38
148338 c.265G>A p.V89M Chr.17: 74588625-74588625 on Build GRCh38
1493982 c.89C>T p.S30L Chr.17: 74588801-74588801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748383823 Chr.17: 74588820-74588820 on Build GRCh38
rs763733780 Chr.17: 74588793-74588793 on Build GRCh38
rs1035901556 Chr.17: 74588637-74588637 on Build GRCh38
rs201241574 Chr.17: 74588480-74588480 on Build GRCh38
rs762282261 Chr.17: 74588587-74588587 on Build GRCh38
rs754428261 Chr.17: 74588703-74588703 on Build GRCh38
rs868469429 Chr.17: 74588529-74588529 on Build GRCh38
rs753724749 Chr.17: 74588797-74588797 on Build GRCh38
rs748078294 Chr.17: 74588542-74588542 on Build GRCh38
rs373845010 Chr.17: 74588457-74588457 on Build GRCh38
rs368705571 Chr.17: 74588749-74588749 on Build GRCh38
rs373622492 Chr.17: 74588549-74588549 on Build GRCh38
rs760255267 Chr.17: 74588792-74588792 on Build GRCh38
rs370521746 Chr.17: 74588652-74588652 on Build GRCh38
rs761013998 Chr.17: 74588579-74588579 on Build GRCh38
rs772670006 Chr.17: 74588507-74588507 on Build GRCh38
rs765806095 Chr.17: 74588691-74588691 on Build GRCh38
rs746716913 Chr.17: 74588522-74588522 on Build GRCh38
rs765790946 Chr.17: 74588588-74588588 on Build GRCh38
rs771577718 Chr.17: 74588872-74588872 on Build GRCh38
rs371226315 Chr.17: 74588721-74588721 on Build GRCh38
rs749294685 Chr.17: 74588648-74588648 on Build GRCh38
rs764890553 Chr.17: 74588886-74588886 on Build GRCh38
rs199533935 Chr.17: 74588738-74588738 on Build GRCh38
rs1009477678 Chr.17: 74588568-74588568 on Build GRCh38
rs558792867 Chr.17: 74588505-74588505 on Build GRCh38
rs763576087 Chr.17: 74588689-74588689 on Build GRCh38
rs959732397 Chr.17: 74588645-74588645 on Build GRCh38
rs564832234 Chr.17: 74588736-74588736 on Build GRCh38
rs1019978456 Chr.17: 74588890-74588890 on Build GRCh38
rs146929471 Chr.17: 74588857-74588857 on Build GRCh38
rs377452480 Chr.17: 74588571-74588571 on Build GRCh38
rs142719069 Chr.17: 74588748-74588748 on Build GRCh38
rs760913950 Chr.17: 74588473-74588473 on Build GRCh38
rs752277663 Chr.17: 74588533-74588533 on Build GRCh38
rs755931050 Chr.17: 74588626-74588626 on Build GRCh38
rs535714190 Chr.17: 74588711-74588711 on Build GRCh38
rs1029153846 Chr.17: 74588911-74588911 on Build GRCh38
rs186264456 Chr.17: 74588591-74588591 on Build GRCh38
rs868497373 Chr.17: 74588750-74588750 on Build GRCh38
rs759981218 Chr.17: 74588859-74588859 on Build GRCh38
rs1029187269 Chr.17: 74588563-74588563 on Build GRCh38
rs777015959 Chr.17: 74588669-74588669 on Build GRCh38
rs763857125 Chr.17: 74588878-74588878 on Build GRCh38
rs529615042 Chr.17: 74588836-74588836 on Build GRCh38
rs200915238 Chr.17: 74588616-74588616 on Build GRCh38
rs771521830 Chr.17: 74588762-74588762 on Build GRCh38
rs373955929 Chr.17: 74588686-74588686 on Build GRCh38
rs781360547 Chr.17: 74588812-74588812 on Build GRCh38
rs780381823 Chr.17: 74588569-74588577 on Build GRCh38
rs375361887 Chr.17: 74588617-74588617 on Build GRCh38
rs750999572 Chr.17: 74588589-74588589 on Build GRCh38
rs374599014 Chr.17: 74588747-74588747 on Build GRCh38
rs783239 Chr.17: 74588625-74588625 on Build GRCh38
rs777023909 Chr.17: 74588739-74588739 on Build GRCh38
rs200235728 Chr.17: 74588523-74588523 on Build GRCh38
rs762229243 Chr.17: 74588676-74588676 on Build GRCh38
rs751143167 Chr.17: 74588694-74588694 on Build GRCh38
rs369529940 Chr.17: 74588881-74588881 on Build GRCh38
rs759621526 Chr.17: 74588467-74588467 on Build GRCh38
rs749896449 Chr.17: 74588498-74588498 on Build GRCh38
rs925736134 Chr.17: 74588824-74588824 on Build GRCh38
rs761698710 Chr.17: 74588883-74588883 on Build GRCh38
rs770203986 Chr.17: 74588835-74588835 on Build GRCh38
rs766967345 Chr.17: 74588787-74588787 on Build GRCh38
rs749707317 Chr.17: 74588840-74588840 on Build GRCh38
rs553248708 Chr.17: 74588482-74588482 on Build GRCh38
rs538792390 Chr.17: 74588468-74588468 on Build GRCh38
rs987422430 Chr.17: 74588781-74588781 on Build GRCh38
rs367737209 Chr.17: 74588462-74588462 on Build GRCh38
rs375291849 Chr.17: 74588460-74588460 on Build GRCh38
rs778949900 Chr.17: 74588801-74588801 on Build GRCh38
rs191085731 Chr.17: 74588724-74588724 on Build GRCh38
rs563559353 Chr.17: 74588893-74588893 on Build GRCh38
rs369631943 Chr.17: 74588891-74588891 on Build GRCh38
rs752992594 Chr.17: 74588470-74588470 on Build GRCh38
rs749188852 Chr.17: 74588733-74588733 on Build GRCh38
rs757905488 Chr.17: 74588491-74588491 on Build GRCh38
rs372269225 Chr.17: 74588782-74588782 on Build GRCh38
rs535122673 Chr.17: 74588552-74588552 on Build GRCh38
rs747063220 Chr.17: 74588658-74588658 on Build GRCh38
rs760165367 Chr.17: 74588874-74588874 on Build GRCh38
rs777654055 Chr.17: 74588646-74588646 on Build GRCh38
rs781234456 Chr.17: 74588735-74588735 on Build GRCh38
rs978596083 Chr.17: 74588475-74588475 on Build GRCh38
rs750286256 Chr.17: 74588804-74588804 on Build GRCh38
rs754381841 Chr.17: 74588490-74588490 on Build GRCh38
rs775049969 Chr.17: 74588873-74588873 on Build GRCh38
rs140321593 Chr.17: 74588728-74588730 on Build GRCh38
rs752361854 Chr.17: 74588791-74588791 on Build GRCh38
rs768796936 Chr.17: 74588858-74588858 on Build GRCh38
rs931441652 Chr.17: 74588842-74588842 on Build GRCh38
rs374553535 Chr.17: 74588583-74588583 on Build GRCh38
rs774693512 Chr.17: 74588776-74588776 on Build GRCh38
rs368714951 Chr.17: 74588575-74588575 on Build GRCh38
rs756980571 Chr.17: 74588800-74588800 on Build GRCh38
rs768955031 Chr.17: 74588864-74588864 on Build GRCh38
rs1037039205 Chr.17: 74588909-74588909 on Build GRCh38
rs757949518 Chr.17: 74588504-74588504 on Build GRCh38
rs372112007 Chr.17: 74588896-74588896 on Build GRCh38
rs571471446 Chr.17: 74588506-74588506 on Build GRCh38
rs752310378 Chr.17: 74588716-74588716 on Build GRCh38
rs775705093 Chr.17: 74588655-74588655 on Build GRCh38
rs996095060 Chr.17: 74588585-74588585 on Build GRCh38
rs747426462 Chr.17: 74588812-74588814 on Build GRCh38
rs765965553 Chr.17: 74588615-74588615 on Build GRCh38
rs747280884 Chr.17: 74588809-74588809 on Build GRCh38
rs765739470 Chr.17: 74588677-74588677 on Build GRCh38
rs180816540 Chr.17: 74588565-74588565 on Build GRCh38
rs757286773 Chr.17: 74588734-74588734 on Build GRCh38

View Full Product Details