Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74924522-74925033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTOP2
Entrez Gene ID
92736
Gene Name
otopetrin 2
Gene Aliases
-
UniGene
Hs.352515
Gene Chromosome Location
Chr.17: 74922963-74933913 on Build GRCh38
Transcripts
NM_178160.2 XM_011525479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645854 c.108G>A p.A36A Chr.17: 74924740-74924740 on Build GRCh38
1479988 c.253G>A p.V85M Chr.17: 74924885-74924885 on Build GRCh38
4913032 c.306G>T p.W102C Chr.17: 74924938-74924938 on Build GRCh38
5630730 c.99G>A p.V33V Chr.17: 74924731-74924731 on Build GRCh38
4978595 c.134C>T p.T45M Chr.17: 74924766-74924766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538005504 Chr.17: 74924789-74924789 on Build GRCh38
rs777555340 Chr.17: 74924623-74924623 on Build GRCh38
rs868163366 Chr.17: 74924688-74924688 on Build GRCh38
rs7224055 Chr.17: 74924574-74924574 on Build GRCh38
rs780531091 Chr.17: 74924606-74924606 on Build GRCh38
rs557988355 Chr.17: 74924799-74924799 on Build GRCh38
rs968394414 Chr.17: 74924972-74924972 on Build GRCh38
rs201029457 Chr.17: 74924869-74924869 on Build GRCh38
rs748893243 Chr.17: 74924701-74924701 on Build GRCh38
rs1039738358 Chr.17: 74924981-74924981 on Build GRCh38
rs974159482 Chr.17: 74924566-74924566 on Build GRCh38
rs751414454 Chr.17: 74924931-74924931 on Build GRCh38
rs766974048 Chr.17: 74924588-74924588 on Build GRCh38
rs777658228 Chr.17: 74924912-74924912 on Build GRCh38
rs151293476 Chr.17: 74924835-74924835 on Build GRCh38
rs753447052 Chr.17: 74924836-74924836 on Build GRCh38
rs778604743 Chr.17: 74924776-74924776 on Build GRCh38
rs774629354 Chr.17: 74924815-74924815 on Build GRCh38
rs1038793108 Chr.17: 74924668-74924668 on Build GRCh38
rs921767063 Chr.17: 74924671-74924671 on Build GRCh38
rs1052456874 Chr.17: 74924551-74924551 on Build GRCh38
rs779017155 Chr.17: 74924864-74924864 on Build GRCh38
rs759936641 Chr.17: 74924746-74924746 on Build GRCh38
rs769895619 Chr.17: 74924801-74924801 on Build GRCh38
rs369382708 Chr.17: 74924652-74924652 on Build GRCh38
rs77742163 Chr.17: 74924766-74924766 on Build GRCh38
rs1012844474 Chr.17: 74924759-74924782 on Build GRCh38
rs767443599 Chr.17: 74924815-74924816 on Build GRCh38
rs758238290 Chr.17: 74924911-74924911 on Build GRCh38
rs564166454 Chr.17: 74924638-74924638 on Build GRCh38
rs934464826 Chr.17: 74924873-74924873 on Build GRCh38
rs899827798 Chr.17: 74924987-74924987 on Build GRCh38
rs372873458 Chr.17: 74924702-74924702 on Build GRCh38
rs759488042 Chr.17: 74924813-74924814 on Build GRCh38
rs998469247 Chr.17: 74925009-74925009 on Build GRCh38
rs758744816 Chr.17: 74924584-74924584 on Build GRCh38
rs769795510 Chr.17: 74924731-74924731 on Build GRCh38
rs751326646 Chr.17: 74924853-74924853 on Build GRCh38
rs934920007 Chr.17: 74924633-74924633 on Build GRCh38
rs748290835 Chr.17: 74924725-74924725 on Build GRCh38
rs548812987 Chr.17: 74924747-74924747 on Build GRCh38
rs180710937 Chr.17: 74924615-74924615 on Build GRCh38
rs777273457 Chr.17: 74924888-74924888 on Build GRCh38
rs752626808 Chr.17: 74924974-74924974 on Build GRCh38
rs750021794 Chr.17: 74924840-74924840 on Build GRCh38
rs747354085 Chr.17: 74924879-74924879 on Build GRCh38
rs751941715 Chr.17: 74924596-74924596 on Build GRCh38
rs764861558 Chr.17: 74924838-74924838 on Build GRCh38
rs755682591 Chr.17: 74924695-74924695 on Build GRCh38
rs756898818 Chr.17: 74924775-74924775 on Build GRCh38
rs764744021 Chr.17: 74924892-74924892 on Build GRCh38
rs752313299 Chr.17: 74924694-74924694 on Build GRCh38
rs749163676 Chr.17: 74924626-74924626 on Build GRCh38
rs762709751 Chr.17: 74924917-74924917 on Build GRCh38
rs752694917 Chr.17: 74924830-74924831 on Build GRCh38
rs752532668 Chr.17: 74924807-74924807 on Build GRCh38
rs771157606 Chr.17: 74924742-74924742 on Build GRCh38
rs754541581 Chr.17: 74924602-74924602 on Build GRCh38
rs761229321 Chr.17: 74924889-74924889 on Build GRCh38
rs760276656 Chr.17: 74924758-74924758 on Build GRCh38
rs777574018 Chr.17: 74924699-74924699 on Build GRCh38
rs768050085 Chr.17: 74924689-74924689 on Build GRCh38
rs774360383 Chr.17: 74924639-74924639 on Build GRCh38
rs369224163 Chr.17: 74924740-74924740 on Build GRCh38
rs772425018 Chr.17: 74924823-74924823 on Build GRCh38
rs573377512 Chr.17: 74924630-74924630 on Build GRCh38
rs767451528 Chr.17: 74924962-74924962 on Build GRCh38
rs202245064 Chr.17: 74924832-74924832 on Build GRCh38
rs893853028 Chr.17: 74924908-74924908 on Build GRCh38
rs909610408 Chr.17: 74924723-74924723 on Build GRCh38
rs774631961 Chr.17: 74924743-74924743 on Build GRCh38
rs776614076 Chr.17: 74924659-74924659 on Build GRCh38
rs998834716 Chr.17: 74924640-74924640 on Build GRCh38
rs772758854 Chr.17: 74924901-74924901 on Build GRCh38
rs750243041 Chr.17: 74924778-74924778 on Build GRCh38
rs532804031 Chr.17: 74924673-74924673 on Build GRCh38
rs763125176 Chr.17: 74924739-74924739 on Build GRCh38
rs202221330 Chr.17: 74924837-74924837 on Build GRCh38
rs373149079 Chr.17: 74924863-74924863 on Build GRCh38
rs765995235 Chr.17: 74924918-74924918 on Build GRCh38
rs749398664 Chr.17: 74924860-74924860 on Build GRCh38
rs781098215 Chr.17: 74924859-74924859 on Build GRCh38
rs753573991 Chr.17: 74924770-74924770 on Build GRCh38
rs566463973 Chr.17: 74924706-74924706 on Build GRCh38
rs776035420 Chr.17: 74924878-74924878 on Build GRCh38
rs1052490426 Chr.17: 74924899-74924899 on Build GRCh38
rs375036660 Chr.17: 74924718-74924718 on Build GRCh38
rs755221850 Chr.17: 74924852-74924852 on Build GRCh38
rs376129549 Chr.17: 74924867-74924867 on Build GRCh38
rs1052105417 Chr.17: 74924685-74924685 on Build GRCh38
rs747667091 Chr.17: 74924611-74924611 on Build GRCh38
rs756029019 Chr.17: 74924982-74924982 on Build GRCh38
rs914231335 Chr.17: 74924969-74924969 on Build GRCh38
rs779773298 Chr.17: 74924844-74924844 on Build GRCh38
rs777873109 Chr.17: 74924806-74924806 on Build GRCh38
rs140803752 Chr.17: 74924711-74924711 on Build GRCh38
rs555455500 Chr.17: 74924687-74924687 on Build GRCh38
rs765497904 Chr.17: 74924665-74924665 on Build GRCh38
rs759824861 Chr.17: 74924818-74924818 on Build GRCh38
rs989475118 Chr.17: 74924897-74924897 on Build GRCh38
rs754764725 Chr.17: 74924958-74924958 on Build GRCh38
rs773415911 Chr.17: 74924669-74924669 on Build GRCh38
rs766606023 Chr.17: 74924682-74924682 on Build GRCh38
rs769344633 Chr.17: 74924886-74924886 on Build GRCh38
rs367611435 Chr.17: 74924779-74924779 on Build GRCh38
rs754960311 Chr.17: 74924858-74924858 on Build GRCh38
rs778530745 Chr.17: 74924708-74924708 on Build GRCh38
rs761859462 Chr.17: 74924661-74924661 on Build GRCh38
rs746055801 Chr.17: 74924866-74924866 on Build GRCh38
rs535352918 Chr.17: 74924715-74924715 on Build GRCh38
rs758138929 Chr.17: 74924843-74924843 on Build GRCh38
rs962766189 Chr.17: 74924558-74924558 on Build GRCh38
rs776164275 Chr.17: 74924824-74924824 on Build GRCh38
rs753189877 Chr.17: 74924755-74924755 on Build GRCh38

View Full Product Details