Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75638973-75639514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL5
Entrez Gene ID
9400
Gene Name
RecQ like helicase 5
Gene Aliases
RECQ5
UniGene
Hs.632229
Gene Chromosome Location
Chr.17: 75626845-75667202 on Build GRCh38
Transcripts
NM_004259.6 XM_005257818.3 XM_005257823.3 XM_011525482.2 XM_011525484.1 XM_011525485.2 XM_011525486.2 XM_017025343.1 NM_001162995.2 NR_028439.1 XM_011525118.2 XM_011525119.2 XM_017024945.1 XM_017024946.1 XM_017024947.1 XM_017024948.1

Gene Symbol
SMIM5
Entrez Gene ID
643008
Gene Name
small integral membrane protein 5
Gene Aliases
C17orf109, PP12104
UniGene
Hs.528605
Gene Chromosome Location
Chr.17: 75633434-75641406 on Build GRCh38
Transcripts
NM_004259.6 XM_005257818.3 XM_005257823.3 XM_011525482.2 XM_011525484.1 XM_011525485.2 XM_011525486.2 XM_017025343.1 NM_001162995.2 NR_028439.1 XM_011525118.2 XM_011525119.2 XM_017024945.1 XM_017024946.1 XM_017024947.1 XM_017024948.1

SNP Detail

SNP ID Chromosome Location
rs912914743 Chr.17: 75639484-75639484 on Build GRCh38
rs993961155 Chr.17: 75639310-75639310 on Build GRCh38
rs551919465 Chr.17: 75639075-75639075 on Build GRCh38
rs979721863 Chr.17: 75639081-75639081 on Build GRCh38
rs571633882 Chr.17: 75639076-75639076 on Build GRCh38
rs938426991 Chr.17: 75639193-75639193 on Build GRCh38
rs944053647 Chr.17: 75639271-75639271 on Build GRCh38
rs997350430 Chr.17: 75639098-75639098 on Build GRCh38
rs938322897 Chr.17: 75639008-75639008 on Build GRCh38
rs912883363 Chr.17: 75639242-75639242 on Build GRCh38
rs552882693 Chr.17: 75639184-75639184 on Build GRCh38
rs928297860 Chr.17: 75638994-75638994 on Build GRCh38
rs1052717843 Chr.17: 75639019-75639019 on Build GRCh38
rs1042614197 Chr.17: 75639165-75639165 on Build GRCh38
rs746950689 Chr.17: 75639171-75639171 on Build GRCh38
rs574911216 Chr.17: 75639274-75639274 on Build GRCh38
rs540859967 Chr.17: 75639472-75639472 on Build GRCh38
rs891243032 Chr.17: 75639352-75639352 on Build GRCh38
rs184270297 Chr.17: 75639110-75639110 on Build GRCh38
rs776525928 Chr.17: 75639347-75639347 on Build GRCh38
rs1027251095 Chr.17: 75639174-75639174 on Build GRCh38
rs888353373 Chr.17: 75639374-75639374 on Build GRCh38
rs572953582 Chr.17: 75639227-75639227 on Build GRCh38
rs897278414 Chr.17: 75639309-75639309 on Build GRCh38
rs777567206 Chr.17: 75639124-75639124 on Build GRCh38
rs577658931 Chr.17: 75639391-75639391 on Build GRCh38
rs1020446174 Chr.17: 75639436-75639436 on Build GRCh38
rs1008269353 Chr.17: 75639169-75639169 on Build GRCh38
rs866074678 Chr.17: 75639143-75639143 on Build GRCh38
rs549979207 Chr.17: 75639456-75639456 on Build GRCh38
rs961460982 Chr.17: 75639021-75639021 on Build GRCh38
rs903999870 Chr.17: 75639095-75639095 on Build GRCh38
rs868035707 Chr.17: 75639141-75639141 on Build GRCh38
rs1026864524 Chr.17: 75639392-75639392 on Build GRCh38
rs979995783 Chr.17: 75639409-75639409 on Build GRCh38
rs973967370 Chr.17: 75639172-75639172 on Build GRCh38
rs959807431 Chr.17: 75639470-75639470 on Build GRCh38
rs535366404 Chr.17: 75639232-75639232 on Build GRCh38
rs995431181 Chr.17: 75639379-75639379 on Build GRCh38
rs868147090 Chr.17: 75639191-75639191 on Build GRCh38
rs139802510 Chr.17: 75639250-75639250 on Build GRCh38
rs543792032 Chr.17: 75639281-75639281 on Build GRCh38
rs897135288 Chr.17: 75639377-75639377 on Build GRCh38
rs1008699209 Chr.17: 75638992-75638992 on Build GRCh38
rs980262244 Chr.17: 75639190-75639190 on Build GRCh38
rs978241390 Chr.17: 75639487-75639487 on Build GRCh38
rs1015594513 Chr.17: 75639170-75639170 on Build GRCh38
rs538213708 Chr.17: 75639025-75639025 on Build GRCh38
rs771203455 Chr.17: 75638997-75638998 on Build GRCh38
rs904024351 Chr.17: 75639284-75639284 on Build GRCh38
rs1041491148 Chr.17: 75639278-75639278 on Build GRCh38
rs749425954 Chr.17: 75639403-75639403 on Build GRCh38
rs749834485 Chr.17: 75639214-75639214 on Build GRCh38
rs892755879 Chr.17: 75639044-75639044 on Build GRCh38
rs1035920756 Chr.17: 75639413-75639413 on Build GRCh38
rs1043929752 Chr.17: 75639252-75639252 on Build GRCh38
rs1008302002 Chr.17: 75639366-75639366 on Build GRCh38
rs563870327 Chr.17: 75639291-75639291 on Build GRCh38
rs758133129 Chr.17: 75639105-75639105 on Build GRCh38
rs1015625552 Chr.17: 75639372-75639372 on Build GRCh38
rs1009763313 Chr.17: 75639067-75639067 on Build GRCh38
rs992965231 Chr.17: 75639133-75639133 on Build GRCh38
rs1050344792 Chr.17: 75639351-75639351 on Build GRCh38
rs945495083 Chr.17: 75639148-75639148 on Build GRCh38
rs912984020 Chr.17: 75639142-75639142 on Build GRCh38

View Full Product Details