Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49822700-49823201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO5B
Entrez Gene ID
4645
Gene Name
myosin VB
Gene Aliases
-
UniGene
Hs.720076
Gene Chromosome Location
Chr.18: 49822786-50195081 on Build GRCh38
Transcripts
NM_001080467.2 NR_117096.1

Gene Symbol
SNHG22
Entrez Gene ID
103091864
Gene Name
small nucleolar RNA host gene 22
Gene Aliases
SCARNA17, SCARNA17HG
UniGene
Hs.646459
Gene Chromosome Location
Chr.18: 49814023-49851059 on Build GRCh38
Transcripts
NM_001080467.2 NR_117096.1

SNP Detail

SNP ID Chromosome Location
rs905004781 Chr.18: 49822810-49822810 on Build GRCh38
rs777915881 Chr.18: 49823107-49823107 on Build GRCh38
rs886053853 Chr.18: 49823154-49823154 on Build GRCh38
rs371267343 Chr.18: 49823092-49823092 on Build GRCh38
rs199813194 Chr.18: 49822965-49822965 on Build GRCh38
rs180921152 Chr.18: 49822773-49822773 on Build GRCh38
rs1030811304 Chr.18: 49822823-49822823 on Build GRCh38
rs377694296 Chr.18: 49823090-49823090 on Build GRCh38
rs968127835 Chr.18: 49822805-49822805 on Build GRCh38
rs368155236 Chr.18: 49823179-49823179 on Build GRCh38
rs373429782 Chr.18: 49823080-49823080 on Build GRCh38
rs796439391 Chr.18: 49822875-49822875 on Build GRCh38
rs552752053 Chr.18: 49822746-49822746 on Build GRCh38
rs538422546 Chr.18: 49823150-49823150 on Build GRCh38
rs72925910 Chr.18: 49823158-49823158 on Build GRCh38
rs904312484 Chr.18: 49823177-49823177 on Build GRCh38
rs568315666 Chr.18: 49822969-49822969 on Build GRCh38
rs571821458 Chr.18: 49823175-49823175 on Build GRCh38
rs1048887905 Chr.18: 49822792-49822792 on Build GRCh38
rs140991986 Chr.18: 49822779-49822779 on Build GRCh38
rs202171965 Chr.18: 49823155-49823155 on Build GRCh38
rs369420591 Chr.18: 49823120-49823120 on Build GRCh38
rs553050934 Chr.18: 49822945-49822945 on Build GRCh38
rs992065300 Chr.18: 49822946-49822946 on Build GRCh38
rs199830730 Chr.18: 49823139-49823139 on Build GRCh38
rs201994842 Chr.18: 49822882-49822882 on Build GRCh38
rs375099918 Chr.18: 49823132-49823133 on Build GRCh38
rs186286356 Chr.18: 49823087-49823087 on Build GRCh38
rs558262946 Chr.18: 49823156-49823156 on Build GRCh38
rs900798281 Chr.18: 49823174-49823175 on Build GRCh38
rs536555299 Chr.18: 49823037-49823037 on Build GRCh38
rs796273976 Chr.18: 49822864-49822864 on Build GRCh38
rs879173424 Chr.18: 49823048-49823048 on Build GRCh38
rs368075985 Chr.18: 49823176-49823177 on Build GRCh38
rs564840373 Chr.18: 49822863-49822863 on Build GRCh38
rs996393879 Chr.18: 49823175-49823175 on Build GRCh38
rs1040601442 Chr.18: 49823174-49823174 on Build GRCh38
rs367570644 Chr.18: 49823171-49823171 on Build GRCh38
rs867604296 Chr.18: 49822759-49822759 on Build GRCh38
rs1047262643 Chr.18: 49823112-49823112 on Build GRCh38
rs996172529 Chr.18: 49822768-49822768 on Build GRCh38
rs371306335 Chr.18: 49823130-49823130 on Build GRCh38
rs1040633183 Chr.18: 49822738-49822738 on Build GRCh38
rs9947614 Chr.18: 49823103-49823103 on Build GRCh38
rs375545150 Chr.18: 49823106-49823106 on Build GRCh38
rs969042743 Chr.18: 49822988-49822988 on Build GRCh38
rs897569878 Chr.18: 49822761-49822761 on Build GRCh38

View Full Product Details