Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6901622-6902125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE1
Entrez Gene ID
2015
Gene Name
adhesion G protein-coupled receptor E1
Gene Aliases
EMR1, TM7LN3
UniGene
Hs.2375
Gene Chromosome Location
Chr.19: 6887549-6940453 on Build GRCh38
Transcripts
NM_001256252.1 NM_001256253.1 NM_001256254.1 NM_001256255.1 NM_001974.4 XM_011527794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887035 c.515-3C>T p.? Chr.19: 6901872-6901872 on Build GRCh38
349694 c.636C>G p.F212L Chr.19: 6901996-6901996 on Build GRCh38
713969 c.610G>A p.E204K Chr.19: 6901970-6901970 on Build GRCh38
3423345 c.605G>A p.G202E Chr.19: 6901965-6901965 on Build GRCh38
3938370 c.645C>A p.L215L Chr.19: 6902005-6902005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751741117 Chr.19: 6902055-6902055 on Build GRCh38
rs531805943 Chr.19: 6901706-6901706 on Build GRCh38
rs188197334 Chr.19: 6901650-6901650 on Build GRCh38
rs546368454 Chr.19: 6901774-6901774 on Build GRCh38
rs928463947 Chr.19: 6901758-6901758 on Build GRCh38
rs961524136 Chr.19: 6902089-6902089 on Build GRCh38
rs981495623 Chr.19: 6902048-6902048 on Build GRCh38
rs199783244 Chr.19: 6901927-6901927 on Build GRCh38
rs1023315324 Chr.19: 6901656-6901656 on Build GRCh38
rs371679836 Chr.19: 6901926-6901926 on Build GRCh38
rs749191912 Chr.19: 6901948-6901948 on Build GRCh38
rs180751858 Chr.19: 6902080-6902080 on Build GRCh38
rs757661323 Chr.19: 6901905-6901905 on Build GRCh38
rs912477028 Chr.19: 6901746-6901746 on Build GRCh38
rs780235055 Chr.19: 6901920-6901920 on Build GRCh38
rs747078010 Chr.19: 6901921-6901921 on Build GRCh38
rs750736236 Chr.19: 6902040-6902040 on Build GRCh38
rs367846052 Chr.19: 6901915-6901915 on Build GRCh38
rs919835300 Chr.19: 6901800-6901800 on Build GRCh38
rs376248323 Chr.19: 6901961-6901961 on Build GRCh38
rs149592085 Chr.19: 6902014-6902014 on Build GRCh38
rs138193492 Chr.19: 6902013-6902013 on Build GRCh38
rs931279679 Chr.19: 6901840-6901840 on Build GRCh38
rs970063838 Chr.19: 6902036-6902036 on Build GRCh38
rs764582668 Chr.19: 6901882-6901882 on Build GRCh38
rs750950000 Chr.19: 6901660-6901660 on Build GRCh38
rs2228533 Chr.19: 6901912-6901912 on Build GRCh38
rs777153696 Chr.19: 6902027-6902027 on Build GRCh38
rs754247451 Chr.19: 6901902-6901902 on Build GRCh38
rs750662133 Chr.19: 6901914-6901914 on Build GRCh38
rs369530344 Chr.19: 6902051-6902051 on Build GRCh38
rs897739 Chr.19: 6901704-6901704 on Build GRCh38
rs1049089633 Chr.19: 6901806-6901806 on Build GRCh38
rs1036896054 Chr.19: 6901794-6901794 on Build GRCh38
rs897738 Chr.19: 6901880-6901880 on Build GRCh38
rs771611173 Chr.19: 6902000-6902000 on Build GRCh38
rs747787708 Chr.19: 6901727-6901727 on Build GRCh38
rs774078797 Chr.19: 6901862-6901862 on Build GRCh38
rs1030710247 Chr.19: 6901988-6901988 on Build GRCh38
rs754418887 Chr.19: 6901695-6901695 on Build GRCh38
rs752146835 Chr.19: 6901877-6901877 on Build GRCh38
rs147011666 Chr.19: 6901876-6901876 on Build GRCh38
rs532730206 Chr.19: 6901649-6901649 on Build GRCh38
rs773205522 Chr.19: 6901838-6901838 on Build GRCh38
rs777833540 Chr.19: 6901945-6901945 on Build GRCh38
rs1022932232 Chr.19: 6902032-6902032 on Build GRCh38
rs763544449 Chr.19: 6902023-6902023 on Build GRCh38
rs781384823 Chr.19: 6902062-6902062 on Build GRCh38
rs61733001 Chr.19: 6901950-6901950 on Build GRCh38
rs981525383 Chr.19: 6901752-6901752 on Build GRCh38
rs202053872 Chr.19: 6902056-6902056 on Build GRCh38
rs151167763 Chr.19: 6901773-6901773 on Build GRCh38
rs200151344 Chr.19: 6901703-6901704 on Build GRCh38
rs762814050 Chr.19: 6901856-6901856 on Build GRCh38
rs930750644 Chr.19: 6901803-6901803 on Build GRCh38
rs1032329160 Chr.19: 6901658-6901658 on Build GRCh38
rs745547615 Chr.19: 6901983-6901983 on Build GRCh38
rs766674508 Chr.19: 6902053-6902053 on Build GRCh38
rs140292485 Chr.19: 6901861-6901861 on Build GRCh38
rs547840627 Chr.19: 6901736-6901736 on Build GRCh38
rs957665105 Chr.19: 6901681-6901681 on Build GRCh38
rs915946482 Chr.19: 6901716-6901716 on Build GRCh38
rs889453975 Chr.19: 6901867-6901867 on Build GRCh38
rs759074855 Chr.19: 6901864-6901864 on Build GRCh38

View Full Product Details