Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7607687-7608188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP3
Entrez Gene ID
57662
Gene Name
calmodulin regulated spectrin associated protein family member 3
Gene Aliases
KIAA1543, NEZHA, PPP1R80
UniGene
Hs.17686
Gene Chromosome Location
Chr.19: 7595902-7618310 on Build GRCh38
Transcripts
NM_001080429.2 NM_020902.1 XM_005272491.3 XM_011528154.2 XM_017027036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590415 c.664G>A p.A222T Chr.19: 7607830-7607830 on Build GRCh38
1397593 c.732G>A p.A244A Chr.19: 7608155-7608155 on Build GRCh38
5691343 c.658G>A p.A220T Chr.19: 7607824-7607824 on Build GRCh38
4557193 c.637G>A p.D213N Chr.19: 7608141-7608141 on Build GRCh38
1397591 c.713G>A p.R238H Chr.19: 7608136-7608136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023918638 Chr.19: 7607944-7607944 on Build GRCh38
rs368094381 Chr.19: 7607823-7607823 on Build GRCh38
rs773909989 Chr.19: 7608102-7608102 on Build GRCh38
rs759263772 Chr.19: 7607729-7607729 on Build GRCh38
rs763101819 Chr.19: 7608036-7608036 on Build GRCh38
rs772170549 Chr.19: 7607780-7607780 on Build GRCh38
rs911841695 Chr.19: 7608109-7608109 on Build GRCh38
rs541294860 Chr.19: 7607897-7607897 on Build GRCh38
rs777647848 Chr.19: 7607873-7607873 on Build GRCh38
rs200413625 Chr.19: 7607878-7607878 on Build GRCh38
rs754629548 Chr.19: 7607850-7607850 on Build GRCh38
rs777047148 Chr.19: 7607806-7607806 on Build GRCh38
rs773808921 Chr.19: 7607910-7607910 on Build GRCh38
rs561613044 Chr.19: 7607916-7607916 on Build GRCh38
rs368361887 Chr.19: 7608147-7608147 on Build GRCh38
rs973853894 Chr.19: 7607901-7607901 on Build GRCh38
rs77772511 Chr.19: 7607713-7607713 on Build GRCh38
rs529751395 Chr.19: 7607924-7607924 on Build GRCh38
rs890353499 Chr.19: 7607764-7607764 on Build GRCh38
rs199552187 Chr.19: 7608159-7608159 on Build GRCh38
rs372713856 Chr.19: 7608154-7608154 on Build GRCh38
rs763852626 Chr.19: 7608136-7608136 on Build GRCh38
rs984105534 Chr.19: 7607938-7607938 on Build GRCh38
rs920666059 Chr.19: 7608029-7608029 on Build GRCh38
rs767144355 Chr.19: 7607852-7607852 on Build GRCh38
rs763534754 Chr.19: 7607918-7607918 on Build GRCh38
rs894161314 Chr.19: 7607927-7607927 on Build GRCh38
rs773466410 Chr.19: 7607776-7607776 on Build GRCh38
rs924627695 Chr.19: 7607765-7607765 on Build GRCh38
rs946751409 Chr.19: 7608017-7608017 on Build GRCh38
rs758532659 Chr.19: 7607787-7607787 on Build GRCh38
rs372278659 Chr.19: 7607824-7607824 on Build GRCh38
rs1015505696 Chr.19: 7607993-7607993 on Build GRCh38
rs755864319 Chr.19: 7607863-7607863 on Build GRCh38
rs377116685 Chr.19: 7608076-7608076 on Build GRCh38
rs767034964 Chr.19: 7608085-7608085 on Build GRCh38
rs536410325 Chr.19: 7607749-7607749 on Build GRCh38
rs74556722 Chr.19: 7607750-7607750 on Build GRCh38
rs1055269709 Chr.19: 7607984-7607984 on Build GRCh38
rs549122093 Chr.19: 7607937-7607937 on Build GRCh38
rs939485333 Chr.19: 7607976-7607976 on Build GRCh38
rs574118963 Chr.19: 7607748-7607748 on Build GRCh38
rs757179247 Chr.19: 7608145-7608145 on Build GRCh38
rs571937717 Chr.19: 7608103-7608103 on Build GRCh38
rs1038417652 Chr.19: 7607743-7607743 on Build GRCh38
rs895258613 Chr.19: 7607763-7607763 on Build GRCh38
rs575107593 Chr.19: 7607783-7607783 on Build GRCh38
rs371424762 Chr.19: 7608160-7608160 on Build GRCh38
rs199695046 Chr.19: 7608144-7608144 on Build GRCh38
rs768798419 Chr.19: 7607792-7607792 on Build GRCh38
rs955190534 Chr.19: 7607827-7607827 on Build GRCh38
rs565216454 Chr.19: 7607767-7607767 on Build GRCh38
rs755853522 Chr.19: 7608134-7608134 on Build GRCh38
rs559545477 Chr.19: 7608044-7608044 on Build GRCh38
rs751082328 Chr.19: 7607849-7607849 on Build GRCh38
rs962199021 Chr.19: 7607996-7607996 on Build GRCh38
rs545335540 Chr.19: 7607751-7607751 on Build GRCh38
rs771550989 Chr.19: 7608164-7608164 on Build GRCh38
rs762138597 Chr.19: 7607808-7607808 on Build GRCh38
rs780073825 Chr.19: 7607883-7607883 on Build GRCh38
rs201927154 Chr.19: 7608072-7608072 on Build GRCh38
rs752524563 Chr.19: 7607859-7607859 on Build GRCh38
rs944621431 Chr.19: 7608116-7608116 on Build GRCh38
rs557153729 Chr.19: 7607740-7607740 on Build GRCh38
rs1032730225 Chr.19: 7607926-7607926 on Build GRCh38
rs752323641 Chr.19: 7608106-7608106 on Build GRCh38
rs940764310 Chr.19: 7607708-7607708 on Build GRCh38
rs767141676 Chr.19: 7608105-7608105 on Build GRCh38
rs941018480 Chr.19: 7607738-7607738 on Build GRCh38
rs770193714 Chr.19: 7608157-7608157 on Build GRCh38
rs775639273 Chr.19: 7607781-7607781 on Build GRCh38
rs374558137 Chr.19: 7607811-7607811 on Build GRCh38
rs973908738 Chr.19: 7608023-7608023 on Build GRCh38
rs199868688 Chr.19: 7608155-7608155 on Build GRCh38
rs764116416 Chr.19: 7607882-7607882 on Build GRCh38
rs773863987 Chr.19: 7607817-7607817 on Build GRCh38
rs1003748216 Chr.19: 7607988-7607988 on Build GRCh38
rs970955523 Chr.19: 7607946-7607946 on Build GRCh38
rs528725744 Chr.19: 7608064-7608064 on Build GRCh38
rs560813675 Chr.19: 7607888-7607888 on Build GRCh38
rs747122417 Chr.19: 7607887-7607887 on Build GRCh38
rs748482600 Chr.19: 7607889-7607889 on Build GRCh38

View Full Product Details