Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9818594-9819086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL12
Entrez Gene ID
54850
Gene Name
F-box and leucine rich repeat protein 12
Gene Aliases
Fbl12
UniGene
Hs.12439
Gene Chromosome Location
Chr.19: 9810267-9820527 on Build GRCh38
Transcripts
NM_001316936.1 NM_001316937.1 NM_001316938.1 NM_001316939.1 NM_001316940.1 NM_001316941.1 NM_001316942.1 NM_017703.2 XM_017026912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
22087 c.97C>T p.R33C Chr.19: 9818607-9818607 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757318304 Chr.19: 9818696-9818696 on Build GRCh38
rs1022388968 Chr.19: 9818657-9818657 on Build GRCh38
rs1003749700 Chr.19: 9818633-9818633 on Build GRCh38
rs776994796 Chr.19: 9818624-9818624 on Build GRCh38
rs760650771 Chr.19: 9818799-9818799 on Build GRCh38
rs550557486 Chr.19: 9818949-9818949 on Build GRCh38
rs755076057 Chr.19: 9818619-9818620 on Build GRCh38
rs781726992 Chr.19: 9818743-9818743 on Build GRCh38
rs750208821 Chr.19: 9818629-9818629 on Build GRCh38
rs151302778 Chr.19: 9818968-9818968 on Build GRCh38
rs950833078 Chr.19: 9818716-9818716 on Build GRCh38
rs770173398 Chr.19: 9818756-9818756 on Build GRCh38
rs370702553 Chr.19: 9818644-9818644 on Build GRCh38
rs889507452 Chr.19: 9818885-9818885 on Build GRCh38
rs770627579 Chr.19: 9818781-9818781 on Build GRCh38
rs978355949 Chr.19: 9819034-9819034 on Build GRCh38
rs868806959 Chr.19: 9818774-9818774 on Build GRCh38
rs755953779 Chr.19: 9818717-9818717 on Build GRCh38
rs560585457 Chr.19: 9818852-9818853 on Build GRCh38
rs764094932 Chr.19: 9818815-9818815 on Build GRCh38
rs990802472 Chr.19: 9818754-9818754 on Build GRCh38
rs1029383658 Chr.19: 9818621-9818621 on Build GRCh38
rs202104046 Chr.19: 9819037-9819037 on Build GRCh38
rs753846182 Chr.19: 9818925-9818925 on Build GRCh38
rs775521475 Chr.19: 9818796-9818796 on Build GRCh38
rs373173036 Chr.19: 9818701-9818701 on Build GRCh38
rs1010134406 Chr.19: 9818887-9818887 on Build GRCh38
rs533316542 Chr.19: 9818677-9818677 on Build GRCh38
rs749628366 Chr.19: 9818767-9818767 on Build GRCh38
rs915531001 Chr.19: 9818793-9818793 on Build GRCh38
rs1056667247 Chr.19: 9818871-9818871 on Build GRCh38
rs1042604302 Chr.19: 9818829-9818829 on Build GRCh38
rs751671303 Chr.19: 9818847-9818847 on Build GRCh38
rs750708251 Chr.19: 9818728-9818728 on Build GRCh38
rs191599755 Chr.19: 9818875-9818875 on Build GRCh38
rs534332420 Chr.19: 9818792-9818792 on Build GRCh38
rs915228797 Chr.19: 9819007-9819007 on Build GRCh38
rs376394175 Chr.19: 9818737-9818737 on Build GRCh38
rs761691565 Chr.19: 9818822-9818822 on Build GRCh38
rs1007001502 Chr.19: 9818905-9818905 on Build GRCh38
rs975028815 Chr.19: 9818695-9818695 on Build GRCh38
rs763933520 Chr.19: 9818712-9818712 on Build GRCh38
rs778149799 Chr.19: 9818763-9818763 on Build GRCh38
rs939593477 Chr.19: 9819038-9819038 on Build GRCh38
rs946949690 Chr.19: 9818809-9818809 on Build GRCh38
rs905442679 Chr.19: 9818628-9818628 on Build GRCh38
rs911002017 Chr.19: 9819063-9819063 on Build GRCh38
rs773961052 Chr.19: 9818684-9818684 on Build GRCh38
rs1051288699 Chr.19: 9818880-9818880 on Build GRCh38
rs773870634 Chr.19: 9818784-9818784 on Build GRCh38
rs563206130 Chr.19: 9818654-9818654 on Build GRCh38
rs759092390 Chr.19: 9818786-9818786 on Build GRCh38
rs754944045 Chr.19: 9818738-9818738 on Build GRCh38
rs767558848 Chr.19: 9818699-9818699 on Build GRCh38
rs930670699 Chr.19: 9818806-9818806 on Build GRCh38
rs771836733 Chr.19: 9818988-9818988 on Build GRCh38
rs777580105 Chr.19: 9818996-9818996 on Build GRCh38
rs771426901 Chr.19: 9818791-9818791 on Build GRCh38
rs770081792 Chr.19: 9818656-9818656 on Build GRCh38
rs1027911086 Chr.19: 9818945-9818945 on Build GRCh38
rs753738253 Chr.19: 9818819-9818819 on Build GRCh38
rs374354767 Chr.19: 9818686-9818686 on Build GRCh38
rs780405829 Chr.19: 9818735-9818735 on Build GRCh38
rs920658206 Chr.19: 9818790-9818790 on Build GRCh38
rs924148677 Chr.19: 9819045-9819045 on Build GRCh38
rs545599741 Chr.19: 9818707-9818707 on Build GRCh38
rs980773023 Chr.19: 9819026-9819026 on Build GRCh38
rs766694260 Chr.19: 9818823-9818823 on Build GRCh38
rs528908619 Chr.19: 9818926-9818926 on Build GRCh38
rs747308993 Chr.19: 9819013-9819013 on Build GRCh38
rs769259793 Chr.19: 9818623-9818623 on Build GRCh38
rs528174497 Chr.19: 9818873-9818873 on Build GRCh38
rs1035154222 Chr.19: 9818641-9818641 on Build GRCh38
rs553854953 Chr.19: 9818966-9818966 on Build GRCh38
rs867947468 Chr.19: 9818772-9818772 on Build GRCh38
rs928049240 Chr.19: 9818749-9818749 on Build GRCh38
rs557917700 Chr.19: 9819057-9819057 on Build GRCh38
rs1006518642 Chr.19: 9818631-9818631 on Build GRCh38
rs1037997497 Chr.19: 9818915-9818915 on Build GRCh38
rs750488214 Chr.19: 9818842-9818842 on Build GRCh38
rs568056008 Chr.19: 9818879-9818879 on Build GRCh38
rs762549198 Chr.19: 9818661-9818661 on Build GRCh38
rs755952187 Chr.19: 9818708-9818708 on Build GRCh38
rs76722602 Chr.19: 9818753-9818753 on Build GRCh38
rs758736833 Chr.19: 9818729-9818729 on Build GRCh38
rs1022066285 Chr.19: 9818910-9818910 on Build GRCh38
rs939547355 Chr.19: 9818861-9818861 on Build GRCh38
rs772870772 Chr.19: 9818658-9818658 on Build GRCh38
rs1025488892 Chr.19: 9818946-9818946 on Build GRCh38
rs1051263228 Chr.19: 9819061-9819061 on Build GRCh38
rs866135450 Chr.19: 9818783-9818783 on Build GRCh38
rs903596088 Chr.19: 9818918-9818918 on Build GRCh38
rs560558701 Chr.19: 9818854-9818854 on Build GRCh38

View Full Product Details