Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10106412-10106898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPAN
Entrez Gene ID
56342
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
BXDC3, SSF, SSF-1, SSF1, SSF2
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10111299 on Build GRCh38
Transcripts
NM_020230.5 NM_001040664.2 NM_001198690.1

Gene Symbol
PPAN-P2RY11
Entrez Gene ID
692312
Gene Name
PPAN-P2RY11 readthrough
Gene Aliases
-
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10115389 on Build GRCh38
Transcripts
NM_020230.5 NM_001040664.2 NM_001198690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5035813 c.147C>T p.D49D Chr.19: 10106629-10106629 on Build GRCh38
7242353 c.30G>A p.Q10Q Chr.19: 10106512-10106512 on Build GRCh38
6374106 c.90C>A p.H30Q Chr.19: 10106572-10106572 on Build GRCh38
1480502 c.67G>C p.E23Q Chr.19: 10106549-10106549 on Build GRCh38
1480503 c.67G>C p.E23Q Chr.19: 10106549-10106549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019414292 Chr.19: 10106715-10106715 on Build GRCh38
rs953907519 Chr.19: 10106506-10106506 on Build GRCh38
rs771552032 Chr.19: 10106668-10106668 on Build GRCh38
rs11559190 Chr.19: 10106572-10106572 on Build GRCh38
rs749685792 Chr.19: 10106629-10106629 on Build GRCh38
rs771593184 Chr.19: 10106566-10106566 on Build GRCh38
rs938045227 Chr.19: 10106793-10106793 on Build GRCh38
rs985084174 Chr.19: 10106776-10106776 on Build GRCh38
rs944679778 Chr.19: 10106663-10106663 on Build GRCh38
rs371223447 Chr.19: 10106599-10106599 on Build GRCh38
rs781326420 Chr.19: 10106597-10106597 on Build GRCh38
rs757571904 Chr.19: 10106652-10106652 on Build GRCh38
rs1014539812 Chr.19: 10106751-10106751 on Build GRCh38
rs767389658 Chr.19: 10106442-10106442 on Build GRCh38
rs868140660 Chr.19: 10106637-10106637 on Build GRCh38
rs766957053 Chr.19: 10106642-10106643 on Build GRCh38
rs755035078 Chr.19: 10106524-10106524 on Build GRCh38
rs985166863 Chr.19: 10106578-10106578 on Build GRCh38
rs1054861239 Chr.19: 10106457-10106457 on Build GRCh38
rs542820412 Chr.19: 10106465-10106465 on Build GRCh38
rs555510856 Chr.19: 10106546-10106546 on Build GRCh38
rs1037184615 Chr.19: 10106740-10106740 on Build GRCh38
rs758704411 Chr.19: 10106488-10106488 on Build GRCh38
rs11559189 Chr.19: 10106662-10106662 on Build GRCh38
rs868499487 Chr.19: 10106638-10106638 on Build GRCh38
rs567422887 Chr.19: 10106874-10106874 on Build GRCh38
rs746435491 Chr.19: 10106677-10106677 on Build GRCh38
rs957207014 Chr.19: 10106642-10106642 on Build GRCh38
rs373410012 Chr.19: 10106458-10106458 on Build GRCh38
rs560157191 Chr.19: 10106743-10106743 on Build GRCh38
rs758992938 Chr.19: 10106560-10106560 on Build GRCh38
rs970453594 Chr.19: 10106611-10106611 on Build GRCh38
rs867784788 Chr.19: 10106636-10106636 on Build GRCh38
rs545316381 Chr.19: 10106734-10106734 on Build GRCh38
rs780354576 Chr.19: 10106490-10106490 on Build GRCh38
rs775333581 Chr.19: 10106569-10106569 on Build GRCh38
rs966398268 Chr.19: 10106468-10106468 on Build GRCh38
rs768070407 Chr.19: 10106801-10106802 on Build GRCh38
rs1049254030 Chr.19: 10106554-10106554 on Build GRCh38
rs755052688 Chr.19: 10106592-10106592 on Build GRCh38
rs966238402 Chr.19: 10106839-10106839 on Build GRCh38
rs28565143 Chr.19: 10106787-10106787 on Build GRCh38
rs375786293 Chr.19: 10106509-10106509 on Build GRCh38
rs887499995 Chr.19: 10106729-10106729 on Build GRCh38
rs889491581 Chr.19: 10106702-10106702 on Build GRCh38
rs778188593 Chr.19: 10106621-10106621 on Build GRCh38
rs756658300 Chr.19: 10106601-10106601 on Build GRCh38
rs976215248 Chr.19: 10106763-10106763 on Build GRCh38
rs868791919 Chr.19: 10106559-10106559 on Build GRCh38
rs868526544 Chr.19: 10106649-10106649 on Build GRCh38
rs1032021287 Chr.19: 10106484-10106484 on Build GRCh38
rs921622370 Chr.19: 10106690-10106690 on Build GRCh38
rs867921224 Chr.19: 10106647-10106647 on Build GRCh38
rs548724784 Chr.19: 10106842-10106842 on Build GRCh38
rs61064209 Chr.19: 10106704-10106704 on Build GRCh38
rs900165027 Chr.19: 10106522-10106522 on Build GRCh38
rs775075029 Chr.19: 10106671-10106671 on Build GRCh38
rs1040065680 Chr.19: 10106473-10106473 on Build GRCh38
rs905864181 Chr.19: 10106772-10106772 on Build GRCh38
rs866508542 Chr.19: 10106497-10106497 on Build GRCh38
rs768741863 Chr.19: 10106687-10106687 on Build GRCh38
rs757062197 Chr.19: 10106795-10106795 on Build GRCh38
rs1033073810 Chr.19: 10106622-10106622 on Build GRCh38
rs976028934 Chr.19: 10106867-10106867 on Build GRCh38
rs374860278 Chr.19: 10106557-10106557 on Build GRCh38
rs1020860711 Chr.19: 10106464-10106464 on Build GRCh38
rs112406563 Chr.19: 10106801-10106801 on Build GRCh38
rs1044949953 Chr.19: 10106678-10106678 on Build GRCh38
rs753687473 Chr.19: 10106574-10106574 on Build GRCh38
rs773955600 Chr.19: 10106558-10106558 on Build GRCh38
rs749597901 Chr.19: 10106552-10106552 on Build GRCh38
rs751806988 Chr.19: 10106587-10106587 on Build GRCh38
rs746663993 Chr.19: 10106730-10106730 on Build GRCh38
rs370147388 Chr.19: 10106540-10106540 on Build GRCh38
rs574092451 Chr.19: 10106694-10106694 on Build GRCh38
rs770123257 Chr.19: 10106545-10106545 on Build GRCh38
rs750728403 Chr.19: 10106469-10106469 on Build GRCh38
rs1046369211 Chr.19: 10106754-10106754 on Build GRCh38
rs760644168 Chr.19: 10106570-10106570 on Build GRCh38
rs1001914776 Chr.19: 10106782-10106782 on Build GRCh38
rs573733797 Chr.19: 10106556-10106556 on Build GRCh38
rs778589107 Chr.19: 10106659-10106659 on Build GRCh38
rs759021092 Chr.19: 10106436-10106436 on Build GRCh38
rs766729415 Chr.19: 10106586-10106586 on Build GRCh38
rs957466776 Chr.19: 10106789-10106789 on Build GRCh38
rs866124728 Chr.19: 10106624-10106624 on Build GRCh38
rs781598891 Chr.19: 10106536-10106536 on Build GRCh38
rs3841592 Chr.19: 10106766-10106766 on Build GRCh38
rs904727962 Chr.19: 10106683-10106683 on Build GRCh38
rs779615505 Chr.19: 10106670-10106670 on Build GRCh38
rs781390741 Chr.19: 10106847-10106847 on Build GRCh38
rs761656993 Chr.19: 10106584-10106584 on Build GRCh38
rs1002304897 Chr.19: 10106455-10106455 on Build GRCh38

View Full Product Details