Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10394943-10395450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37
Entrez Gene ID
11140
Gene Name
cell division cycle 37
Gene Aliases
P50CDC37
UniGene
Hs.160958
Gene Chromosome Location
Chr.19: 10391128-10403595 on Build GRCh38
Transcripts
NM_007065.3 XM_011527652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1711807 c.691C>T p.R231W Chr.19: 10395056-10395056 on Build GRCh38
7198661 c.526T>A p.S176T Chr.19: 10395305-10395305 on Build GRCh38
5972674 c.522C>T p.Y174Y Chr.19: 10395309-10395309 on Build GRCh38
4503656 c.645C>A p.V215V Chr.19: 10395102-10395102 on Build GRCh38
5549080 c.630C>T p.A210A Chr.19: 10395117-10395117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937100439 Chr.19: 10395162-10395162 on Build GRCh38
rs931105175 Chr.19: 10395005-10395005 on Build GRCh38
rs754408676 Chr.19: 10395179-10395179 on Build GRCh38
rs147547295 Chr.19: 10395107-10395107 on Build GRCh38
rs992473284 Chr.19: 10395166-10395166 on Build GRCh38
rs746798040 Chr.19: 10395156-10395161 on Build GRCh38
rs1025592298 Chr.19: 10395298-10395298 on Build GRCh38
rs916908422 Chr.19: 10395421-10395421 on Build GRCh38
rs779190160 Chr.19: 10395056-10395056 on Build GRCh38
rs764498981 Chr.19: 10395239-10395239 on Build GRCh38
rs751802744 Chr.19: 10395397-10395397 on Build GRCh38
rs753219911 Chr.19: 10395015-10395015 on Build GRCh38
rs139416692 Chr.19: 10395282-10395282 on Build GRCh38
rs756698559 Chr.19: 10395418-10395418 on Build GRCh38
rs746133397 Chr.19: 10395065-10395065 on Build GRCh38
rs771877493 Chr.19: 10395096-10395096 on Build GRCh38
rs146531345 Chr.19: 10395409-10395409 on Build GRCh38
rs529125841 Chr.19: 10394999-10394999 on Build GRCh38
rs943982488 Chr.19: 10395370-10395370 on Build GRCh38
rs150028004 Chr.19: 10395285-10395285 on Build GRCh38
rs931156064 Chr.19: 10395385-10395385 on Build GRCh38
rs994076132 Chr.19: 10395292-10395292 on Build GRCh38
rs189376825 Chr.19: 10395384-10395384 on Build GRCh38
rs375165732 Chr.19: 10395228-10395228 on Build GRCh38
rs762179256 Chr.19: 10395365-10395365 on Build GRCh38
rs768977400 Chr.19: 10395349-10395349 on Build GRCh38
rs761037786 Chr.19: 10395164-10395164 on Build GRCh38
rs757313823 Chr.19: 10395180-10395180 on Build GRCh38
rs768657748 Chr.19: 10395110-10395110 on Build GRCh38
rs752951399 Chr.19: 10395163-10395163 on Build GRCh38
rs754800877 Chr.19: 10395296-10395296 on Build GRCh38
rs761019690 Chr.19: 10395235-10395235 on Build GRCh38
rs762929807 Chr.19: 10395386-10395386 on Build GRCh38
rs770538573 Chr.19: 10395331-10395331 on Build GRCh38
rs750412756 Chr.19: 10395281-10395281 on Build GRCh38
rs766532701 Chr.19: 10395392-10395392 on Build GRCh38
rs773978075 Chr.19: 10395332-10395332 on Build GRCh38
rs55826704 Chr.19: 10395313-10395313 on Build GRCh38
rs965254505 Chr.19: 10395104-10395104 on Build GRCh38
rs765803614 Chr.19: 10395278-10395278 on Build GRCh38
rs768746057 Chr.19: 10394976-10394976 on Build GRCh38
rs370095608 Chr.19: 10395408-10395408 on Build GRCh38
rs372486429 Chr.19: 10395209-10395209 on Build GRCh38
rs769403695 Chr.19: 10395288-10395288 on Build GRCh38
rs772560649 Chr.19: 10395210-10395210 on Build GRCh38
rs781265000 Chr.19: 10395205-10395205 on Build GRCh38
rs778775213 Chr.19: 10395406-10395406 on Build GRCh38
rs776939474 Chr.19: 10395350-10395350 on Build GRCh38
rs761422135 Chr.19: 10394989-10394989 on Build GRCh38
rs774280690 Chr.19: 10395196-10395196 on Build GRCh38
rs761405698 Chr.19: 10395147-10395147 on Build GRCh38
rs140745389 Chr.19: 10395044-10395044 on Build GRCh38
rs779687284 Chr.19: 10395427-10395427 on Build GRCh38
rs367995641 Chr.19: 10395412-10395412 on Build GRCh38
rs769489821 Chr.19: 10395149-10395149 on Build GRCh38
rs373797879 Chr.19: 10394986-10394986 on Build GRCh38
rs952556654 Chr.19: 10395154-10395154 on Build GRCh38
rs771052216 Chr.19: 10395180-10395180 on Build GRCh38
rs550795142 Chr.19: 10395010-10395010 on Build GRCh38
rs371406069 Chr.19: 10395211-10395211 on Build GRCh38
rs755960486 Chr.19: 10395310-10395310 on Build GRCh38
rs199725202 Chr.19: 10395410-10395410 on Build GRCh38
rs147241364 Chr.19: 10395043-10395043 on Build GRCh38
rs912470360 Chr.19: 10394966-10394966 on Build GRCh38
rs754888704 Chr.19: 10395198-10395198 on Build GRCh38
rs781175114 Chr.19: 10395299-10395299 on Build GRCh38
rs770666678 Chr.19: 10395184-10395184 on Build GRCh38
rs570835276 Chr.19: 10395382-10395382 on Build GRCh38
rs776688491 Chr.19: 10395210-10395210 on Build GRCh38
rs748148831 Chr.19: 10395207-10395207 on Build GRCh38
rs376983797 Chr.19: 10395105-10395105 on Build GRCh38
rs777098602 Chr.19: 10395243-10395243 on Build GRCh38
rs766090745 Chr.19: 10395157-10395157 on Build GRCh38
rs943862482 Chr.19: 10394983-10394983 on Build GRCh38
rs755937670 Chr.19: 10395411-10395411 on Build GRCh38
rs369440357 Chr.19: 10395132-10395132 on Build GRCh38
rs758501401 Chr.19: 10395193-10395193 on Build GRCh38
rs764515365 Chr.19: 10395178-10395178 on Build GRCh38
rs762557613 Chr.19: 10395151-10395151 on Build GRCh38
rs746736521 Chr.19: 10395195-10395195 on Build GRCh38
rs759934665 Chr.19: 10395007-10395007 on Build GRCh38
rs927041029 Chr.19: 10395160-10395160 on Build GRCh38
rs771663570 Chr.19: 10395426-10395426 on Build GRCh38
rs776716837 Chr.19: 10395135-10395135 on Build GRCh38
rs765597038 Chr.19: 10395371-10395371 on Build GRCh38
rs777775938 Chr.19: 10395325-10395325 on Build GRCh38
rs780337347 Chr.19: 10395194-10395194 on Build GRCh38
rs745566999 Chr.19: 10395334-10395334 on Build GRCh38
rs770427825 Chr.19: 10395220-10395220 on Build GRCh38
rs866354313 Chr.19: 10395335-10395335 on Build GRCh38
rs762346805 Chr.19: 10395255-10395255 on Build GRCh38
rs372127896 Chr.19: 10394977-10394977 on Build GRCh38
rs376326177 Chr.19: 10395425-10395425 on Build GRCh38
rs750112484 Chr.19: 10395002-10395002 on Build GRCh38
rs778908667 Chr.19: 10395186-10395186 on Build GRCh38
rs760577381 Chr.19: 10394974-10394974 on Build GRCh38
rs975254816 Chr.19: 10395380-10395380 on Build GRCh38
rs372000706 Chr.19: 10395424-10395424 on Build GRCh38
rs748675716 Chr.19: 10395328-10395328 on Build GRCh38
rs201914397 Chr.19: 10395302-10395302 on Build GRCh38
rs757330227 Chr.19: 10395055-10395055 on Build GRCh38
rs750627622 Chr.19: 10395192-10395192 on Build GRCh38

View Full Product Details