Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10517295-10517791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S1PR5
Entrez Gene ID
53637
Gene Name
sphingosine-1-phosphate receptor 5
Gene Aliases
EDG8, Edg-8, S1P5, SPPR-1, SPPR-2
UniGene
Hs.501561
Gene Chromosome Location
Chr.19: 10512742-10517992 on Build GRCh38
Transcripts
NM_001166215.1 NM_030760.4

SNP Detail

SNP ID Chromosome Location
rs940789067 Chr.19: 10517464-10517464 on Build GRCh38
rs903593344 Chr.19: 10517622-10517622 on Build GRCh38
rs905959773 Chr.19: 10517367-10517367 on Build GRCh38
rs1020658897 Chr.19: 10517458-10517458 on Build GRCh38
rs953762399 Chr.19: 10517728-10517728 on Build GRCh38
rs569031050 Chr.19: 10517764-10517764 on Build GRCh38
rs1010228916 Chr.19: 10517451-10517451 on Build GRCh38
rs147279753 Chr.19: 10517489-10517489 on Build GRCh38
rs953149058 Chr.19: 10517505-10517505 on Build GRCh38
rs539753931 Chr.19: 10517402-10517402 on Build GRCh38
rs541554572 Chr.19: 10517357-10517357 on Build GRCh38
rs892269231 Chr.19: 10517590-10517590 on Build GRCh38
rs1027967119 Chr.19: 10517727-10517727 on Build GRCh38
rs926543872 Chr.19: 10517542-10517542 on Build GRCh38
rs372358570 Chr.19: 10517361-10517361 on Build GRCh38
rs1028752176 Chr.19: 10517498-10517498 on Build GRCh38
rs893216492 Chr.19: 10517625-10517625 on Build GRCh38
rs901767881 Chr.19: 10517668-10517668 on Build GRCh38
rs899139579 Chr.19: 10517319-10517319 on Build GRCh38
rs971719513 Chr.19: 10517548-10517548 on Build GRCh38
rs997362554 Chr.19: 10517689-10517689 on Build GRCh38
rs950301750 Chr.19: 10517565-10517565 on Build GRCh38
rs888975971 Chr.19: 10517721-10517721 on Build GRCh38
rs1043413125 Chr.19: 10517701-10517701 on Build GRCh38
rs975088367 Chr.19: 10517399-10517399 on Build GRCh38
rs946040494 Chr.19: 10517647-10517647 on Build GRCh38
rs940348505 Chr.19: 10517339-10517339 on Build GRCh38
rs10418435 Chr.19: 10517365-10517365 on Build GRCh38
rs1041685360 Chr.19: 10517649-10517649 on Build GRCh38
rs927469437 Chr.19: 10517606-10517606 on Build GRCh38
rs939257683 Chr.19: 10517558-10517558 on Build GRCh38
rs879577498 Chr.19: 10517656-10517656 on Build GRCh38
rs866637447 Chr.19: 10517601-10517601 on Build GRCh38
rs1015959873 Chr.19: 10517525-10517525 on Build GRCh38
rs908856666 Chr.19: 10517337-10517337 on Build GRCh38
rs1034830157 Chr.19: 10517355-10517355 on Build GRCh38
rs1006009486 Chr.19: 10517747-10517747 on Build GRCh38
rs369671742 Chr.19: 10517562-10517562 on Build GRCh38
rs901720028 Chr.19: 10517481-10517481 on Build GRCh38
rs547315572 Chr.19: 10517720-10517720 on Build GRCh38
rs550638538 Chr.19: 10517759-10517759 on Build GRCh38
rs1056312851 Chr.19: 10517575-10517575 on Build GRCh38
rs903546820 Chr.19: 10517723-10517723 on Build GRCh38
rs1003813391 Chr.19: 10517344-10517344 on Build GRCh38
rs984527240 Chr.19: 10517507-10517507 on Build GRCh38
rs893156680 Chr.19: 10517414-10517414 on Build GRCh38
rs529322332 Chr.19: 10517752-10517752 on Build GRCh38
rs930659875 Chr.19: 10517522-10517522 on Build GRCh38
rs528782049 Chr.19: 10517536-10517536 on Build GRCh38
rs984432404 Chr.19: 10517318-10517318 on Build GRCh38
rs930639979 Chr.19: 10517321-10517321 on Build GRCh38
rs981625189 Chr.19: 10517567-10517567 on Build GRCh38
rs937481035 Chr.19: 10517620-10517620 on Build GRCh38
rs973430670 Chr.19: 10517350-10517350 on Build GRCh38
rs967740219 Chr.19: 10517372-10517372 on Build GRCh38
rs1054577296 Chr.19: 10517623-10517623 on Build GRCh38
rs1009394908 Chr.19: 10517615-10517615 on Build GRCh38
rs546433617 Chr.19: 10517407-10517407 on Build GRCh38
rs912023342 Chr.19: 10517428-10517428 on Build GRCh38
rs182112172 Chr.19: 10517751-10517751 on Build GRCh38
rs1009118419 Chr.19: 10517742-10517742 on Build GRCh38

View Full Product Details