Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11102283-11102807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLR
Entrez Gene ID
3949
Gene Name
low density lipoprotein receptor
Gene Aliases
FH, FHC, LDLCQ2
UniGene
Hs.213289
Gene Chromosome Location
Chr.19: 11089362-11133830 on Build GRCh38
Transcripts
NM_000527.4 NM_001195798.1 NM_001195799.1 NM_001195800.1 NM_001195803.1 XM_011528010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6700288 c.313+1G>A p.? Chr.19: 11102787-11102787 on Build GRCh38
6700304 c.301G>A p.E101K Chr.19: 11102774-11102774 on Build GRCh38
4074072 c.267C>T p.C89C Chr.19: 11102740-11102740 on Build GRCh38
4074071 c.242G>A p.R81H Chr.19: 11102715-11102715 on Build GRCh38
990774 c.282C>T p.D94D Chr.19: 11102755-11102755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879254450 Chr.19: 11102728-11102728 on Build GRCh38
rs529775182 Chr.19: 11102319-11102319 on Build GRCh38
rs879254452 Chr.19: 11102733-11102733 on Build GRCh38
rs180836942 Chr.19: 11102472-11102472 on Build GRCh38
rs879254447 Chr.19: 11102717-11102717 on Build GRCh38
rs576744963 Chr.19: 11102564-11102564 on Build GRCh38
rs750718763 Chr.19: 11102695-11102695 on Build GRCh38
rs17249225 Chr.19: 11102363-11102363 on Build GRCh38
rs879254437 Chr.19: 11102677-11102677 on Build GRCh38
rs574337291 Chr.19: 11102699-11102699 on Build GRCh38
rs879254444 Chr.19: 11102713-11102713 on Build GRCh38
rs958169798 Chr.19: 11102526-11102526 on Build GRCh38
rs758927662 Chr.19: 11102698-11102698 on Build GRCh38
rs766709484 Chr.19: 11102773-11102773 on Build GRCh38
rs973134754 Chr.19: 11102556-11102556 on Build GRCh38
rs940127734 Chr.19: 11102553-11102553 on Build GRCh38
rs559091708 Chr.19: 11102610-11102610 on Build GRCh38
rs183134488 Chr.19: 11102782-11102782 on Build GRCh38
rs563743121 Chr.19: 11102330-11102330 on Build GRCh38
rs879254433 Chr.19: 11102663-11102663 on Build GRCh38
rs897909667 Chr.19: 11102443-11102443 on Build GRCh38
rs139400379 Chr.19: 11102758-11102758 on Build GRCh38
rs898709202 Chr.19: 11102385-11102385 on Build GRCh38
rs372845091 Chr.19: 11102764-11102764 on Build GRCh38
rs879254461 Chr.19: 11102774-11102774 on Build GRCh38
rs875989895 Chr.19: 11102784-11102784 on Build GRCh38
rs879254456 Chr.19: 11102756-11102756 on Build GRCh38
rs879254446 Chr.19: 11102717-11102717 on Build GRCh38
rs745776977 Chr.19: 11102753-11102753 on Build GRCh38
rs1032460394 Chr.19: 11102474-11102474 on Build GRCh38
rs377271627 Chr.19: 11102769-11102769 on Build GRCh38
rs875989896 Chr.19: 11102786-11102787 on Build GRCh38
rs879254435 Chr.19: 11102668-11102669 on Build GRCh38
rs774016801 Chr.19: 11102664-11102664 on Build GRCh38
rs938806788 Chr.19: 11102681-11102681 on Build GRCh38
rs771019366 Chr.19: 11102742-11102742 on Build GRCh38
rs975764282 Chr.19: 11102595-11102595 on Build GRCh38
rs190725941 Chr.19: 11102466-11102466 on Build GRCh38
rs765699402 Chr.19: 11102692-11102692 on Build GRCh38
rs13306510 Chr.19: 11102786-11102786 on Build GRCh38
rs879254442 Chr.19: 11102705-11102705 on Build GRCh38
rs144172724 Chr.19: 11102774-11102774 on Build GRCh38
rs879254445 Chr.19: 11102715-11102716 on Build GRCh38
rs879254459 Chr.19: 11102771-11102771 on Build GRCh38
rs879254431 Chr.19: 11102410-11102415 on Build GRCh38
rs766903209 Chr.19: 11102684-11102684 on Build GRCh38
rs774858755 Chr.19: 11102759-11102759 on Build GRCh38
rs730882078 Chr.19: 11102714-11102714 on Build GRCh38
rs879254451 Chr.19: 11102730-11102738 on Build GRCh38
rs1040447339 Chr.19: 11102378-11102378 on Build GRCh38
rs112029328 Chr.19: 11102787-11102787 on Build GRCh38
rs879254436 Chr.19: 11102672-11102673 on Build GRCh38
rs775255244 Chr.19: 11102685-11102685 on Build GRCh38
rs879254458 Chr.19: 11102769-11102770 on Build GRCh38
rs887631980 Chr.19: 11102425-11102425 on Build GRCh38
rs922966892 Chr.19: 11102603-11102603 on Build GRCh38
rs374168004 Chr.19: 11102614-11102614 on Build GRCh38
rs875989894 Chr.19: 11102739-11102739 on Build GRCh38
rs748118639 Chr.19: 11102633-11102633 on Build GRCh38
rs979669428 Chr.19: 11102319-11102320 on Build GRCh38
rs879254438 Chr.19: 11102687-11102687 on Build GRCh38
rs896252141 Chr.19: 11102465-11102465 on Build GRCh38
rs121908025 Chr.19: 11102732-11102732 on Build GRCh38
rs774467219 Chr.19: 11102747-11102747 on Build GRCh38
rs879254453 Chr.19: 11102734-11102735 on Build GRCh38
rs931918651 Chr.19: 11102388-11102388 on Build GRCh38
rs879254432 Chr.19: 11102662-11102662 on Build GRCh38
rs989888307 Chr.19: 11102338-11102338 on Build GRCh38
rs749245318 Chr.19: 11102649-11102649 on Build GRCh38
rs535051620 Chr.19: 11102383-11102383 on Build GRCh38
rs541587443 Chr.19: 11102356-11102356 on Build GRCh38
rs879254462 Chr.19: 11102778-11102778 on Build GRCh38
rs770931363 Chr.19: 11102659-11102659 on Build GRCh38
rs372828849 Chr.19: 11102743-11102743 on Build GRCh38
rs555338272 Chr.19: 11102562-11102562 on Build GRCh38
rs74179967 Chr.19: 11102371-11102371 on Build GRCh38
rs879254443 Chr.19: 11102709-11102710 on Build GRCh38
rs931614846 Chr.19: 11102395-11102395 on Build GRCh38
rs875989890 Chr.19: 11102697-11102697 on Build GRCh38
rs576204177 Chr.19: 11102480-11102480 on Build GRCh38
rs769691180 Chr.19: 11102638-11102638 on Build GRCh38
rs759075066 Chr.19: 11102668-11102668 on Build GRCh38
rs760378478 Chr.19: 11102762-11102762 on Build GRCh38
rs547030615 Chr.19: 11102439-11102439 on Build GRCh38
rs912986005 Chr.19: 11102339-11102339 on Build GRCh38
rs879254465 Chr.19: 11102785-11102785 on Build GRCh38
rs986072954 Chr.19: 11102623-11102623 on Build GRCh38
rs879254460 Chr.19: 11102772-11102772 on Build GRCh38
rs986876236 Chr.19: 11102337-11102337 on Build GRCh38
rs531128076 Chr.19: 11102348-11102348 on Build GRCh38
rs1006073690 Chr.19: 11102429-11102429 on Build GRCh38
rs879254439 Chr.19: 11102696-11102696 on Build GRCh38
rs769534124 Chr.19: 11102471-11102471 on Build GRCh38
rs879254463 Chr.19: 11102783-11102786 on Build GRCh38
rs875989893 Chr.19: 11102726-11102726 on Build GRCh38
rs575172638 Chr.19: 11102594-11102594 on Build GRCh38
rs875989892 Chr.19: 11102724-11102724 on Build GRCh38
rs370860696 Chr.19: 11102705-11102705 on Build GRCh38
rs146675823 Chr.19: 11102706-11102706 on Build GRCh38
rs875989891 Chr.19: 11102719-11102719 on Build GRCh38
rs1057516129 Chr.19: 11102706-11102706 on Build GRCh38
rs372094141 Chr.19: 11102648-11102648 on Build GRCh38
rs1050245577 Chr.19: 11102391-11102391 on Build GRCh38
rs879254454 Chr.19: 11102737-11102737 on Build GRCh38
rs879254466 Chr.19: 11102787-11102788 on Build GRCh38
rs875989889 Chr.19: 11102669-11102670 on Build GRCh38
rs879254448 Chr.19: 11102718-11102718 on Build GRCh38
rs544203837 Chr.19: 11102662-11102662 on Build GRCh38
rs548224532 Chr.19: 11102321-11102321 on Build GRCh38
rs199570811 Chr.19: 11102734-11102734 on Build GRCh38
rs879254449 Chr.19: 11102723-11102723 on Build GRCh38
rs552948476 Chr.19: 11102352-11102352 on Build GRCh38
rs781518425 Chr.19: 11102627-11102627 on Build GRCh38
rs1044579531 Chr.19: 11102397-11102397 on Build GRCh38
rs879254457 Chr.19: 11102757-11102757 on Build GRCh38
rs879254434 Chr.19: 11102666-11102674 on Build GRCh38
rs563390335 Chr.19: 11102777-11102777 on Build GRCh38
rs777640882 Chr.19: 11102740-11102740 on Build GRCh38
rs568413051 Chr.19: 11102464-11102464 on Build GRCh38
rs879254441 Chr.19: 11102703-11102704 on Build GRCh38
rs190990789 Chr.19: 11102561-11102561 on Build GRCh38
rs879254455 Chr.19: 11102738-11102738 on Build GRCh38
rs760229862 Chr.19: 11102687-11102687 on Build GRCh38
rs750474121 Chr.19: 11102765-11102765 on Build GRCh38
rs879254464 Chr.19: 11102783-11102783 on Build GRCh38
rs979342361 Chr.19: 11102525-11102525 on Build GRCh38
rs764797225 Chr.19: 11102776-11102776 on Build GRCh38
rs185255158 Chr.19: 11102551-11102551 on Build GRCh38
rs200660051 Chr.19: 11102735-11102735 on Build GRCh38
rs17242850 Chr.19: 11102497-11102497 on Build GRCh38
rs1013556017 Chr.19: 11102448-11102448 on Build GRCh38
rs368074156 Chr.19: 11102635-11102635 on Build GRCh38
rs953852884 Chr.19: 11102550-11102550 on Build GRCh38
rs749038326 Chr.19: 11102741-11102741 on Build GRCh38
rs879254440 Chr.19: 11102703-11102703 on Build GRCh38

View Full Product Details