Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11172839-11173379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK2
Entrez Gene ID
25959
Gene Name
KN motif and ankyrin repeat domains 2
Gene Aliases
ANKRD25, MXRA3, PPKWH, SIP
UniGene
Hs.284208
Gene Chromosome Location
Chr.19: 11164267-11197567 on Build GRCh38
Transcripts
NM_001136191.2 NM_015493.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6277348 c.2176G>A p.D726N Chr.19: 11173040-11173040 on Build GRCh38
5755243 c.2105T>G p.V702G Chr.19: 11173111-11173111 on Build GRCh38
3692344 c.2124C>T p.T708T Chr.19: 11173068-11173068 on Build GRCh38
438535 c.2207G>C p.S736T Chr.19: 11172985-11172985 on Build GRCh38
6690000 c.2194A>G p.N732D Chr.19: 11172998-11172998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576348444 Chr.19: 11172968-11172968 on Build GRCh38
rs763338230 Chr.19: 11173160-11173160 on Build GRCh38
rs889525922 Chr.19: 11172917-11172917 on Build GRCh38
rs771164084 Chr.19: 11173034-11173034 on Build GRCh38
rs143339573 Chr.19: 11173097-11173097 on Build GRCh38
rs758993599 Chr.19: 11173087-11173087 on Build GRCh38
rs773071028 Chr.19: 11173053-11173053 on Build GRCh38
rs901214147 Chr.19: 11173306-11173306 on Build GRCh38
rs772183646 Chr.19: 11172944-11172944 on Build GRCh38
rs4804149 Chr.19: 11173352-11173352 on Build GRCh38
rs372687614 Chr.19: 11173089-11173089 on Build GRCh38
rs751200516 Chr.19: 11173177-11173177 on Build GRCh38
rs138941748 Chr.19: 11173012-11173012 on Build GRCh38
rs965630736 Chr.19: 11173081-11173081 on Build GRCh38
rs376926134 Chr.19: 11173096-11173096 on Build GRCh38
rs149228688 Chr.19: 11172937-11172937 on Build GRCh38
rs746006286 Chr.19: 11172943-11172943 on Build GRCh38
rs760409016 Chr.19: 11173149-11173149 on Build GRCh38
rs750720130 Chr.19: 11173135-11173135 on Build GRCh38
rs531057207 Chr.19: 11173321-11173321 on Build GRCh38
rs893912974 Chr.19: 11173222-11173222 on Build GRCh38
rs377487306 Chr.19: 11173143-11173143 on Build GRCh38
rs375561800 Chr.19: 11173035-11173035 on Build GRCh38
rs780475485 Chr.19: 11172946-11172946 on Build GRCh38
rs775823105 Chr.19: 11173005-11173005 on Build GRCh38
rs773196270 Chr.19: 11172970-11172970 on Build GRCh38
rs758979865 Chr.19: 11173065-11173065 on Build GRCh38
rs773865789 Chr.19: 11173061-11173061 on Build GRCh38
rs762873476 Chr.19: 11173142-11173143 on Build GRCh38
rs189623046 Chr.19: 11173186-11173186 on Build GRCh38
rs369583093 Chr.19: 11172975-11172975 on Build GRCh38
rs1013428935 Chr.19: 11173019-11173019 on Build GRCh38
rs897077132 Chr.19: 11172934-11172934 on Build GRCh38
rs201723514 Chr.19: 11173122-11173122 on Build GRCh38
rs200093308 Chr.19: 11172967-11172967 on Build GRCh38
rs767055554 Chr.19: 11173161-11173161 on Build GRCh38
rs577079831 Chr.19: 11173150-11173150 on Build GRCh38
rs73504667 Chr.19: 11173193-11173193 on Build GRCh38
rs775199754 Chr.19: 11173068-11173068 on Build GRCh38
rs762427020 Chr.19: 11172973-11172973 on Build GRCh38
rs765607833 Chr.19: 11173082-11173082 on Build GRCh38
rs1050640382 Chr.19: 11172954-11172954 on Build GRCh38
rs1046223879 Chr.19: 11173026-11173026 on Build GRCh38
rs965598518 Chr.19: 11172924-11172924 on Build GRCh38
rs181846053 Chr.19: 11172894-11172894 on Build GRCh38
rs779800581 Chr.19: 11173162-11173162 on Build GRCh38
rs1046783449 Chr.19: 11173187-11173187 on Build GRCh38
rs777595303 Chr.19: 11173239-11173239 on Build GRCh38
rs200538226 Chr.19: 11172942-11172942 on Build GRCh38
rs769618777 Chr.19: 11172969-11172969 on Build GRCh38
rs888346895 Chr.19: 11173296-11173296 on Build GRCh38
rs370976104 Chr.19: 11173159-11173159 on Build GRCh38
rs988132136 Chr.19: 11172908-11172908 on Build GRCh38
rs563033225 Chr.19: 11173252-11173252 on Build GRCh38
rs908981446 Chr.19: 11173108-11173108 on Build GRCh38
rs756387589 Chr.19: 11173176-11173176 on Build GRCh38
rs144561976 Chr.19: 11173025-11173025 on Build GRCh38
rs749216928 Chr.19: 11173055-11173055 on Build GRCh38
rs770860594 Chr.19: 11173120-11173120 on Build GRCh38
rs777653062 Chr.19: 11173111-11173111 on Build GRCh38
rs763602191 Chr.19: 11173151-11173151 on Build GRCh38
rs775817924 Chr.19: 11173308-11173308 on Build GRCh38
rs750023451 Chr.19: 11172910-11172910 on Build GRCh38
rs775048871 Chr.19: 11173148-11173148 on Build GRCh38
rs146334556 Chr.19: 11173093-11173093 on Build GRCh38
rs779350628 Chr.19: 11173013-11173013 on Build GRCh38
rs747204915 Chr.19: 11172948-11172948 on Build GRCh38
rs778143762 Chr.19: 11172936-11172936 on Build GRCh38
rs759810616 Chr.19: 11173257-11173258 on Build GRCh38
rs931753981 Chr.19: 11172951-11172951 on Build GRCh38
rs868612770 Chr.19: 11173202-11173202 on Build GRCh38
rs750923584 Chr.19: 11173029-11173029 on Build GRCh38
rs752050429 Chr.19: 11173040-11173040 on Build GRCh38
rs185203180 Chr.19: 11173138-11173138 on Build GRCh38
rs564521244 Chr.19: 11173197-11173197 on Build GRCh38
rs781419443 Chr.19: 11173305-11173305 on Build GRCh38
rs758208101 Chr.19: 11173231-11173231 on Build GRCh38
rs374892729 Chr.19: 11172963-11172963 on Build GRCh38
rs138964602 Chr.19: 11173185-11173185 on Build GRCh38
rs373009999 Chr.19: 11172995-11172995 on Build GRCh38
rs954302290 Chr.19: 11173332-11173332 on Build GRCh38
rs1048329485 Chr.19: 11173269-11173269 on Build GRCh38
rs926069953 Chr.19: 11172865-11172865 on Build GRCh38
rs759210703 Chr.19: 11172998-11172998 on Build GRCh38
rs752098596 Chr.19: 11173167-11173167 on Build GRCh38
rs148739151 Chr.19: 11173042-11173042 on Build GRCh38
rs762136808 Chr.19: 11173071-11173071 on Build GRCh38
rs958317506 Chr.19: 11172870-11172870 on Build GRCh38
rs377363480 Chr.19: 11172993-11172993 on Build GRCh38
rs1001343713 Chr.19: 11173315-11173315 on Build GRCh38
rs117383959 Chr.19: 11173347-11173347 on Build GRCh38
rs750715955 Chr.19: 11173083-11173083 on Build GRCh38
rs767224525 Chr.19: 11173067-11173067 on Build GRCh38
rs768956638 Chr.19: 11172955-11172955 on Build GRCh38
rs769617642 Chr.19: 11173043-11173043 on Build GRCh38
rs766932665 Chr.19: 11173088-11173088 on Build GRCh38
rs958133088 Chr.19: 11173057-11173057 on Build GRCh38
rs1040690612 Chr.19: 11172925-11172925 on Build GRCh38
rs755900841 Chr.19: 11173098-11173098 on Build GRCh38
rs748980415 Chr.19: 11173116-11173116 on Build GRCh38
rs955595985 Chr.19: 11173039-11173039 on Build GRCh38
rs1029021304 Chr.19: 11173329-11173329 on Build GRCh38
rs141494405 Chr.19: 11173060-11173060 on Build GRCh38
rs200321828 Chr.19: 11173041-11173041 on Build GRCh38

View Full Product Details