Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34300670-34301167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0355
Entrez Gene ID
9710
Gene Name
KIAA0355
Gene Aliases
-
UniGene
Hs.330073
Gene Chromosome Location
Chr.19: 34254551-34355566 on Build GRCh38
Transcripts
NM_014686.3 XM_005259444.1 XM_017027525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234256 c.466C>T p.Q156* Chr.19: 34300939-34300939 on Build GRCh38
5964878 c.495+2T>C p.? Chr.19: 34300970-34300970 on Build GRCh38
3532290 c.472G>A p.E158K Chr.19: 34300945-34300945 on Build GRCh38
4076831 c.223G>A p.D75N Chr.19: 34300696-34300696 on Build GRCh38
710357 c.431T>A p.M144K Chr.19: 34300904-34300904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764020643 Chr.19: 34300696-34300696 on Build GRCh38
rs768101617 Chr.19: 34300977-34300977 on Build GRCh38
rs146815070 Chr.19: 34300830-34300830 on Build GRCh38
rs753131048 Chr.19: 34300826-34300826 on Build GRCh38
rs754003690 Chr.19: 34300729-34300729 on Build GRCh38
rs891316055 Chr.19: 34301057-34301057 on Build GRCh38
rs760940741 Chr.19: 34300799-34300799 on Build GRCh38
rs770572065 Chr.19: 34300755-34300755 on Build GRCh38
rs778258992 Chr.19: 34300748-34300748 on Build GRCh38
rs757281425 Chr.19: 34300872-34300872 on Build GRCh38
rs376785044 Chr.19: 34300806-34300806 on Build GRCh38
rs773154057 Chr.19: 34301006-34301006 on Build GRCh38
rs893745805 Chr.19: 34301095-34301095 on Build GRCh38
rs751405374 Chr.19: 34300798-34300798 on Build GRCh38
rs747401571 Chr.19: 34300842-34300842 on Build GRCh38
rs750602156 Chr.19: 34300699-34300699 on Build GRCh38
rs769388748 Chr.19: 34300784-34300784 on Build GRCh38
rs529496099 Chr.19: 34300973-34300973 on Build GRCh38
rs892222973 Chr.19: 34301122-34301122 on Build GRCh38
rs903192757 Chr.19: 34300708-34300708 on Build GRCh38
rs1052346063 Chr.19: 34301087-34301087 on Build GRCh38
rs950097548 Chr.19: 34300997-34300997 on Build GRCh38
rs376530523 Chr.19: 34300990-34300990 on Build GRCh38
rs759067421 Chr.19: 34300783-34300783 on Build GRCh38
rs369572498 Chr.19: 34300740-34300740 on Build GRCh38
rs111551327 Chr.19: 34300790-34300790 on Build GRCh38
rs746152945 Chr.19: 34300978-34300978 on Build GRCh38
rs777929074 Chr.19: 34300833-34300833 on Build GRCh38
rs768671183 Chr.19: 34300875-34300875 on Build GRCh38
rs757616651 Chr.19: 34300947-34300947 on Build GRCh38
rs752106417 Chr.19: 34300940-34300940 on Build GRCh38
rs755800148 Chr.19: 34300963-34300963 on Build GRCh38
rs779115366 Chr.19: 34300811-34300811 on Build GRCh38
rs1010732348 Chr.19: 34301114-34301114 on Build GRCh38
rs181213766 Chr.19: 34301094-34301094 on Build GRCh38
rs528640076 Chr.19: 34301107-34301107 on Build GRCh38
rs780217364 Chr.19: 34300718-34300718 on Build GRCh38
rs373316791 Chr.19: 34300864-34300864 on Build GRCh38
rs759859175 Chr.19: 34300916-34300916 on Build GRCh38
rs759610631 Chr.19: 34301009-34301009 on Build GRCh38
rs770519034 Chr.19: 34300855-34300855 on Build GRCh38
rs530620364 Chr.19: 34300890-34300890 on Build GRCh38
rs781598440 Chr.19: 34300949-34300949 on Build GRCh38
rs763063958 Chr.19: 34301016-34301016 on Build GRCh38
rs745589597 Chr.19: 34300862-34300862 on Build GRCh38
rs765115581 Chr.19: 34300693-34300693 on Build GRCh38
rs146007478 Chr.19: 34301066-34301066 on Build GRCh38
rs756418226 Chr.19: 34300713-34300713 on Build GRCh38
rs980676510 Chr.19: 34300801-34300801 on Build GRCh38
rs62122214 Chr.19: 34300917-34300917 on Build GRCh38
rs776076607 Chr.19: 34300780-34300780 on Build GRCh38
rs149799888 Chr.19: 34300738-34300738 on Build GRCh38
rs767458251 Chr.19: 34300896-34300896 on Build GRCh38
rs764603418 Chr.19: 34300935-34300935 on Build GRCh38
rs754107575 Chr.19: 34300913-34300913 on Build GRCh38
rs140551420 Chr.19: 34300876-34300876 on Build GRCh38
rs932936766 Chr.19: 34301007-34301007 on Build GRCh38
rs572714541 Chr.19: 34300794-34300794 on Build GRCh38
rs200592725 Chr.19: 34300757-34300757 on Build GRCh38
rs752731347 Chr.19: 34300694-34300694 on Build GRCh38
rs779391650 Chr.19: 34300972-34300972 on Build GRCh38
rs953283950 Chr.19: 34300785-34300785 on Build GRCh38
rs908640012 Chr.19: 34300888-34300888 on Build GRCh38
rs941410226 Chr.19: 34300897-34300897 on Build GRCh38
rs778928865 Chr.19: 34300824-34300825 on Build GRCh38
rs370579561 Chr.19: 34301017-34301017 on Build GRCh38
rs774240213 Chr.19: 34300887-34300887 on Build GRCh38
rs146643118 Chr.19: 34300934-34300934 on Build GRCh38
rs566381372 Chr.19: 34301062-34301062 on Build GRCh38
rs754164774 Chr.19: 34300800-34300800 on Build GRCh38
rs551356719 Chr.19: 34301077-34301077 on Build GRCh38
rs763743064 Chr.19: 34300796-34300796 on Build GRCh38
rs201207182 Chr.19: 34300692-34300692 on Build GRCh38
rs369099995 Chr.19: 34300861-34300861 on Build GRCh38
rs752822969 Chr.19: 34300927-34300927 on Build GRCh38
rs769176897 Chr.19: 34300863-34300863 on Build GRCh38
rs765523205 Chr.19: 34301010-34301010 on Build GRCh38
rs571345352 Chr.19: 34301089-34301089 on Build GRCh38
rs745469726 Chr.19: 34300951-34300951 on Build GRCh38
rs774401718 Chr.19: 34300985-34300985 on Build GRCh38
rs202077029 Chr.19: 34300695-34300695 on Build GRCh38
rs775380576 Chr.19: 34301011-34301011 on Build GRCh38
rs188387566 Chr.19: 34300824-34300824 on Build GRCh38
rs373446167 Chr.19: 34300747-34300747 on Build GRCh38

View Full Product Details