Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36015734-36016231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP3
Entrez Gene ID
25999
Gene Name
CAP-Gly domain containing linker protein 3
Gene Aliases
CLIPR-59, CLIPR59, RSNL1
UniGene
Hs.466539
Gene Chromosome Location
Chr.19: 36014660-36032895 on Build GRCh38
Transcripts
NM_001199570.1 NM_015526.2 NM_001290056.1

Gene Symbol
LOC101927572
Entrez Gene ID
101927572
Gene Name
uncharacterized LOC101927572
Gene Aliases
-
UniGene
Hs.676116
Gene Chromosome Location
Chr.19: 36014499-36045997 on Build GRCh38
Transcripts
NM_001199570.1 NM_015526.2 NM_001290056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630870 c.1625G>C p.R542T Chr.19: 36016177-36016177 on Build GRCh38
1712198 c.1628C>T p.A543V Chr.19: 36016174-36016174 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751326856 Chr.19: 36016173-36016173 on Build GRCh38
rs566256774 Chr.19: 36016164-36016164 on Build GRCh38
rs755707156 Chr.19: 36015883-36015883 on Build GRCh38
rs867674014 Chr.19: 36016083-36016083 on Build GRCh38
rs747943491 Chr.19: 36016153-36016153 on Build GRCh38
rs745476868 Chr.19: 36015785-36015785 on Build GRCh38
rs900493625 Chr.19: 36016032-36016032 on Build GRCh38
rs531704483 Chr.19: 36016020-36016020 on Build GRCh38
rs373129417 Chr.19: 36016121-36016121 on Build GRCh38
rs777021304 Chr.19: 36016123-36016123 on Build GRCh38
rs970017130 Chr.19: 36016208-36016208 on Build GRCh38
rs752957221 Chr.19: 36016149-36016149 on Build GRCh38
rs926993196 Chr.19: 36015864-36015864 on Build GRCh38
rs549954209 Chr.19: 36016100-36016100 on Build GRCh38
rs990776899 Chr.19: 36015875-36015875 on Build GRCh38
rs984288275 Chr.19: 36015780-36015780 on Build GRCh38
rs375085266 Chr.19: 36016103-36016103 on Build GRCh38
rs572372222 Chr.19: 36015935-36015935 on Build GRCh38
rs754473727 Chr.19: 36016034-36016034 on Build GRCh38
rs970355183 Chr.19: 36016206-36016206 on Build GRCh38
rs904423055 Chr.19: 36015991-36015991 on Build GRCh38
rs1048368899 Chr.19: 36015967-36015967 on Build GRCh38
rs191206537 Chr.19: 36015946-36015946 on Build GRCh38
rs574161520 Chr.19: 36015800-36015815 on Build GRCh38
rs377246800 Chr.19: 36016139-36016139 on Build GRCh38
rs980162574 Chr.19: 36015806-36015806 on Build GRCh38
rs942273777 Chr.19: 36016023-36016023 on Build GRCh38
rs1039295175 Chr.19: 36016027-36016027 on Build GRCh38
rs1056906350 Chr.19: 36015893-36015893 on Build GRCh38
rs997491670 Chr.19: 36016089-36016089 on Build GRCh38
rs779478161 Chr.19: 36015891-36015891 on Build GRCh38
rs560272920 Chr.19: 36015916-36015916 on Build GRCh38
rs901182557 Chr.19: 36016076-36016076 on Build GRCh38
rs910122017 Chr.19: 36015791-36015791 on Build GRCh38
rs1023578298 Chr.19: 36016174-36016174 on Build GRCh38
rs186051220 Chr.19: 36015871-36015871 on Build GRCh38
rs916244192 Chr.19: 36015888-36015888 on Build GRCh38
rs200757066 Chr.19: 36016096-36016096 on Build GRCh38
rs913037577 Chr.19: 36015947-36015947 on Build GRCh38
rs981836216 Chr.19: 36015908-36015908 on Build GRCh38
rs1006819349 Chr.19: 36016026-36016026 on Build GRCh38
rs372057691 Chr.19: 36016102-36016102 on Build GRCh38
rs957933433 Chr.19: 36016152-36016152 on Build GRCh38
rs918356113 Chr.19: 36015928-36015928 on Build GRCh38
rs757021638 Chr.19: 36016176-36016176 on Build GRCh38
rs183209495 Chr.19: 36015983-36015983 on Build GRCh38
rs547716033 Chr.19: 36016126-36016126 on Build GRCh38

View Full Product Details