Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36891472-36891973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF345
Entrez Gene ID
25850
Gene Name
zinc finger protein 345
Gene Aliases
HZF10
UniGene
Hs.362324
Gene Chromosome Location
Chr.19: 36850350-36893218 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Gene Symbol
ZNF829
Entrez Gene ID
374899
Gene Name
zinc finger protein 829
Gene Aliases
-
UniGene
Hs.720312
Gene Chromosome Location
Chr.19: 36888124-36916387 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612126 c.977G>C p.C326S Chr.19: 36891814-36891814 on Build GRCh38
4752986 c.966A>C p.K322N Chr.19: 36891825-36891825 on Build GRCh38
4767291 c.1033A>C p.I345L Chr.19: 36891758-36891758 on Build GRCh38
1612127 c.829T>A p.F277I Chr.19: 36891962-36891962 on Build GRCh38
7078875 c.1216A>C p.K406Q Chr.19: 36891575-36891575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748693239 Chr.19: 36891718-36891718 on Build GRCh38
rs200748250 Chr.19: 36891602-36891602 on Build GRCh38
rs759428106 Chr.19: 36891646-36891646 on Build GRCh38
rs757970073 Chr.19: 36891541-36891541 on Build GRCh38
rs771101060 Chr.19: 36891760-36891760 on Build GRCh38
rs370190247 Chr.19: 36891616-36891616 on Build GRCh38
rs756169350 Chr.19: 36891501-36891519 on Build GRCh38
rs751222241 Chr.19: 36891561-36891561 on Build GRCh38
rs572267939 Chr.19: 36891815-36891815 on Build GRCh38
rs370987908 Chr.19: 36891766-36891766 on Build GRCh38
rs780815980 Chr.19: 36891581-36891581 on Build GRCh38
rs377686659 Chr.19: 36891653-36891653 on Build GRCh38
rs756758300 Chr.19: 36891667-36891667 on Build GRCh38
rs778461996 Chr.19: 36891916-36891916 on Build GRCh38
rs561580495 Chr.19: 36891903-36891903 on Build GRCh38
rs764914093 Chr.19: 36891936-36891936 on Build GRCh38
rs538972636 Chr.19: 36891777-36891777 on Build GRCh38
rs200398691 Chr.19: 36891817-36891817 on Build GRCh38
rs756736847 Chr.19: 36891518-36891518 on Build GRCh38
rs751130418 Chr.19: 36891655-36891655 on Build GRCh38
rs900526065 Chr.19: 36891865-36891865 on Build GRCh38
rs757376817 Chr.19: 36891942-36891942 on Build GRCh38
rs752558777 Chr.19: 36891539-36891539 on Build GRCh38
rs902455550 Chr.19: 36891885-36891885 on Build GRCh38
rs779514046 Chr.19: 36891598-36891598 on Build GRCh38
rs749717750 Chr.19: 36891584-36891584 on Build GRCh38
rs753169453 Chr.19: 36891888-36891888 on Build GRCh38
rs1024427937 Chr.19: 36891737-36891737 on Build GRCh38
rs954107135 Chr.19: 36891890-36891890 on Build GRCh38
rs79277718 Chr.19: 36891516-36891516 on Build GRCh38
rs767854488 Chr.19: 36891621-36891622 on Build GRCh38
rs949068706 Chr.19: 36891725-36891725 on Build GRCh38
rs554063383 Chr.19: 36891789-36891789 on Build GRCh38
rs556562482 Chr.19: 36891533-36891533 on Build GRCh38
rs760178925 Chr.19: 36891846-36891846 on Build GRCh38
rs1046003831 Chr.19: 36891728-36891728 on Build GRCh38
rs199766242 Chr.19: 36891806-36891806 on Build GRCh38
rs778409148 Chr.19: 36891729-36891729 on Build GRCh38
rs774705661 Chr.19: 36891514-36891514 on Build GRCh38
rs750930987 Chr.19: 36891503-36891503 on Build GRCh38
rs758917124 Chr.19: 36891719-36891719 on Build GRCh38
rs773600149 Chr.19: 36891517-36891517 on Build GRCh38
rs760930868 Chr.19: 36891520-36891520 on Build GRCh38
rs750851502 Chr.19: 36891636-36891644 on Build GRCh38
rs756833731 Chr.19: 36891580-36891580 on Build GRCh38
rs542821566 Chr.19: 36891839-36891839 on Build GRCh38
rs776706408 Chr.19: 36891638-36891638 on Build GRCh38
rs755393020 Chr.19: 36891698-36891698 on Build GRCh38
rs766291147 Chr.19: 36891529-36891529 on Build GRCh38
rs372663992 Chr.19: 36891601-36891601 on Build GRCh38
rs868863246 Chr.19: 36891642-36891642 on Build GRCh38
rs750022943 Chr.19: 36891696-36891696 on Build GRCh38
rs752088177 Chr.19: 36891934-36891934 on Build GRCh38
rs746024713 Chr.19: 36891771-36891771 on Build GRCh38
rs369348025 Chr.19: 36891592-36891592 on Build GRCh38
rs1048738550 Chr.19: 36891691-36891691 on Build GRCh38
rs764913013 Chr.19: 36891648-36891648 on Build GRCh38
rs767241898 Chr.19: 36891688-36891688 on Build GRCh38
rs374018238 Chr.19: 36891798-36891798 on Build GRCh38
rs966547273 Chr.19: 36891578-36891578 on Build GRCh38
rs997915354 Chr.19: 36891868-36891868 on Build GRCh38
rs771348288 Chr.19: 36891631-36891631 on Build GRCh38
rs866477046 Chr.19: 36891664-36891664 on Build GRCh38
rs990493512 Chr.19: 36891630-36891630 on Build GRCh38
rs748307111 Chr.19: 36891515-36891515 on Build GRCh38
rs999357901 Chr.19: 36891907-36891907 on Build GRCh38
rs765647992 Chr.19: 36891872-36891872 on Build GRCh38
rs747151341 Chr.19: 36891755-36891755 on Build GRCh38
rs779107045 Chr.19: 36891699-36891699 on Build GRCh38
rs776421835 Chr.19: 36891532-36891532 on Build GRCh38
rs763558897 Chr.19: 36891557-36891557 on Build GRCh38
rs374436128 Chr.19: 36891651-36891651 on Build GRCh38
rs578174327 Chr.19: 36891654-36891654 on Build GRCh38
rs769658336 Chr.19: 36891776-36891776 on Build GRCh38
rs376922609 Chr.19: 36891603-36891603 on Build GRCh38
rs759155383 Chr.19: 36891730-36891730 on Build GRCh38
rs762238694 Chr.19: 36891561-36891562 on Build GRCh38

View Full Product Details