Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37152945-37153455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF585A
Entrez Gene ID
199704
Gene Name
zinc finger protein 585A
Gene Aliases
-
UniGene
Hs.659236
Gene Chromosome Location
Chr.19: 37147438-37172741 on Build GRCh38
Transcripts
NM_001288800.1 NM_152655.3 NM_199126.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524932 c.697C>T p.H233Y Chr.19: 37153037-37153037 on Build GRCh38
3533101 c.557T>A p.F186Y Chr.19: 37153342-37153342 on Build GRCh38
4077524 c.291T>C p.L97L Chr.19: 37153443-37153443 on Build GRCh38
1712211 c.589C>T p.Q197* Chr.19: 37153310-37153310 on Build GRCh38
188369 c.701G>A p.R234Q Chr.19: 37153033-37153033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998998404 Chr.19: 37153093-37153093 on Build GRCh38
rs143906060 Chr.19: 37153133-37153133 on Build GRCh38
rs778025597 Chr.19: 37153073-37153073 on Build GRCh38
rs780553869 Chr.19: 37153305-37153305 on Build GRCh38
rs375962026 Chr.19: 37153021-37153021 on Build GRCh38
rs748424020 Chr.19: 37153332-37153332 on Build GRCh38
rs148585024 Chr.19: 37153270-37153270 on Build GRCh38
rs149338317 Chr.19: 37153211-37153211 on Build GRCh38
rs755681123 Chr.19: 37153067-37153067 on Build GRCh38
rs748959106 Chr.19: 37152997-37152997 on Build GRCh38
rs781041269 Chr.19: 37153054-37153054 on Build GRCh38
rs763843729 Chr.19: 37153044-37153044 on Build GRCh38
rs141892107 Chr.19: 37153122-37153122 on Build GRCh38
rs766304050 Chr.19: 37153257-37153257 on Build GRCh38
rs867868491 Chr.19: 37153198-37153198 on Build GRCh38
rs757172889 Chr.19: 37153052-37153052 on Build GRCh38
rs182259444 Chr.19: 37153302-37153302 on Build GRCh38
rs771520159 Chr.19: 37153023-37153023 on Build GRCh38
rs750839137 Chr.19: 37153409-37153409 on Build GRCh38
rs369819757 Chr.19: 37153140-37153140 on Build GRCh38
rs986842065 Chr.19: 37153225-37153225 on Build GRCh38
rs139703545 Chr.19: 37153191-37153191 on Build GRCh38
rs377263650 Chr.19: 37153088-37153088 on Build GRCh38
rs748937252 Chr.19: 37153071-37153071 on Build GRCh38
rs750429570 Chr.19: 37153131-37153131 on Build GRCh38
rs776673917 Chr.19: 37153344-37153344 on Build GRCh38
rs370889139 Chr.19: 37153229-37153229 on Build GRCh38
rs768336087 Chr.19: 37153072-37153072 on Build GRCh38
rs746194945 Chr.19: 37153282-37153282 on Build GRCh38
rs144430542 Chr.19: 37153271-37153271 on Build GRCh38
rs763202419 Chr.19: 37153042-37153042 on Build GRCh38
rs774287804 Chr.19: 37153316-37153316 on Build GRCh38
rs770613625 Chr.19: 37153340-37153340 on Build GRCh38
rs760633008 Chr.19: 37153254-37153254 on Build GRCh38
rs768774194 Chr.19: 37153218-37153218 on Build GRCh38
rs371659218 Chr.19: 37152995-37152995 on Build GRCh38
rs368107459 Chr.19: 37152969-37152969 on Build GRCh38
rs754572050 Chr.19: 37153260-37153260 on Build GRCh38
rs560374823 Chr.19: 37153103-37153103 on Build GRCh38
rs757760523 Chr.19: 37153188-37153188 on Build GRCh38
rs747739310 Chr.19: 37153187-37153187 on Build GRCh38
rs756549472 Chr.19: 37153412-37153412 on Build GRCh38
rs779625830 Chr.19: 37152990-37152990 on Build GRCh38
rs779537700 Chr.19: 37153068-37153068 on Build GRCh38
rs764336682 Chr.19: 37153101-37153101 on Build GRCh38
rs57916401 Chr.19: 37153149-37153149 on Build GRCh38
rs758391518 Chr.19: 37153406-37153406 on Build GRCh38
rs775617175 Chr.19: 37153089-37153089 on Build GRCh38
rs1011666825 Chr.19: 37153145-37153145 on Build GRCh38
rs571853976 Chr.19: 37153337-37153337 on Build GRCh38
rs780248991 Chr.19: 37153417-37153417 on Build GRCh38
rs770375388 Chr.19: 37153204-37153204 on Build GRCh38
rs368991030 Chr.19: 37153033-37153033 on Build GRCh38
rs773599256 Chr.19: 37153013-37153013 on Build GRCh38
rs760418433 Chr.19: 37153339-37153339 on Build GRCh38
rs769443221 Chr.19: 37152982-37152982 on Build GRCh38
rs779060372 Chr.19: 37153425-37153425 on Build GRCh38
rs369952455 Chr.19: 37153272-37153272 on Build GRCh38
rs372852597 Chr.19: 37153251-37153251 on Build GRCh38
rs754433642 Chr.19: 37153420-37153420 on Build GRCh38
rs375276322 Chr.19: 37153333-37153333 on Build GRCh38
rs755424594 Chr.19: 37153422-37153422 on Build GRCh38
rs186098323 Chr.19: 37153397-37153397 on Build GRCh38
rs868468724 Chr.19: 37153362-37153362 on Build GRCh38
rs74605992 Chr.19: 37153311-37153311 on Build GRCh38
rs764645813 Chr.19: 37153263-37153263 on Build GRCh38
rs746425707 Chr.19: 37153197-37153197 on Build GRCh38
rs767343183 Chr.19: 37153246-37153246 on Build GRCh38
rs201793121 Chr.19: 37153189-37153189 on Build GRCh38
rs753032748 Chr.19: 37153035-37153035 on Build GRCh38
rs774021931 Chr.19: 37153102-37153102 on Build GRCh38
rs756017006 Chr.19: 37153132-37153132 on Build GRCh38
rs762700640 Chr.19: 37153403-37153403 on Build GRCh38
rs749735548 Chr.19: 37153307-37153307 on Build GRCh38
rs868588800 Chr.19: 37153249-37153249 on Build GRCh38
rs200310362 Chr.19: 37153405-37153405 on Build GRCh38
rs776852199 Chr.19: 37153027-37153027 on Build GRCh38
rs772387782 Chr.19: 37153002-37153002 on Build GRCh38
rs772972019 Chr.19: 37153248-37153248 on Build GRCh38
rs752198586 Chr.19: 37153400-37153400 on Build GRCh38
rs771575283 Chr.19: 37153081-37153081 on Build GRCh38
rs367893624 Chr.19: 37153426-37153426 on Build GRCh38
rs763106021 Chr.19: 37153100-37153100 on Build GRCh38
rs749950759 Chr.19: 37153066-37153066 on Build GRCh38
rs759389942 Chr.19: 37153384-37153384 on Build GRCh38
rs752432653 Chr.19: 37153266-37153266 on Build GRCh38
rs373038145 Chr.19: 37153074-37153074 on Build GRCh38
rs749323324 Chr.19: 37153217-37153217 on Build GRCh38
rs751318956 Chr.19: 37153046-37153046 on Build GRCh38
rs765392534 Chr.19: 37153034-37153034 on Build GRCh38
rs144861208 Chr.19: 37153258-37153258 on Build GRCh38
rs376706809 Chr.19: 37153168-37153168 on Build GRCh38
rs760114914 Chr.19: 37153082-37153082 on Build GRCh38

View Full Product Details