Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37885468-37885977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR87
Entrez Gene ID
83889
Gene Name
WD repeat domain 87
Gene Aliases
NYD-SP11
UniGene
Hs.664194
Gene Chromosome Location
Chr.19: 37884577-37906706 on Build GRCh38
Transcripts
NM_001291088.1 NM_031951.4 XM_011527360.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
995840 c.7984G>A p.E2662K Chr.19: 37885570-37885570 on Build GRCh38
5921617 c.7625C>T p.S2542F Chr.19: 37885929-37885929 on Build GRCh38
5346532 c.7594C>T p.L2532L Chr.19: 37885960-37885960 on Build GRCh38
4282377 c.8044G>A p.E2682K Chr.19: 37885510-37885510 on Build GRCh38
5056596 c.7741C>A p.L2581M Chr.19: 37885813-37885813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558935370 Chr.19: 37885836-37885836 on Build GRCh38
rs10408510 Chr.19: 37885845-37885845 on Build GRCh38
rs932347615 Chr.19: 37885699-37885699 on Build GRCh38
rs149523077 Chr.19: 37885585-37885585 on Build GRCh38
rs896964318 Chr.19: 37885554-37885554 on Build GRCh38
rs978736320 Chr.19: 37885634-37885634 on Build GRCh38
rs557031994 Chr.19: 37885907-37885907 on Build GRCh38
rs968631762 Chr.19: 37885618-37885618 on Build GRCh38
rs759967163 Chr.19: 37885595-37885595 on Build GRCh38
rs751228063 Chr.19: 37885539-37885540 on Build GRCh38
rs762069466 Chr.19: 37885898-37885898 on Build GRCh38
rs1006382077 Chr.19: 37885810-37885810 on Build GRCh38
rs758479311 Chr.19: 37885785-37885785 on Build GRCh38
rs201341562 Chr.19: 37885690-37885690 on Build GRCh38
rs140620408 Chr.19: 37885944-37885944 on Build GRCh38
rs751749158 Chr.19: 37885692-37885692 on Build GRCh38
rs867447893 Chr.19: 37885939-37885939 on Build GRCh38
rs142857310 Chr.19: 37885954-37885954 on Build GRCh38
rs909527447 Chr.19: 37885794-37885794 on Build GRCh38
rs960710861 Chr.19: 37885855-37885855 on Build GRCh38
rs757145397 Chr.19: 37885680-37885680 on Build GRCh38
rs552274186 Chr.19: 37885913-37885913 on Build GRCh38
rs376695008 Chr.19: 37885514-37885514 on Build GRCh38
rs943633751 Chr.19: 37885670-37885670 on Build GRCh38
rs960678220 Chr.19: 37885574-37885574 on Build GRCh38
rs771246610 Chr.19: 37885576-37885576 on Build GRCh38
rs770257593 Chr.19: 37885840-37885840 on Build GRCh38
rs143991202 Chr.19: 37885649-37885649 on Build GRCh38
rs529997341 Chr.19: 37885827-37885827 on Build GRCh38
rs778463046 Chr.19: 37885804-37885804 on Build GRCh38
rs544012769 Chr.19: 37885597-37885597 on Build GRCh38
rs200024701 Chr.19: 37885621-37885621 on Build GRCh38
rs756748251 Chr.19: 37885559-37885559 on Build GRCh38
rs762960595 Chr.19: 37885867-37885867 on Build GRCh38
rs370935622 Chr.19: 37885631-37885631 on Build GRCh38
rs1047946861 Chr.19: 37885778-37885778 on Build GRCh38
rs988206133 Chr.19: 37885578-37885578 on Build GRCh38
rs866428498 Chr.19: 37885683-37885683 on Build GRCh38
rs764197978 Chr.19: 37885639-37885639 on Build GRCh38
rs532766383 Chr.19: 37885640-37885640 on Build GRCh38
rs190158380 Chr.19: 37885694-37885694 on Build GRCh38
rs756107216 Chr.19: 37885738-37885738 on Build GRCh38
rs1035013050 Chr.19: 37885568-37885568 on Build GRCh38
rs552860128 Chr.19: 37885773-37885773 on Build GRCh38
rs1003341073 Chr.19: 37885842-37885842 on Build GRCh38
rs867506203 Chr.19: 37885495-37885495 on Build GRCh38
rs201555125 Chr.19: 37885943-37885943 on Build GRCh38
rs764366822 Chr.19: 37885668-37885668 on Build GRCh38
rs751825059 Chr.19: 37885832-37885832 on Build GRCh38
rs577755444 Chr.19: 37885829-37885829 on Build GRCh38
rs754464380 Chr.19: 37885797-37885797 on Build GRCh38
rs115178885 Chr.19: 37885828-37885828 on Build GRCh38
rs1015459360 Chr.19: 37885549-37885549 on Build GRCh38
rs750399394 Chr.19: 37885731-37885731 on Build GRCh38
rs990783961 Chr.19: 37885617-37885617 on Build GRCh38
rs748714625 Chr.19: 37885781-37885781 on Build GRCh38
rs371409897 Chr.19: 37885821-37885821 on Build GRCh38
rs765362570 Chr.19: 37885607-37885607 on Build GRCh38
rs1049439881 Chr.19: 37885706-37885706 on Build GRCh38
rs1023310809 Chr.19: 37885494-37885494 on Build GRCh38
rs752669541 Chr.19: 37885627-37885627 on Build GRCh38
rs779931233 Chr.19: 37885869-37885869 on Build GRCh38
rs774437076 Chr.19: 37885896-37885896 on Build GRCh38

View Full Product Details