Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38327286-38327775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK6
Entrez Gene ID
9424
Gene Name
potassium two pore domain channel subfamily K member 6
Gene Aliases
K2p6.1, KCNK8, TOSS, TWIK-2, TWIK2
UniGene
Hs.240395
Gene Chromosome Location
Chr.19: 38319814-38329014 on Build GRCh38
Transcripts
NM_004823.1 XM_011527526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5389630 c.867C>T p.I289I Chr.19: 38327328-38327328 on Build GRCh38
6427018 c.891C>A p.H297Q Chr.19: 38327352-38327352 on Build GRCh38
3202034 c.924C>T p.Y308Y Chr.19: 38327385-38327385 on Build GRCh38
7302762 c.838A>G p.N280D Chr.19: 38327299-38327299 on Build GRCh38
6244841 c.872G>T p.G291V Chr.19: 38327333-38327333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375236784 Chr.19: 38327313-38327313 on Build GRCh38
rs924080776 Chr.19: 38327751-38327751 on Build GRCh38
rs927740655 Chr.19: 38327682-38327682 on Build GRCh38
rs140227388 Chr.19: 38327318-38327318 on Build GRCh38
rs781437400 Chr.19: 38327399-38327399 on Build GRCh38
rs952623675 Chr.19: 38327615-38327615 on Build GRCh38
rs573672675 Chr.19: 38327487-38327487 on Build GRCh38
rs774513893 Chr.19: 38327446-38327446 on Build GRCh38
rs867848648 Chr.19: 38327404-38327404 on Build GRCh38
rs776991765 Chr.19: 38327335-38327335 on Build GRCh38
rs990480709 Chr.19: 38327707-38327707 on Build GRCh38
rs201279966 Chr.19: 38327386-38327386 on Build GRCh38
rs374630252 Chr.19: 38327349-38327349 on Build GRCh38
rs749443797 Chr.19: 38327428-38327428 on Build GRCh38
rs371433357 Chr.19: 38327378-38327378 on Build GRCh38
rs760768471 Chr.19: 38327594-38327594 on Build GRCh38
rs781253725 Chr.19: 38327306-38327306 on Build GRCh38
rs940672455 Chr.19: 38327481-38327481 on Build GRCh38
rs900468075 Chr.19: 38327582-38327582 on Build GRCh38
rs920863789 Chr.19: 38327750-38327753 on Build GRCh38
rs1019979336 Chr.19: 38327568-38327568 on Build GRCh38
rs568626349 Chr.19: 38327736-38327736 on Build GRCh38
rs981909890 Chr.19: 38327410-38327410 on Build GRCh38
rs199734308 Chr.19: 38327385-38327385 on Build GRCh38
rs1034099967 Chr.19: 38327343-38327343 on Build GRCh38
rs549369957 Chr.19: 38327348-38327348 on Build GRCh38
rs780036066 Chr.19: 38327331-38327331 on Build GRCh38
rs778517642 Chr.19: 38327320-38327320 on Build GRCh38
rs746207668 Chr.19: 38327406-38327406 on Build GRCh38
rs546003759 Chr.19: 38327352-38327352 on Build GRCh38
rs746380227 Chr.19: 38327337-38327337 on Build GRCh38
rs907898067 Chr.19: 38327461-38327461 on Build GRCh38
rs143217284 Chr.19: 38327593-38327593 on Build GRCh38
rs773964608 Chr.19: 38327436-38327437 on Build GRCh38
rs1055349360 Chr.19: 38327467-38327467 on Build GRCh38
rs767646161 Chr.19: 38327557-38327557 on Build GRCh38
rs371025331 Chr.19: 38327437-38327437 on Build GRCh38
rs762059409 Chr.19: 38327351-38327351 on Build GRCh38
rs769244044 Chr.19: 38327328-38327328 on Build GRCh38
rs769008976 Chr.19: 38327434-38327434 on Build GRCh38
rs749223512 Chr.19: 38327346-38327348 on Build GRCh38
rs776123206 Chr.19: 38327427-38327427 on Build GRCh38
rs982241444 Chr.19: 38327672-38327672 on Build GRCh38
rs145593084 Chr.19: 38327341-38327341 on Build GRCh38
rs756021437 Chr.19: 38327314-38327314 on Build GRCh38
rs754474011 Chr.19: 38327380-38327380 on Build GRCh38
rs975257853 Chr.19: 38327623-38327623 on Build GRCh38
rs902708690 Chr.19: 38327521-38327521 on Build GRCh38
rs151147940 Chr.19: 38327317-38327317 on Build GRCh38
rs996136592 Chr.19: 38327592-38327592 on Build GRCh38
rs761967593 Chr.19: 38327518-38327518 on Build GRCh38
rs1037990665 Chr.19: 38327484-38327484 on Build GRCh38
rs1013984474 Chr.19: 38327527-38327527 on Build GRCh38
rs768472826 Chr.19: 38327359-38327360 on Build GRCh38
rs955171045 Chr.19: 38327663-38327663 on Build GRCh38
rs3745952 Chr.19: 38327659-38327659 on Build GRCh38
rs186101876 Chr.19: 38327395-38327395 on Build GRCh38
rs753396263 Chr.19: 38327379-38327379 on Build GRCh38
rs758430268 Chr.19: 38327739-38327739 on Build GRCh38
rs371030201 Chr.19: 38327334-38327334 on Build GRCh38
rs1015826426 Chr.19: 38327651-38327651 on Build GRCh38
rs973771478 Chr.19: 38327741-38327741 on Build GRCh38
rs747432115 Chr.19: 38327330-38327330 on Build GRCh38
rs73041296 Chr.19: 38327304-38327304 on Build GRCh38
rs1009379138 Chr.19: 38327562-38327562 on Build GRCh38
rs1042941932 Chr.19: 38327520-38327520 on Build GRCh38
rs779446626 Chr.19: 38327490-38327490 on Build GRCh38
rs561771204 Chr.19: 38327579-38327579 on Build GRCh38
rs578120117 Chr.19: 38327342-38327342 on Build GRCh38
rs367679159 Chr.19: 38327355-38327355 on Build GRCh38
rs774706372 Chr.19: 38327350-38327350 on Build GRCh38
rs958985214 Chr.19: 38327687-38327687 on Build GRCh38
rs148325841 Chr.19: 38327737-38327737 on Build GRCh38
rs559702897 Chr.19: 38327632-38327632 on Build GRCh38
rs944901917 Chr.19: 38327752-38327752 on Build GRCh38
rs763561578 Chr.19: 38327376-38327376 on Build GRCh38
rs770249632 Chr.19: 38327409-38327409 on Build GRCh38
rs765130387 Chr.19: 38327625-38327625 on Build GRCh38
rs1005455142 Chr.19: 38327634-38327634 on Build GRCh38
rs775672766 Chr.19: 38327356-38327356 on Build GRCh38
rs914943766 Chr.19: 38327724-38327724 on Build GRCh38

View Full Product Details