Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38918015-38918512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARS2
Entrez Gene ID
54938
Gene Name
seryl-tRNA synthetase 2, mitochondrial
Gene Aliases
SARS, SARSM, SERS, SYS, SerRS, SerRSmt, mtSerRS
UniGene
Hs.709416
Gene Chromosome Location
Chr.19: 38915264-38930896 on Build GRCh38
Transcripts
NM_001145901.1 NM_017827.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6771945 c.916G>A p.G306S Chr.19: 38918422-38918422 on Build GRCh38
4769459 c.922+3G>A p.? Chr.19: 38918419-38918419 on Build GRCh38
2157410 c.950A>C p.D317A Chr.19: 38918106-38918106 on Build GRCh38
5517512 c.899G>A p.G300E Chr.19: 38918445-38918445 on Build GRCh38
6771944 c.922G>A p.G308S Chr.19: 38918422-38918422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017302120 Chr.19: 38918411-38918411 on Build GRCh38
rs60352446 Chr.19: 38918411-38918412 on Build GRCh38
rs954446202 Chr.19: 38918077-38918077 on Build GRCh38
rs542117712 Chr.19: 38918143-38918143 on Build GRCh38
rs192230421 Chr.19: 38918435-38918435 on Build GRCh38
rs182484523 Chr.19: 38918450-38918450 on Build GRCh38
rs757034837 Chr.19: 38918467-38918467 on Build GRCh38
rs539232990 Chr.19: 38918369-38918369 on Build GRCh38
rs369982598 Chr.19: 38918047-38918047 on Build GRCh38
rs926114893 Chr.19: 38918197-38918197 on Build GRCh38
rs569078516 Chr.19: 38918351-38918351 on Build GRCh38
rs763514016 Chr.19: 38918043-38918043 on Build GRCh38
rs139600976 Chr.19: 38918119-38918119 on Build GRCh38
rs942882713 Chr.19: 38918216-38918216 on Build GRCh38
rs778973299 Chr.19: 38918089-38918089 on Build GRCh38
rs772816444 Chr.19: 38918448-38918448 on Build GRCh38
rs745358885 Chr.19: 38918107-38918107 on Build GRCh38
rs549369011 Chr.19: 38918246-38918246 on Build GRCh38
rs778297773 Chr.19: 38918484-38918484 on Build GRCh38
rs752816058 Chr.19: 38918440-38918440 on Build GRCh38
rs754023830 Chr.19: 38918184-38918184 on Build GRCh38
rs758199702 Chr.19: 38918096-38918096 on Build GRCh38
rs140851346 Chr.19: 38918475-38918475 on Build GRCh38
rs4803034 Chr.19: 38918048-38918048 on Build GRCh38
rs772633018 Chr.19: 38918416-38918416 on Build GRCh38
rs1047045695 Chr.19: 38918247-38918247 on Build GRCh38
rs777495435 Chr.19: 38918384-38918384 on Build GRCh38
rs201472832 Chr.19: 38918066-38918066 on Build GRCh38
rs1001935298 Chr.19: 38918085-38918085 on Build GRCh38
rs747367586 Chr.19: 38918485-38918485 on Build GRCh38
rs746697410 Chr.19: 38918101-38918101 on Build GRCh38
rs766596120 Chr.19: 38918175-38918175 on Build GRCh38
rs377236830 Chr.19: 38918415-38918415 on Build GRCh38
rs1029967366 Chr.19: 38918362-38918362 on Build GRCh38
rs773492726 Chr.19: 38918420-38918420 on Build GRCh38
rs756907209 Chr.19: 38918396-38918396 on Build GRCh38
rs769171598 Chr.19: 38918115-38918115 on Build GRCh38
rs374265064 Chr.19: 38918324-38918324 on Build GRCh38
rs762119532 Chr.19: 38918052-38918052 on Build GRCh38
rs918019366 Chr.19: 38918248-38918248 on Build GRCh38
rs765267941 Chr.19: 38918374-38918374 on Build GRCh38
rs941256664 Chr.19: 38918328-38918328 on Build GRCh38
rs777493834 Chr.19: 38918099-38918099 on Build GRCh38
rs149720096 Chr.19: 38918434-38918434 on Build GRCh38
rs910348716 Chr.19: 38918142-38918142 on Build GRCh38
rs767814478 Chr.19: 38918056-38918056 on Build GRCh38
rs762170958 Chr.19: 38918120-38918120 on Build GRCh38
rs949424844 Chr.19: 38918295-38918295 on Build GRCh38
rs760877000 Chr.19: 38918165-38918165 on Build GRCh38
rs892698905 Chr.19: 38918346-38918346 on Build GRCh38
rs933976558 Chr.19: 38918269-38918269 on Build GRCh38
rs774853873 Chr.19: 38918407-38918407 on Build GRCh38
rs780828600 Chr.19: 38918469-38918469 on Build GRCh38
rs761507700 Chr.19: 38918391-38918391 on Build GRCh38
rs752619987 Chr.19: 38918382-38918382 on Build GRCh38
rs761081540 Chr.19: 38918429-38918429 on Build GRCh38
rs936814207 Chr.19: 38918326-38918326 on Build GRCh38
rs919179409 Chr.19: 38918206-38918206 on Build GRCh38
rs776187528 Chr.19: 38918041-38918041 on Build GRCh38
rs550590004 Chr.19: 38918342-38918342 on Build GRCh38
rs753964044 Chr.19: 38918074-38918074 on Build GRCh38
rs1038709279 Chr.19: 38918345-38918345 on Build GRCh38
rs755006378 Chr.19: 38918086-38918086 on Build GRCh38
rs917884456 Chr.19: 38918461-38918461 on Build GRCh38
rs145232405 Chr.19: 38918296-38918296 on Build GRCh38
rs144697966 Chr.19: 38918480-38918480 on Build GRCh38
rs758339707 Chr.19: 38918383-38918383 on Build GRCh38
rs1033683967 Chr.19: 38918476-38918476 on Build GRCh38
rs868083911 Chr.19: 38918407-38918408 on Build GRCh38
rs79702490 Chr.19: 38918470-38918470 on Build GRCh38
rs1009673826 Chr.19: 38918358-38918358 on Build GRCh38
rs745416138 Chr.19: 38918400-38918400 on Build GRCh38
rs527701185 Chr.19: 38918186-38918186 on Build GRCh38
rs200998412 Chr.19: 38918070-38918070 on Build GRCh38
rs144529043 Chr.19: 38918438-38918438 on Build GRCh38
rs973243823 Chr.19: 38918422-38918422 on Build GRCh38
rs746788057 Chr.19: 38918392-38918392 on Build GRCh38
rs150429988 Chr.19: 38918067-38918067 on Build GRCh38
rs760235346 Chr.19: 38918038-38918038 on Build GRCh38
rs770288697 Chr.19: 38918039-38918039 on Build GRCh38
rs1007231424 Chr.19: 38918403-38918403 on Build GRCh38
rs957005475 Chr.19: 38918122-38918122 on Build GRCh38
rs574003951 Chr.19: 38918486-38918486 on Build GRCh38
rs769228801 Chr.19: 38918406-38918406 on Build GRCh38
rs776041054 Chr.19: 38918106-38918106 on Build GRCh38
rs780846488 Chr.19: 38918399-38918399 on Build GRCh38
rs377287317 Chr.19: 38918477-38918477 on Build GRCh38
rs773613553 Chr.19: 38918162-38918162 on Build GRCh38
rs1020494109 Chr.19: 38918163-38918163 on Build GRCh38
rs770608203 Chr.19: 38918104-38918104 on Build GRCh38
rs765461892 Chr.19: 38918439-38918439 on Build GRCh38

View Full Product Details