Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46545902-46546397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNCN
Entrez Gene ID
148930
Gene Name
kinocilin
Gene Aliases
Kino, L5
UniGene
Hs.350764
Gene Chromosome Location
Chr.1: 46545638-46551657 on Build GRCh38
Transcripts
NM_001097611.1 NM_001322255.1 NR_038403.1

Gene Symbol
MKNK1-AS1
Entrez Gene ID
100507423
Gene Name
MKNK1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.601863
Gene Chromosome Location
Chr.1: 46538696-46570255 on Build GRCh38
Transcripts
NM_001097611.1 NM_001322255.1 NR_038403.1

SNP Detail

SNP ID Chromosome Location
rs1004706600 Chr.1: 46546052-46546052 on Build GRCh38
rs963272479 Chr.1: 46546314-46546314 on Build GRCh38
rs992316436 Chr.1: 46546143-46546143 on Build GRCh38
rs150442078 Chr.1: 46546078-46546078 on Build GRCh38
rs928708958 Chr.1: 46546038-46546038 on Build GRCh38
rs149201351 Chr.1: 46546309-46546309 on Build GRCh38
rs557937107 Chr.1: 46546189-46546189 on Build GRCh38
rs969961356 Chr.1: 46546292-46546292 on Build GRCh38
rs1007321793 Chr.1: 46546247-46546247 on Build GRCh38
rs1258036 Chr.1: 46546015-46546015 on Build GRCh38
rs565514633 Chr.1: 46546156-46546156 on Build GRCh38
rs189468736 Chr.1: 46546026-46546026 on Build GRCh38
rs886392141 Chr.1: 46546041-46546041 on Build GRCh38
rs930289452 Chr.1: 46546163-46546163 on Build GRCh38
rs924181106 Chr.1: 46546116-46546116 on Build GRCh38
rs956973910 Chr.1: 46546124-46546124 on Build GRCh38
rs949858824 Chr.1: 46545991-46545991 on Build GRCh38
rs758447973 Chr.1: 46545929-46545929 on Build GRCh38
rs763388147 Chr.1: 46546030-46546030 on Build GRCh38
rs967380198 Chr.1: 46546177-46546177 on Build GRCh38
rs888746246 Chr.1: 46546246-46546246 on Build GRCh38
rs374149107 Chr.1: 46546153-46546153 on Build GRCh38
rs893327476 Chr.1: 46546264-46546264 on Build GRCh38
rs981398389 Chr.1: 46546180-46546180 on Build GRCh38
rs949795584 Chr.1: 46546147-46546147 on Build GRCh38
rs114617541 Chr.1: 46546257-46546257 on Build GRCh38
rs935493753 Chr.1: 46545985-46545985 on Build GRCh38
rs766674629 Chr.1: 46546234-46546234 on Build GRCh38
rs1044542833 Chr.1: 46546150-46546150 on Build GRCh38
rs990434223 Chr.1: 46546149-46546149 on Build GRCh38
rs1012145620 Chr.1: 46546287-46546287 on Build GRCh38
rs886331642 Chr.1: 46546182-46546182 on Build GRCh38
rs569952025 Chr.1: 46546188-46546188 on Build GRCh38
rs878922924 Chr.1: 46545993-46545993 on Build GRCh38
rs145328823 Chr.1: 46546146-46546146 on Build GRCh38
rs1056585407 Chr.1: 46545986-46545986 on Build GRCh38
rs773938327 Chr.1: 46546179-46546179 on Build GRCh38
rs535942707 Chr.1: 46546351-46546351 on Build GRCh38
rs555055943 Chr.1: 46546203-46546203 on Build GRCh38
rs1027573128 Chr.1: 46546031-46546031 on Build GRCh38
rs990773846 Chr.1: 46546001-46546001 on Build GRCh38
rs781167607 Chr.1: 46545962-46545962 on Build GRCh38
rs1020474689 Chr.1: 46546172-46546172 on Build GRCh38
rs930405457 Chr.1: 46546315-46546315 on Build GRCh38
rs885125 Chr.1: 46546090-46546090 on Build GRCh38
rs958930128 Chr.1: 46546294-46546294 on Build GRCh38
rs180694444 Chr.1: 46546087-46546087 on Build GRCh38
rs967599879 Chr.1: 46546291-46546291 on Build GRCh38
rs895549086 Chr.1: 46545990-46545990 on Build GRCh38
rs553731981 Chr.1: 46546290-46546290 on Build GRCh38
rs754190252 Chr.1: 46546261-46546261 on Build GRCh38
rs1000802485 Chr.1: 46546212-46546212 on Build GRCh38
rs1019211570 Chr.1: 46546054-46546054 on Build GRCh38
rs756280539 Chr.1: 46546284-46546284 on Build GRCh38
rs755403507 Chr.1: 46546283-46546283 on Build GRCh38
rs574121627 Chr.1: 46546220-46546220 on Build GRCh38
rs924234850 Chr.1: 46545954-46545954 on Build GRCh38
rs1048728141 Chr.1: 46546340-46546340 on Build GRCh38
rs766962606 Chr.1: 46546190-46546190 on Build GRCh38
rs879410254 Chr.1: 46546191-46546191 on Build GRCh38
rs1018976920 Chr.1: 46545927-46545929 on Build GRCh38
rs1048790414 Chr.1: 46546173-46546173 on Build GRCh38
rs1018447293 Chr.1: 46546145-46546145 on Build GRCh38
rs545601780 Chr.1: 46546352-46546352 on Build GRCh38
rs765468319 Chr.1: 46546308-46546308 on Build GRCh38
rs565834402 Chr.1: 46546044-46546044 on Build GRCh38
rs570143981 Chr.1: 46546195-46546195 on Build GRCh38
rs979435625 Chr.1: 46546112-46546112 on Build GRCh38
rs885126 Chr.1: 46546200-46546200 on Build GRCh38
rs887565359 Chr.1: 46546062-46546063 on Build GRCh38
rs971883917 Chr.1: 46546319-46546319 on Build GRCh38
rs571375656 Chr.1: 46546083-46546083 on Build GRCh38
rs1055627096 Chr.1: 46546058-46546058 on Build GRCh38
rs867621783 Chr.1: 46546117-46546117 on Build GRCh38
rs1051647507 Chr.1: 46546365-46546365 on Build GRCh38
rs1030116704 Chr.1: 46546098-46546098 on Build GRCh38
rs529149447 Chr.1: 46546091-46546091 on Build GRCh38
rs761178790 Chr.1: 46546204-46546204 on Build GRCh38

View Full Product Details