Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51501546-51502079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIGLEC12
Entrez Gene ID
89858
Gene Name
sialic acid binding Ig like lectin 12 (gene/pseudogene)
Gene Aliases
S2V, SIGLECL1, SLG, Siglec-XII
UniGene
Hs.249741
Gene Chromosome Location
Chr.19: 51491227-51501789 on Build GRCh38
Transcripts
NM_053003.3 XM_011527442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6899499 c.167C>T p.S56F Chr.19: 51501567-51501567 on Build GRCh38
1000067 c.9G>A p.L3L Chr.19: 51501725-51501725 on Build GRCh38
4648323 c.183C>A p.G61G Chr.19: 51501551-51501551 on Build GRCh38
1000068 c.6A>G p.L2L Chr.19: 51501728-51501728 on Build GRCh38
6868890 c.31C>A p.L11M Chr.19: 51501703-51501703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562282804 Chr.19: 51501581-51501581 on Build GRCh38
rs754281543 Chr.19: 51501935-51501935 on Build GRCh38
rs779174939 Chr.19: 51501640-51501640 on Build GRCh38
rs986051437 Chr.19: 51501817-51501817 on Build GRCh38
rs780128292 Chr.19: 51501566-51501566 on Build GRCh38
rs150618212 Chr.19: 51501642-51501642 on Build GRCh38
rs73557353 Chr.19: 51501973-51501973 on Build GRCh38
rs1005578418 Chr.19: 51501671-51501671 on Build GRCh38
rs932861744 Chr.19: 51501846-51501846 on Build GRCh38
rs1030128365 Chr.19: 51501958-51501958 on Build GRCh38
rs554159585 Chr.19: 51501923-51501923 on Build GRCh38
rs948938952 Chr.19: 51501899-51501899 on Build GRCh38
rs1024821828 Chr.19: 51501730-51501730 on Build GRCh38
rs747330609 Chr.19: 51501641-51501641 on Build GRCh38
rs368298593 Chr.19: 51501592-51501592 on Build GRCh38
rs954585483 Chr.19: 51501800-51501800 on Build GRCh38
rs924409216 Chr.19: 51501854-51501854 on Build GRCh38
rs989176246 Chr.19: 51501748-51501748 on Build GRCh38
rs867229008 Chr.19: 51501622-51501622 on Build GRCh38
rs16982743 Chr.19: 51501649-51501649 on Build GRCh38
rs773986532 Chr.19: 51501684-51501684 on Build GRCh38
rs761260283 Chr.19: 51501612-51501612 on Build GRCh38
rs755122986 Chr.19: 51501713-51501713 on Build GRCh38
rs756013982 Chr.19: 51501738-51501738 on Build GRCh38
rs572600906 Chr.19: 51501933-51501933 on Build GRCh38
rs1043197809 Chr.19: 51501839-51501839 on Build GRCh38
rs766722884 Chr.19: 51501592-51501594 on Build GRCh38
rs73051368 Chr.19: 51502010-51502010 on Build GRCh38
rs989533456 Chr.19: 51501588-51501588 on Build GRCh38
rs746889063 Chr.19: 51501573-51501573 on Build GRCh38
rs957729266 Chr.19: 51501587-51501587 on Build GRCh38
rs201603484 Chr.19: 51501666-51501666 on Build GRCh38
rs766642019 Chr.19: 51501705-51501705 on Build GRCh38
rs765933544 Chr.19: 51501596-51501596 on Build GRCh38
rs773548774 Chr.19: 51501782-51501782 on Build GRCh38
rs771121123 Chr.19: 51502045-51502045 on Build GRCh38
rs771560541 Chr.19: 51501763-51501763 on Build GRCh38
rs910188872 Chr.19: 51501829-51501829 on Build GRCh38
rs536985630 Chr.19: 51501772-51501772 on Build GRCh38
rs144228959 Chr.19: 51501613-51501613 on Build GRCh38
rs770777994 Chr.19: 51501683-51501683 on Build GRCh38
rs764719892 Chr.19: 51501615-51501615 on Build GRCh38
rs200523709 Chr.19: 51501704-51501704 on Build GRCh38
rs546182902 Chr.19: 51501654-51501654 on Build GRCh38
rs10409414 Chr.19: 51501883-51501883 on Build GRCh38
rs769585114 Chr.19: 51501664-51501664 on Build GRCh38
rs1005573688 Chr.19: 51501922-51501922 on Build GRCh38
rs774190349 Chr.19: 51501754-51501754 on Build GRCh38
rs565504103 Chr.19: 51501679-51501679 on Build GRCh38
rs745538990 Chr.19: 51501756-51501756 on Build GRCh38
rs986040249 Chr.19: 51501989-51501989 on Build GRCh38
rs770686048 Chr.19: 51501743-51501743 on Build GRCh38
rs371419669 Chr.19: 51501739-51501739 on Build GRCh38
rs768582710 Chr.19: 51501574-51501574 on Build GRCh38
rs139701790 Chr.19: 51501690-51501690 on Build GRCh38
rs1057297852 Chr.19: 51501628-51501628 on Build GRCh38
rs772884464 Chr.19: 51501591-51501591 on Build GRCh38
rs1006041770 Chr.19: 51501718-51501718 on Build GRCh38
rs776282780 Chr.19: 51501580-51501580 on Build GRCh38
rs369228775 Chr.19: 51501838-51501838 on Build GRCh38
rs781237187 Chr.19: 51501656-51501656 on Build GRCh38
rs752417488 Chr.19: 51501611-51501612 on Build GRCh38
rs947088142 Chr.19: 51501586-51501586 on Build GRCh38
rs781156649 Chr.19: 51501719-51501719 on Build GRCh38
rs976875777 Chr.19: 51501826-51501826 on Build GRCh38
rs923109932 Chr.19: 51501774-51501774 on Build GRCh38
rs376073863 Chr.19: 51501783-51501783 on Build GRCh38
rs542311484 Chr.19: 51501900-51501900 on Build GRCh38
rs764479890 Chr.19: 51501686-51501686 on Build GRCh38
rs377448422 Chr.19: 51501685-51501685 on Build GRCh38
rs1037035175 Chr.19: 51501891-51501891 on Build GRCh38
rs148809065 Chr.19: 51501624-51501624 on Build GRCh38
rs73557352 Chr.19: 51501912-51501912 on Build GRCh38
rs192306330 Chr.19: 51501736-51501736 on Build GRCh38
rs751613292 Chr.19: 51501706-51501706 on Build GRCh38
rs773483529 Chr.19: 51501946-51501946 on Build GRCh38
rs765580092 Chr.19: 51501698-51501698 on Build GRCh38
rs781695255 Chr.19: 51501651-51501651 on Build GRCh38
rs769846346 Chr.19: 51501836-51501836 on Build GRCh38
rs769305374 Chr.19: 51501589-51501589 on Build GRCh38
rs747862814 Chr.19: 51501661-51501661 on Build GRCh38
rs1029392606 Chr.19: 51502011-51502011 on Build GRCh38
rs1057200375 Chr.19: 51501869-51501869 on Build GRCh38
rs143800743 Chr.19: 51501600-51501600 on Build GRCh38
rs141817270 Chr.19: 51501636-51501636 on Build GRCh38
rs76945124 Chr.19: 51501584-51501584 on Build GRCh38
rs192276046 Chr.19: 51502038-51502038 on Build GRCh38
rs888441086 Chr.19: 51501921-51501921 on Build GRCh38
rs897113270 Chr.19: 51501909-51501909 on Build GRCh38
rs772674474 Chr.19: 51501676-51501684 on Build GRCh38
rs957929080 Chr.19: 51501741-51501741 on Build GRCh38
rs567183867 Chr.19: 51501682-51501682 on Build GRCh38
rs868019260 Chr.19: 51501776-51501776 on Build GRCh38
rs774873541 Chr.19: 51501764-51501764 on Build GRCh38
rs756443355 Chr.19: 51501585-51501586 on Build GRCh38
rs1033483905 Chr.19: 51501934-51501934 on Build GRCh38
rs373307996 Chr.19: 51502008-51502008 on Build GRCh38
rs934812259 Chr.19: 51501855-51501855 on Build GRCh38
rs559743761 Chr.19: 51501765-51501765 on Build GRCh38

View Full Product Details