Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52046665-52047178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF432
Entrez Gene ID
9668
Gene Name
zinc finger protein 432
Gene Aliases
-
UniGene
Hs.655934
Gene Chromosome Location
Chr.19: 52031379-52049124 on Build GRCh38
Transcripts
NM_001322284.1 NM_001322285.1 NM_014650.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5600907 c.13C>T p.Q5* Chr.19: 52046856-52046856 on Build GRCh38
5073523 c.3G>T p.M1I Chr.19: 52046866-52046866 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751194654 Chr.19: 52047155-52047155 on Build GRCh38
rs752993675 Chr.19: 52046816-52046816 on Build GRCh38
rs115184158 Chr.19: 52046831-52046831 on Build GRCh38
rs771784760 Chr.19: 52046853-52046853 on Build GRCh38
rs1020775604 Chr.19: 52046833-52046833 on Build GRCh38
rs929567704 Chr.19: 52046694-52046694 on Build GRCh38
rs199524883 Chr.19: 52046912-52046912 on Build GRCh38
rs988230769 Chr.19: 52046811-52046811 on Build GRCh38
rs1018012759 Chr.19: 52047103-52047103 on Build GRCh38
rs774736328 Chr.19: 52046913-52046913 on Build GRCh38
rs766722539 Chr.19: 52047096-52047096 on Build GRCh38
rs184490417 Chr.19: 52046971-52046971 on Build GRCh38
rs1032550060 Chr.19: 52046720-52046720 on Build GRCh38
rs761753128 Chr.19: 52046835-52046835 on Build GRCh38
rs377603622 Chr.19: 52047030-52047030 on Build GRCh38
rs980814776 Chr.19: 52046875-52046875 on Build GRCh38
rs558167572 Chr.19: 52047013-52047013 on Build GRCh38
rs781420406 Chr.19: 52046806-52046806 on Build GRCh38
rs763368344 Chr.19: 52046910-52046910 on Build GRCh38
rs746837272 Chr.19: 52046843-52046843 on Build GRCh38
rs926645286 Chr.19: 52046897-52046897 on Build GRCh38
rs942002274 Chr.19: 52046945-52046945 on Build GRCh38
rs530663650 Chr.19: 52046706-52046706 on Build GRCh38
rs1002328473 Chr.19: 52046765-52046765 on Build GRCh38
rs776177011 Chr.19: 52046901-52046901 on Build GRCh38
rs555430068 Chr.19: 52047122-52047122 on Build GRCh38
rs778044976 Chr.19: 52046822-52046822 on Build GRCh38
rs113457206 Chr.19: 52046894-52046894 on Build GRCh38
rs576337953 Chr.19: 52047104-52047104 on Build GRCh38
rs775084967 Chr.19: 52046859-52046859 on Build GRCh38
rs541279009 Chr.19: 52047002-52047002 on Build GRCh38
rs55932673 Chr.19: 52046949-52046949 on Build GRCh38
rs998078576 Chr.19: 52046944-52046944 on Build GRCh38
rs567509634 Chr.19: 52046827-52046827 on Build GRCh38
rs759899818 Chr.19: 52046920-52046920 on Build GRCh38
rs374757637 Chr.19: 52046987-52046987 on Build GRCh38
rs1046690542 Chr.19: 52047121-52047122 on Build GRCh38
rs1032559778 Chr.19: 52047138-52047138 on Build GRCh38
rs768161864 Chr.19: 52046738-52046738 on Build GRCh38
rs113173256 Chr.19: 52046796-52046796 on Build GRCh38
rs954734129 Chr.19: 52046963-52046963 on Build GRCh38
rs756227983 Chr.19: 52046817-52046817 on Build GRCh38
rs17779341 Chr.19: 52047148-52047148 on Build GRCh38
rs746706797 Chr.19: 52046860-52046860 on Build GRCh38
rs890894904 Chr.19: 52047043-52047043 on Build GRCh38
rs762480037 Chr.19: 52046995-52046995 on Build GRCh38
rs768132710 Chr.19: 52046884-52046884 on Build GRCh38
rs756151745 Chr.19: 52046902-52046902 on Build GRCh38
rs745616994 Chr.19: 52046851-52046851 on Build GRCh38
rs1046072102 Chr.19: 52046702-52046702 on Build GRCh38
rs1047305699 Chr.19: 52047021-52047021 on Build GRCh38
rs16983408 Chr.19: 52047111-52047111 on Build GRCh38
rs1008419164 Chr.19: 52047083-52047083 on Build GRCh38

View Full Product Details