Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52804829-52805346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF28
Entrez Gene ID
7576
Gene Name
zinc finger protein 28
Gene Aliases
KOX24
UniGene
Hs.732407
Gene Chromosome Location
Chr.19: 52797408-52821686 on Build GRCh38
Transcripts
NM_006969.3 NR_036599.1 NR_036600.1 XM_006723354.3 XM_011527262.2

SNP Detail

SNP ID Chromosome Location
rs7259738 Chr.19: 52805077-52805077 on Build GRCh38
rs774812977 Chr.19: 52805274-52805274 on Build GRCh38
rs369935614 Chr.19: 52805273-52805273 on Build GRCh38
rs535712252 Chr.19: 52805001-52805001 on Build GRCh38
rs901936803 Chr.19: 52805256-52805256 on Build GRCh38
rs7259580 Chr.19: 52804996-52804996 on Build GRCh38
rs375524185 Chr.19: 52805272-52805272 on Build GRCh38
rs1041749052 Chr.19: 52805196-52805196 on Build GRCh38
rs538998132 Chr.19: 52804864-52804864 on Build GRCh38
rs572282151 Chr.19: 52805063-52805063 on Build GRCh38
rs1006573532 Chr.19: 52805300-52805300 on Build GRCh38
rs758261275 Chr.19: 52805307-52805307 on Build GRCh38
rs35221997 Chr.19: 52805201-52805202 on Build GRCh38
rs1054904157 Chr.19: 52804865-52804865 on Build GRCh38
rs1054449306 Chr.19: 52805132-52805132 on Build GRCh38
rs937321188 Chr.19: 52804857-52804857 on Build GRCh38
rs149281556 Chr.19: 52805004-52805004 on Build GRCh38
rs552621861 Chr.19: 52804914-52804914 on Build GRCh38
rs538229151 Chr.19: 52804943-52804943 on Build GRCh38
rs1020057789 Chr.19: 52804909-52804909 on Build GRCh38
rs543406445 Chr.19: 52805193-52805193 on Build GRCh38
rs570794434 Chr.19: 52804923-52804923 on Build GRCh38
rs575999108 Chr.19: 52805157-52805157 on Build GRCh38
rs190495593 Chr.19: 52805228-52805228 on Build GRCh38
rs556630822 Chr.19: 52804954-52804954 on Build GRCh38
rs547689119 Chr.19: 52805264-52805264 on Build GRCh38
rs774095853 Chr.19: 52805127-52805127 on Build GRCh38
rs113268535 Chr.19: 52805162-52805162 on Build GRCh38
rs7259973 Chr.19: 52805128-52805128 on Build GRCh38
rs893162853 Chr.19: 52805150-52805150 on Build GRCh38
rs144324536 Chr.19: 52805273-52805274 on Build GRCh38
rs953458365 Chr.19: 52804990-52804990 on Build GRCh38
rs370514534 Chr.19: 52804967-52804967 on Build GRCh38
rs1029274635 Chr.19: 52805289-52805289 on Build GRCh38
rs186946393 Chr.19: 52805130-52805130 on Build GRCh38
rs7260017 Chr.19: 52805317-52805317 on Build GRCh38
rs946070900 Chr.19: 52805168-52805168 on Build GRCh38
rs893215057 Chr.19: 52804869-52804869 on Build GRCh38
rs917688053 Chr.19: 52805071-52805071 on Build GRCh38
rs867679521 Chr.19: 52804983-52804983 on Build GRCh38
rs997765120 Chr.19: 52804972-52804972 on Build GRCh38
rs529938029 Chr.19: 52804882-52804882 on Build GRCh38
rs200444053 Chr.19: 52804989-52804989 on Build GRCh38
rs566190327 Chr.19: 52805257-52805257 on Build GRCh38
rs889023666 Chr.19: 52805299-52805299 on Build GRCh38

View Full Product Details