Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53175313-53175712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF665
Entrez Gene ID
79788
Gene Name
zinc finger protein 665
Gene Aliases
ZFP160L
UniGene
Hs.745230
Gene Chromosome Location
Chr.19: 53163299-53193404 on Build GRCh38
Transcripts
NM_024733.3 XM_005259266.4 XM_011527325.2 XM_011527327.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6583924 c.111G>T p.M37I Chr.19: 53175476-53175476 on Build GRCh38
265115 c.87G>T p.Q29H Chr.19: 53175500-53175500 on Build GRCh38
1631074 c.87G>A p.Q29Q Chr.19: 53175500-53175500 on Build GRCh38
6085907 c.17G>T p.G6V Chr.19: 53175570-53175570 on Build GRCh38
1396073 c.53C>A p.S18Y Chr.19: 53175534-53175534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369778289 Chr.19: 53175440-53175440 on Build GRCh38
rs770797220 Chr.19: 53175557-53175558 on Build GRCh38
rs555028037 Chr.19: 53175679-53175679 on Build GRCh38
rs915188084 Chr.19: 53175365-53175365 on Build GRCh38
rs781135341 Chr.19: 53175589-53175589 on Build GRCh38
rs538663862 Chr.19: 53175492-53175493 on Build GRCh38
rs75067138 Chr.19: 53175352-53175352 on Build GRCh38
rs959436950 Chr.19: 53175355-53175355 on Build GRCh38
rs550462893 Chr.19: 53175377-53175377 on Build GRCh38
rs769859180 Chr.19: 53175520-53175520 on Build GRCh38
rs757507068 Chr.19: 53175438-53175438 on Build GRCh38
rs1032418725 Chr.19: 53175404-53175404 on Build GRCh38
rs372493765 Chr.19: 53175550-53175550 on Build GRCh38
rs1035152009 Chr.19: 53175524-53175526 on Build GRCh38
rs79131937 Chr.19: 53175405-53175405 on Build GRCh38
rs1001281122 Chr.19: 53175397-53175397 on Build GRCh38
rs772873273 Chr.19: 53175606-53175606 on Build GRCh38
rs373897910 Chr.19: 53175592-53175592 on Build GRCh38
rs746037212 Chr.19: 53175456-53175456 on Build GRCh38
rs200205628 Chr.19: 53175415-53175415 on Build GRCh38
rs780093659 Chr.19: 53175471-53175471 on Build GRCh38
rs762494459 Chr.19: 53175504-53175504 on Build GRCh38
rs141117052 Chr.19: 53175588-53175588 on Build GRCh38
rs575700570 Chr.19: 53175651-53175651 on Build GRCh38
rs754350804 Chr.19: 53175526-53175526 on Build GRCh38
rs866978994 Chr.19: 53175485-53175485 on Build GRCh38
rs759052818 Chr.19: 53175515-53175515 on Build GRCh38
rs747104790 Chr.19: 53175576-53175576 on Build GRCh38
rs181136298 Chr.19: 53175615-53175615 on Build GRCh38
rs779117155 Chr.19: 53175539-53175539 on Build GRCh38
rs199747280 Chr.19: 53175508-53175508 on Build GRCh38
rs4289099 Chr.19: 53175671-53175671 on Build GRCh38
rs980691898 Chr.19: 53175436-53175436 on Build GRCh38
rs377197349 Chr.19: 53175477-53175477 on Build GRCh38
rs769676753 Chr.19: 53175604-53175604 on Build GRCh38
rs372741529 Chr.19: 53175461-53175461 on Build GRCh38
rs374548324 Chr.19: 53175529-53175529 on Build GRCh38
rs80042365 Chr.19: 53175421-53175421 on Build GRCh38
rs987915199 Chr.19: 53175406-53175406 on Build GRCh38
rs143559625 Chr.19: 53175482-53175482 on Build GRCh38
rs772234934 Chr.19: 53175464-53175464 on Build GRCh38
rs146504762 Chr.19: 53175543-53175543 on Build GRCh38
rs1000870151 Chr.19: 53175633-53175633 on Build GRCh38
rs370109452 Chr.19: 53175481-53175481 on Build GRCh38
rs772954894 Chr.19: 53175501-53175501 on Build GRCh38
rs557609672 Chr.19: 53175617-53175617 on Build GRCh38
rs576042075 Chr.19: 53175641-53175641 on Build GRCh38
rs964037841 Chr.19: 53175367-53175367 on Build GRCh38
rs1032780683 Chr.19: 53175640-53175640 on Build GRCh38
rs758544425 Chr.19: 53175548-53175548 on Build GRCh38
rs900117307 Chr.19: 53175626-53175626 on Build GRCh38
rs376420963 Chr.19: 53175493-53175493 on Build GRCh38
rs768541502 Chr.19: 53175416-53175416 on Build GRCh38
rs774448274 Chr.19: 53175521-53175521 on Build GRCh38

View Full Product Details