Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53454523-53455036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF761
Entrez Gene ID
388561
Gene Name
zinc finger protein 761
Gene Aliases
ZNF468
UniGene
Hs.433293
Gene Chromosome Location
Chr.19: 53431974-53458261 on Build GRCh38
Transcripts
NM_001008401.3 NM_001289951.1 NM_001289952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7139563 c.275C>A p.S92* Chr.19: 53454944-53454944 on Build GRCh38
6355804 c.250G>A p.D84N Chr.19: 53454919-53454919 on Build GRCh38
368716 c.260G>T p.G87V Chr.19: 53454929-53454929 on Build GRCh38
126492 c.151G>A p.E51K Chr.19: 53454820-53454820 on Build GRCh38
712895 c.141G>T p.W47C Chr.19: 53454810-53454810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760471909 Chr.19: 53454975-53454975 on Build GRCh38
rs145308414 Chr.19: 53454912-53454912 on Build GRCh38
rs369482628 Chr.19: 53454684-53454684 on Build GRCh38
rs759096681 Chr.19: 53454693-53454693 on Build GRCh38
rs774535946 Chr.19: 53454768-53454768 on Build GRCh38
rs547390147 Chr.19: 53454817-53454817 on Build GRCh38
rs755121807 Chr.19: 53454845-53454845 on Build GRCh38
rs749493648 Chr.19: 53454757-53454757 on Build GRCh38
rs376099451 Chr.19: 53454766-53454766 on Build GRCh38
rs753853741 Chr.19: 53454920-53454920 on Build GRCh38
rs1984432 Chr.19: 53455009-53455009 on Build GRCh38
rs781761316 Chr.19: 53454961-53454961 on Build GRCh38
rs546012135 Chr.19: 53454580-53454580 on Build GRCh38
rs757125235 Chr.19: 53454951-53454951 on Build GRCh38
rs766988495 Chr.19: 53454818-53454818 on Build GRCh38
rs115009768 Chr.19: 53454895-53454895 on Build GRCh38
rs763232497 Chr.19: 53454674-53454674 on Build GRCh38
rs940081469 Chr.19: 53454579-53454579 on Build GRCh38
rs752478792 Chr.19: 53454642-53454642 on Build GRCh38
rs367989673 Chr.19: 53454972-53454972 on Build GRCh38
rs553863305 Chr.19: 53454944-53454944 on Build GRCh38
rs374389762 Chr.19: 53454962-53454962 on Build GRCh38
rs764896268 Chr.19: 53454696-53454696 on Build GRCh38
rs745751809 Chr.19: 53454997-53454997 on Build GRCh38
rs200311936 Chr.19: 53454664-53454664 on Build GRCh38
rs756184648 Chr.19: 53454749-53454749 on Build GRCh38
rs764807877 Chr.19: 53454789-53454789 on Build GRCh38
rs781204680 Chr.19: 53454743-53454743 on Build GRCh38
rs150807379 Chr.19: 53454848-53454848 on Build GRCh38
rs531744761 Chr.19: 53454974-53454974 on Build GRCh38
rs769946281 Chr.19: 53454667-53454667 on Build GRCh38
rs377263462 Chr.19: 53454675-53454675 on Build GRCh38
rs764503301 Chr.19: 53454621-53454621 on Build GRCh38
rs893449218 Chr.19: 53454900-53454900 on Build GRCh38
rs552789932 Chr.19: 53454956-53454956 on Build GRCh38
rs770088588 Chr.19: 53454609-53454609 on Build GRCh38
rs754544707 Chr.19: 53454634-53454634 on Build GRCh38
rs766216657 Chr.19: 53454976-53454976 on Build GRCh38
rs148252402 Chr.19: 53454692-53454692 on Build GRCh38
rs1020607122 Chr.19: 53454924-53454924 on Build GRCh38
rs762645345 Chr.19: 53454796-53454796 on Build GRCh38
rs367690444 Chr.19: 53454632-53454632 on Build GRCh38
rs180718989 Chr.19: 53454597-53454597 on Build GRCh38
rs778603918 Chr.19: 53454971-53454971 on Build GRCh38
rs370359488 Chr.19: 53454992-53454992 on Build GRCh38
rs752061189 Chr.19: 53454627-53454627 on Build GRCh38
rs757076604 Chr.19: 53454723-53454723 on Build GRCh38
rs370203710 Chr.19: 53454858-53454858 on Build GRCh38
rs753767903 Chr.19: 53454999-53454999 on Build GRCh38
rs768097146 Chr.19: 53454690-53454690 on Build GRCh38
rs373350015 Chr.19: 53454880-53454880 on Build GRCh38
rs780063567 Chr.19: 53454831-53454831 on Build GRCh38
rs777778106 Chr.19: 53454653-53454653 on Build GRCh38
rs376501395 Chr.19: 53454897-53454897 on Build GRCh38
rs141985937 Chr.19: 53454850-53454850 on Build GRCh38
rs202218689 Chr.19: 53454701-53454701 on Build GRCh38
rs780150832 Chr.19: 53454752-53454752 on Build GRCh38
rs752391056 Chr.19: 53454697-53454697 on Build GRCh38
rs777688365 Chr.19: 53454710-53454710 on Build GRCh38
rs770177327 Chr.19: 53454866-53454867 on Build GRCh38
rs751402595 Chr.19: 53454715-53454715 on Build GRCh38
rs748546093 Chr.19: 53454770-53454770 on Build GRCh38
rs778924770 Chr.19: 53454934-53454934 on Build GRCh38
rs771587441 Chr.19: 53454875-53454875 on Build GRCh38
rs750245548 Chr.19: 53454910-53454910 on Build GRCh38
rs773752751 Chr.19: 53454782-53454782 on Build GRCh38
rs749397502 Chr.19: 53454842-53454842 on Build GRCh38
rs759629266 Chr.19: 53454604-53454604 on Build GRCh38
rs375085830 Chr.19: 53454631-53454631 on Build GRCh38
rs201558114 Chr.19: 53454683-53454683 on Build GRCh38
rs369430266 Chr.19: 53454781-53454781 on Build GRCh38
rs148779203 Chr.19: 53454947-53454947 on Build GRCh38
rs770949279 Chr.19: 53454656-53454656 on Build GRCh38
rs757063871 Chr.19: 53454808-53454808 on Build GRCh38
rs370693285 Chr.19: 53454948-53454948 on Build GRCh38
rs147142607 Chr.19: 53454882-53454882 on Build GRCh38
rs761329848 Chr.19: 53454784-53454784 on Build GRCh38
rs141314537 Chr.19: 53454807-53454807 on Build GRCh38
rs758595093 Chr.19: 53454945-53454945 on Build GRCh38
rs267605656 Chr.19: 53454946-53454946 on Build GRCh38
rs771507880 Chr.19: 53454960-53454960 on Build GRCh38
rs752272159 Chr.19: 53454837-53454837 on Build GRCh38
rs373360886 Chr.19: 53454795-53454795 on Build GRCh38
rs34553879 Chr.19: 53454849-53454849 on Build GRCh38
rs762425731 Chr.19: 53454630-53454630 on Build GRCh38
rs148293146 Chr.19: 53454611-53454611 on Build GRCh38
rs1009231862 Chr.19: 53454699-53454699 on Build GRCh38
rs778709212 Chr.19: 53454637-53454637 on Build GRCh38
rs187302990 Chr.19: 53454855-53454855 on Build GRCh38
rs762566457 Chr.19: 53454881-53454881 on Build GRCh38
rs565786134 Chr.19: 53454830-53454830 on Build GRCh38
rs563859985 Chr.19: 53454765-53454765 on Build GRCh38
rs143107546 Chr.19: 53454668-53454668 on Build GRCh38
rs761480131 Chr.19: 53454964-53454964 on Build GRCh38
rs773785083 Chr.19: 53454963-53454963 on Build GRCh38
rs781093345 Chr.19: 53454660-53454660 on Build GRCh38
rs753654707 Chr.19: 53454890-53454890 on Build GRCh38
rs146306324 Chr.19: 53454894-53454894 on Build GRCh38
rs758253192 Chr.19: 53454652-53454652 on Build GRCh38
rs2708743 Chr.19: 53454872-53454872 on Build GRCh38
rs755035632 Chr.19: 53454923-53454923 on Build GRCh38
rs756008918 Chr.19: 53454911-53454911 on Build GRCh38

View Full Product Details