Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54145509-54146016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843744 c.522G>C p.E174D Chr.19: 54145636-54145636 on Build GRCh38
3797499 c.211G>A p.E71K Chr.19: 54145960-54145960 on Build GRCh38
1578380 c.808delA p.I270fs*65 Chr.19: 54146014-54146014 on Build GRCh38
7349751 c.562C>T p.R188C Chr.19: 54145676-54145676 on Build GRCh38
3721294 c.259T>C p.S87P Chr.19: 54146008-54146008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767913658 Chr.19: 54145916-54145916 on Build GRCh38
rs764309033 Chr.19: 54145937-54145937 on Build GRCh38
rs199874706 Chr.19: 54145856-54145856 on Build GRCh38
rs773254399 Chr.19: 54145855-54145855 on Build GRCh38
rs754508390 Chr.19: 54145961-54145961 on Build GRCh38
rs774146815 Chr.19: 54145576-54145576 on Build GRCh38
rs375211815 Chr.19: 54145863-54145863 on Build GRCh38
rs760901337 Chr.19: 54145684-54145684 on Build GRCh38
rs747901070 Chr.19: 54145853-54145853 on Build GRCh38
rs368181282 Chr.19: 54145600-54145600 on Build GRCh38
rs748810728 Chr.19: 54145780-54145780 on Build GRCh38
rs149814896 Chr.19: 54145651-54145651 on Build GRCh38
rs11541604 Chr.19: 54145979-54145979 on Build GRCh38
rs754139680 Chr.19: 54145626-54145626 on Build GRCh38
rs373987800 Chr.19: 54145559-54145559 on Build GRCh38
rs1042601186 Chr.19: 54145589-54145589 on Build GRCh38
rs144796513 Chr.19: 54145708-54145708 on Build GRCh38
rs758067096 Chr.19: 54145971-54145971 on Build GRCh38
rs751930267 Chr.19: 54145570-54145571 on Build GRCh38
rs143683346 Chr.19: 54145926-54145926 on Build GRCh38
rs772043379 Chr.19: 54145854-54145854 on Build GRCh38
rs371650779 Chr.19: 54145603-54145603 on Build GRCh38
rs191843066 Chr.19: 54145825-54145825 on Build GRCh38
rs73936620 Chr.19: 54145835-54145835 on Build GRCh38
rs764910094 Chr.19: 54145963-54145963 on Build GRCh38
rs778260176 Chr.19: 54145669-54145669 on Build GRCh38
rs952488775 Chr.19: 54145720-54145720 on Build GRCh38
rs753193649 Chr.19: 54145654-54145654 on Build GRCh38
rs142169688 Chr.19: 54145943-54145943 on Build GRCh38
rs369073653 Chr.19: 54145867-54145867 on Build GRCh38
rs755778475 Chr.19: 54145865-54145865 on Build GRCh38
rs777770096 Chr.19: 54145930-54145930 on Build GRCh38
rs767574789 Chr.19: 54145590-54145590 on Build GRCh38
rs751333148 Chr.19: 54145986-54145986 on Build GRCh38
rs587608163 Chr.19: 54145729-54145729 on Build GRCh38
rs764506972 Chr.19: 54145888-54145888 on Build GRCh38
rs112551303 Chr.19: 54145833-54145833 on Build GRCh38
rs765395773 Chr.19: 54145807-54145807 on Build GRCh38
rs587764978 Chr.19: 54145965-54145965 on Build GRCh38
rs779847200 Chr.19: 54145892-54145894 on Build GRCh38
rs769266626 Chr.19: 54145544-54145544 on Build GRCh38
rs775228409 Chr.19: 54145557-54145557 on Build GRCh38
rs753285609 Chr.19: 54145892-54145892 on Build GRCh38
rs373632528 Chr.19: 54145655-54145655 on Build GRCh38
rs372705849 Chr.19: 54145810-54145810 on Build GRCh38
rs868238552 Chr.19: 54145937-54145938 on Build GRCh38
rs765407340 Chr.19: 54145874-54145874 on Build GRCh38
rs375948136 Chr.19: 54145817-54145817 on Build GRCh38
rs779539051 Chr.19: 54145703-54145703 on Build GRCh38
rs775714798 Chr.19: 54145878-54145878 on Build GRCh38
rs34286651 Chr.19: 54145702-54145702 on Build GRCh38
rs1004235021 Chr.19: 54145628-54145628 on Build GRCh38
rs141264166 Chr.19: 54145792-54145792 on Build GRCh38
rs759861030 Chr.19: 54145793-54145793 on Build GRCh38
rs776719450 Chr.19: 54145866-54145866 on Build GRCh38
rs111840559 Chr.19: 54145650-54145650 on Build GRCh38
rs761899974 Chr.19: 54145579-54145579 on Build GRCh38
rs768532413 Chr.19: 54145561-54145561 on Build GRCh38
rs753475896 Chr.19: 54145930-54145931 on Build GRCh38
rs745399657 Chr.19: 54145534-54145534 on Build GRCh38
rs777576063 Chr.19: 54145973-54145973 on Build GRCh38
rs34936413 Chr.19: 54145765-54145765 on Build GRCh38
rs774229944 Chr.19: 54145733-54145733 on Build GRCh38
rs1012509242 Chr.19: 54145656-54145656 on Build GRCh38
rs755267492 Chr.19: 54145633-54145633 on Build GRCh38
rs754467152 Chr.19: 54145923-54145923 on Build GRCh38
rs757720625 Chr.19: 54145903-54145903 on Build GRCh38
rs373852003 Chr.19: 54145841-54145841 on Build GRCh38
rs746793484 Chr.19: 54145932-54145932 on Build GRCh38
rs200727273 Chr.19: 54145675-54145675 on Build GRCh38
rs770901969 Chr.19: 54145862-54145862 on Build GRCh38
rs369057695 Chr.19: 54145838-54145838 on Build GRCh38
rs772094308 Chr.19: 54145746-54145746 on Build GRCh38
rs35511193 Chr.19: 54145771-54145771 on Build GRCh38
rs758265246 Chr.19: 54145927-54145927 on Build GRCh38
rs778684787 Chr.19: 54145924-54145924 on Build GRCh38
rs906687418 Chr.19: 54145594-54145594 on Build GRCh38
rs36665 Chr.19: 54145935-54145935 on Build GRCh38
rs765894239 Chr.19: 54145850-54145850 on Build GRCh38
rs200821584 Chr.19: 54145917-54145917 on Build GRCh38
rs778738017 Chr.19: 54145845-54145845 on Build GRCh38
rs367791191 Chr.19: 54145744-54145744 on Build GRCh38
rs762116071 Chr.19: 54145938-54145938 on Build GRCh38
rs921159903 Chr.19: 54145823-54145823 on Build GRCh38
rs879222513 Chr.19: 54145869-54145869 on Build GRCh38

View Full Product Details