Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54875263-54875768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAR
Entrez Gene ID
2204
Gene Name
Fc fragment of IgA receptor
Gene Aliases
CD89, CTB-61M7.2, FcalphaRI
UniGene
Hs.659872
Gene Chromosome Location
Chr.19: 54874093-54891420 on Build GRCh38
Transcripts
NM_002000.3 NM_133269.3 NM_133271.3 NM_133272.3 NM_133273.3 NM_133274.3 NM_133277.3 NM_133278.3 XM_011526625.2 XM_017026473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903999 c.35-4C>T p.? Chr.19: 54875326-54875326 on Build GRCh38
7105997 c.35-1G>C p.? Chr.19: 54875329-54875329 on Build GRCh38
5505453 c.54G>T p.R18S Chr.19: 54875349-54875349 on Build GRCh38
4127882 c.35-10T>C p.? Chr.19: 54875320-54875320 on Build GRCh38
5505452 c.54G>T p.R18S Chr.19: 54875349-54875349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375350117 Chr.19: 54875322-54875322 on Build GRCh38
rs868002531 Chr.19: 54875353-54875353 on Build GRCh38
rs992996020 Chr.19: 54875356-54875356 on Build GRCh38
rs1008591480 Chr.19: 54875726-54875726 on Build GRCh38
rs769869252 Chr.19: 54875321-54875321 on Build GRCh38
rs746861847 Chr.19: 54875301-54875301 on Build GRCh38
rs762348116 Chr.19: 54875522-54875522 on Build GRCh38
rs994700114 Chr.19: 54875603-54875603 on Build GRCh38
rs1016584224 Chr.19: 54875374-54875374 on Build GRCh38
rs900304544 Chr.19: 54875549-54875549 on Build GRCh38
rs111848981 Chr.19: 54875716-54875716 on Build GRCh38
rs374865895 Chr.19: 54875344-54875344 on Build GRCh38
rs895834117 Chr.19: 54875602-54875602 on Build GRCh38
rs904774020 Chr.19: 54875393-54875393 on Build GRCh38
rs587623569 Chr.19: 54875714-54875714 on Build GRCh38
rs1025954192 Chr.19: 54875715-54875715 on Build GRCh38
rs77887839 Chr.19: 54875402-54875402 on Build GRCh38
rs766346316 Chr.19: 54875324-54875324 on Build GRCh38
rs749131959 Chr.19: 54875398-54875398 on Build GRCh38
rs561636353 Chr.19: 54875365-54875365 on Build GRCh38
rs775528912 Chr.19: 54875389-54875389 on Build GRCh38
rs765932099 Chr.19: 54875346-54875346 on Build GRCh38
rs184438611 Chr.19: 54875641-54875641 on Build GRCh38
rs763057330 Chr.19: 54875396-54875396 on Build GRCh38
rs587725836 Chr.19: 54875677-54875677 on Build GRCh38
rs147036661 Chr.19: 54875336-54875336 on Build GRCh38
rs1008625008 Chr.19: 54875505-54875505 on Build GRCh38
rs948926498 Chr.19: 54875371-54875371 on Build GRCh38
rs950621322 Chr.19: 54875639-54875639 on Build GRCh38
rs746135976 Chr.19: 54875381-54875381 on Build GRCh38
rs1027947839 Chr.19: 54875617-54875617 on Build GRCh38
rs553154190 Chr.19: 54875350-54875350 on Build GRCh38
rs762022307 Chr.19: 54875404-54875404 on Build GRCh38
rs1044443251 Chr.19: 54875608-54875608 on Build GRCh38
rs1017757314 Chr.19: 54875730-54875730 on Build GRCh38
rs201138186 Chr.19: 54875362-54875362 on Build GRCh38
rs764631672 Chr.19: 54875333-54875333 on Build GRCh38
rs926189121 Chr.19: 54875542-54875542 on Build GRCh38
rs781208047 Chr.19: 54875376-54875376 on Build GRCh38
rs189710867 Chr.19: 54875645-54875645 on Build GRCh38
rs960095397 Chr.19: 54875687-54875687 on Build GRCh38
rs905977323 Chr.19: 54875667-54875667 on Build GRCh38
rs769820073 Chr.19: 54875387-54875387 on Build GRCh38
rs753390737 Chr.19: 54875413-54875413 on Build GRCh38
rs766062961 Chr.19: 54875713-54875713 on Build GRCh38
rs978980473 Chr.19: 54875541-54875541 on Build GRCh38
rs181014076 Chr.19: 54875528-54875528 on Build GRCh38
rs587737640 Chr.19: 54875738-54875738 on Build GRCh38
rs770247959 Chr.19: 54875596-54875596 on Build GRCh38
rs587751198 Chr.19: 54875569-54875569 on Build GRCh38
rs11084377 Chr.19: 54875434-54875434 on Build GRCh38
rs746768907 Chr.19: 54875368-54875368 on Build GRCh38
rs776971511 Chr.19: 54875316-54875316 on Build GRCh38
rs759479854 Chr.19: 54875319-54875319 on Build GRCh38
rs771056955 Chr.19: 54875311-54875311 on Build GRCh38
rs770775372 Chr.19: 54875375-54875375 on Build GRCh38
rs148076512 Chr.19: 54875510-54875510 on Build GRCh38
rs773694557 Chr.19: 54875605-54875605 on Build GRCh38
rs771811507 Chr.19: 54875501-54875503 on Build GRCh38
rs777925365 Chr.19: 54875366-54875366 on Build GRCh38
rs146580216 Chr.19: 54875349-54875349 on Build GRCh38
rs8112766 Chr.19: 54875464-54875464 on Build GRCh38
rs987230997 Chr.19: 54875441-54875441 on Build GRCh38
rs1004209738 Chr.19: 54875664-54875664 on Build GRCh38
rs979300685 Chr.19: 54875741-54875741 on Build GRCh38
rs763477424 Chr.19: 54875712-54875712 on Build GRCh38
rs747027258 Chr.19: 54875452-54875452 on Build GRCh38
rs763477126 Chr.19: 54875326-54875326 on Build GRCh38
rs777731467 Chr.19: 54875300-54875300 on Build GRCh38
rs371586148 Chr.19: 54875287-54875287 on Build GRCh38
rs1053016618 Chr.19: 54875585-54875585 on Build GRCh38
rs587715649 Chr.19: 54875533-54875533 on Build GRCh38

View Full Product Details