Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46576479-46576979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506142
Entrez Gene ID
100506142
Gene Name
uncharacterized LOC100506142
Gene Aliases
-
UniGene
Hs.658541
Gene Chromosome Location
Chr.2: 46568256-46580238 on Build GRCh38
Transcripts
NR_104182.1 NM_012249.3 XM_005264229.2 XM_011532726.2 XM_011532728.2 XM_017003716.1

Gene Symbol
RHOQ
Entrez Gene ID
23433
Gene Name
ras homolog family member Q
Gene Aliases
ARHQ, HEL-S-42, RASL7A, TC10, TC10A
UniGene
Hs.709193
Gene Chromosome Location
Chr.2: 46542409-46584688 on Build GRCh38
Transcripts
NR_104182.1 NM_012249.3 XM_005264229.2 XM_011532726.2 XM_011532728.2 XM_017003716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839601 c.401G>T p.R134I Chr.2: 46576595-46576595 on Build GRCh38
3372794 c.409G>C p.D137H Chr.2: 46576603-46576603 on Build GRCh38
1176707 c.457A>C p.K153Q Chr.2: 46576651-46576651 on Build GRCh38
1408216 c.384C>T p.D128D Chr.2: 46576578-46576578 on Build GRCh38
6402386 c.406A>G p.N136D Chr.2: 46576600-46576600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776060237 Chr.2: 46576538-46576538 on Build GRCh38
rs1039884171 Chr.2: 46576604-46576604 on Build GRCh38
rs919265104 Chr.2: 46576819-46576819 on Build GRCh38
rs757089034 Chr.2: 46576570-46576570 on Build GRCh38
rs1041031811 Chr.2: 46576941-46576941 on Build GRCh38
rs766509302 Chr.2: 46576662-46576662 on Build GRCh38
rs764371724 Chr.2: 46576817-46576817 on Build GRCh38
rs750487279 Chr.2: 46576578-46576578 on Build GRCh38
rs997908018 Chr.2: 46576712-46576712 on Build GRCh38
rs749708433 Chr.2: 46576517-46576517 on Build GRCh38
rs926733670 Chr.2: 46576748-46576748 on Build GRCh38
rs980949014 Chr.2: 46576740-46576740 on Build GRCh38
rs770095456 Chr.2: 46576514-46576514 on Build GRCh38
rs557938105 Chr.2: 46576953-46576953 on Build GRCh38
rs1054625376 Chr.2: 46576927-46576932 on Build GRCh38
rs780593963 Chr.2: 46576516-46576516 on Build GRCh38
rs762863999 Chr.2: 46576543-46576543 on Build GRCh38
rs763415592 Chr.2: 46576512-46576512 on Build GRCh38
rs936171950 Chr.2: 46576597-46576597 on Build GRCh38
rs199805157 Chr.2: 46576696-46576696 on Build GRCh38
rs774977046 Chr.2: 46576521-46576521 on Build GRCh38
rs756317849 Chr.2: 46576700-46576700 on Build GRCh38
rs571024797 Chr.2: 46576683-46576683 on Build GRCh38
rs76875301 Chr.2: 46576873-46576873 on Build GRCh38
rs186583689 Chr.2: 46576869-46576869 on Build GRCh38
rs762449415 Chr.2: 46576647-46576647 on Build GRCh38
rs1022525969 Chr.2: 46576692-46576692 on Build GRCh38
rs374080824 Chr.2: 46576654-46576654 on Build GRCh38
rs780471058 Chr.2: 46576592-46576592 on Build GRCh38
rs1050798045 Chr.2: 46576952-46576952 on Build GRCh38
rs762222260 Chr.2: 46576524-46576524 on Build GRCh38
rs956321344 Chr.2: 46576668-46576668 on Build GRCh38
rs748701592 Chr.2: 46576605-46576605 on Build GRCh38
rs899621783 Chr.2: 46576917-46576917 on Build GRCh38
rs570102350 Chr.2: 46576829-46576829 on Build GRCh38
rs781448226 Chr.2: 46576571-46576571 on Build GRCh38
rs759208401 Chr.2: 46576539-46576539 on Build GRCh38
rs188741708 Chr.2: 46576939-46576939 on Build GRCh38
rs575355824 Chr.2: 46576929-46576929 on Build GRCh38
rs991848149 Chr.2: 46576846-46576846 on Build GRCh38
rs755435974 Chr.2: 46576596-46576596 on Build GRCh38
rs535877672 Chr.2: 46576880-46576880 on Build GRCh38
rs766559792 Chr.2: 46576703-46576703 on Build GRCh38
rs779078356 Chr.2: 46576603-46576603 on Build GRCh38
rs917223151 Chr.2: 46576857-46576857 on Build GRCh38
rs368002446 Chr.2: 46576542-46576542 on Build GRCh38
rs900814140 Chr.2: 46576949-46576949 on Build GRCh38
rs199596438 Chr.2: 46576645-46576645 on Build GRCh38
rs997821825 Chr.2: 46576951-46576951 on Build GRCh38
rs751623190 Chr.2: 46576670-46576671 on Build GRCh38
rs773972844 Chr.2: 46576682-46576682 on Build GRCh38
rs767599247 Chr.2: 46576689-46576689 on Build GRCh38
rs143446856 Chr.2: 46576621-46576621 on Build GRCh38
rs770479250 Chr.2: 46576533-46576533 on Build GRCh38
rs534618101 Chr.2: 46576863-46576863 on Build GRCh38
rs960819853 Chr.2: 46576784-46576784 on Build GRCh38
rs768876248 Chr.2: 46576519-46576519 on Build GRCh38
rs745432428 Chr.2: 46576627-46576627 on Build GRCh38
rs764911792 Chr.2: 46576540-46576540 on Build GRCh38
rs542346871 Chr.2: 46576545-46576545 on Build GRCh38
rs550279558 Chr.2: 46576729-46576729 on Build GRCh38
rs1046929166 Chr.2: 46576504-46576504 on Build GRCh38
rs1016744749 Chr.2: 46576823-46576823 on Build GRCh38
rs1029330742 Chr.2: 46576652-46576652 on Build GRCh38
rs1005327360 Chr.2: 46576754-46576754 on Build GRCh38
rs899570225 Chr.2: 46576606-46576606 on Build GRCh38
rs557013698 Chr.2: 46576928-46576928 on Build GRCh38
rs773679481 Chr.2: 46576624-46576624 on Build GRCh38
rs909786317 Chr.2: 46576864-46576864 on Build GRCh38
rs749455524 Chr.2: 46576593-46576593 on Build GRCh38
rs754094245 Chr.2: 46576704-46576704 on Build GRCh38
rs769282097 Chr.2: 46576644-46576644 on Build GRCh38
rs936095140 Chr.2: 46576884-46576884 on Build GRCh38
rs751522974 Chr.2: 46576563-46576563 on Build GRCh38

View Full Product Details