Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105269517-105270011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFBRAP1
Entrez Gene ID
9392
Gene Name
transforming growth factor beta receptor associated protein 1
Gene Aliases
TRAP-1, TRAP1, VPS3
UniGene
Hs.446350
Gene Chromosome Location
Chr.2: 105249404-105329714 on Build GRCh38
Transcripts
NM_001142621.2 NM_004257.5 XM_011512193.2 XM_017005339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019819 c.2023A>C p.K675Q Chr.2: 105269655-105269655 on Build GRCh38
4934072 c.2050C>T p.H684Y Chr.2: 105269628-105269628 on Build GRCh38
1564880 c.2008G>A p.A670T Chr.2: 105269670-105269670 on Build GRCh38
1669189 c.2128C>A p.P710T Chr.2: 105269550-105269550 on Build GRCh38
7103027 c.2145C>G p.L715L Chr.2: 105269533-105269533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374268101 Chr.2: 105269756-105269756 on Build GRCh38
rs1040822093 Chr.2: 105269975-105269975 on Build GRCh38
rs756255038 Chr.2: 105269551-105269551 on Build GRCh38
rs769114421 Chr.2: 105269685-105269685 on Build GRCh38
rs750427203 Chr.2: 105269545-105269545 on Build GRCh38
rs779014006 Chr.2: 105269657-105269657 on Build GRCh38
rs936338297 Chr.2: 105269893-105269893 on Build GRCh38
rs913398643 Chr.2: 105269849-105269849 on Build GRCh38
rs1022138131 Chr.2: 105269547-105269547 on Build GRCh38
rs535283754 Chr.2: 105269783-105269783 on Build GRCh38
rs767692666 Chr.2: 105269613-105269613 on Build GRCh38
rs771800045 Chr.2: 105269566-105269566 on Build GRCh38
rs913504497 Chr.2: 105269698-105269698 on Build GRCh38
rs199861425 Chr.2: 105269670-105269670 on Build GRCh38
rs963835212 Chr.2: 105269603-105269603 on Build GRCh38
rs899349665 Chr.2: 105269705-105269705 on Build GRCh38
rs778783019 Chr.2: 105269559-105269559 on Build GRCh38
rs2241798 Chr.2: 105269762-105269762 on Build GRCh38
rs1054892059 Chr.2: 105269750-105269750 on Build GRCh38
rs114212410 Chr.2: 105269739-105269739 on Build GRCh38
rs767978457 Chr.2: 105269702-105269702 on Build GRCh38
rs78321172 Chr.2: 105269926-105269926 on Build GRCh38
rs1054774781 Chr.2: 105269902-105269902 on Build GRCh38
rs200479465 Chr.2: 105269679-105269679 on Build GRCh38
rs557358647 Chr.2: 105269828-105269828 on Build GRCh38
rs1013393695 Chr.2: 105269544-105269544 on Build GRCh38
rs116051351 Chr.2: 105269891-105269891 on Build GRCh38
rs1029057949 Chr.2: 105269816-105269816 on Build GRCh38
rs756130720 Chr.2: 105269626-105269626 on Build GRCh38
rs760884013 Chr.2: 105269707-105269707 on Build GRCh38
rs758263311 Chr.2: 105269745-105269745 on Build GRCh38
rs779830123 Chr.2: 105269556-105269556 on Build GRCh38
rs745348624 Chr.2: 105269757-105269757 on Build GRCh38
rs979047411 Chr.2: 105269871-105269871 on Build GRCh38
rs776152944 Chr.2: 105269682-105269682 on Build GRCh38
rs974801503 Chr.2: 105269647-105269647 on Build GRCh38
rs781418446 Chr.2: 105269543-105269543 on Build GRCh38
rs777278959 Chr.2: 105269671-105269671 on Build GRCh38
rs921991624 Chr.2: 105269678-105269678 on Build GRCh38
rs746027004 Chr.2: 105269801-105269801 on Build GRCh38
rs996110479 Chr.2: 105269809-105269809 on Build GRCh38
rs144071743 Chr.2: 105269692-105269692 on Build GRCh38
rs370211705 Chr.2: 105269683-105269683 on Build GRCh38
rs900519441 Chr.2: 105269977-105269977 on Build GRCh38
rs1004798959 Chr.2: 105269763-105269763 on Build GRCh38
rs142371913 Chr.2: 105269646-105269646 on Build GRCh38
rs762193471 Chr.2: 105269601-105269601 on Build GRCh38
rs960486044 Chr.2: 105269859-105269859 on Build GRCh38
rs936290886 Chr.2: 105269748-105269748 on Build GRCh38
rs980391035 Chr.2: 105269903-105269903 on Build GRCh38
rs766523815 Chr.2: 105269709-105269709 on Build GRCh38
rs747923465 Chr.2: 105269659-105269659 on Build GRCh38
rs762140292 Chr.2: 105269696-105269696 on Build GRCh38
rs772959340 Chr.2: 105269569-105269569 on Build GRCh38
rs749301041 Chr.2: 105269557-105269557 on Build GRCh38
rs1047922182 Chr.2: 105269955-105269955 on Build GRCh38
rs375991312 Chr.2: 105269728-105269728 on Build GRCh38
rs565390607 Chr.2: 105269593-105269593 on Build GRCh38
rs1046215415 Chr.2: 105269758-105269758 on Build GRCh38
rs764601182 Chr.2: 105269594-105269594 on Build GRCh38
rs967887316 Chr.2: 105269819-105269819 on Build GRCh38
rs755138827 Chr.2: 105269730-105269730 on Build GRCh38
rs780247252 Chr.2: 105269633-105269633 on Build GRCh38
rs923656393 Chr.2: 105269585-105269585 on Build GRCh38
rs537324905 Chr.2: 105269755-105269755 on Build GRCh38
rs992282860 Chr.2: 105269878-105269878 on Build GRCh38
rs373915154 Chr.2: 105269706-105269706 on Build GRCh38
rs140106209 Chr.2: 105269589-105269589 on Build GRCh38
rs777558887 Chr.2: 105269746-105269746 on Build GRCh38
rs916250245 Chr.2: 105269885-105269885 on Build GRCh38
rs752742217 Chr.2: 105269744-105269744 on Build GRCh38
rs1024913487 Chr.2: 105269550-105269550 on Build GRCh38
rs770534140 Chr.2: 105269590-105269590 on Build GRCh38
rs754903144 Chr.2: 105269650-105269650 on Build GRCh38
rs750702972 Chr.2: 105269614-105269614 on Build GRCh38
rs944801551 Chr.2: 105269965-105269965 on Build GRCh38
rs746797490 Chr.2: 105269754-105269754 on Build GRCh38
rs916360621 Chr.2: 105269908-105269908 on Build GRCh38
rs1046698601 Chr.2: 105269979-105269979 on Build GRCh38
rs769945483 Chr.2: 105269837-105269837 on Build GRCh38
rs967913279 Chr.2: 105269866-105269866 on Build GRCh38
rs145351527 Chr.2: 105269565-105269565 on Build GRCh38
rs768567840 Chr.2: 105269558-105269558 on Build GRCh38

View Full Product Details